Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10531265-10531810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMP2
Entrez Gene ID
2013
Gene Name
epithelial membrane protein 2
Gene Aliases
XMP
UniGene
Hs.531561
Gene Chromosome Location
Chr.16: 10528422-10580716 on Build GRCh38
Transcripts
NM_001424.5 XM_006720864.3

SNP Detail

SNP ID Chromosome Location
rs538892101 Chr.16: 10531577-10531577 on Build GRCh38
rs1000191118 Chr.16: 10531410-10531410 on Build GRCh38
rs1057236222 Chr.16: 10531693-10531693 on Build GRCh38
rs185600717 Chr.16: 10531708-10531708 on Build GRCh38
rs1016321548 Chr.16: 10531649-10531649 on Build GRCh38
rs149380057 Chr.16: 10531749-10531756 on Build GRCh38
rs531622159 Chr.16: 10531402-10531402 on Build GRCh38
rs561079188 Chr.16: 10531674-10531674 on Build GRCh38
rs997758743 Chr.16: 10531576-10531576 on Build GRCh38
rs367909312 Chr.16: 10531434-10531434 on Build GRCh38
rs935458842 Chr.16: 10531553-10531553 on Build GRCh38
rs111364972 Chr.16: 10531749-10531760 on Build GRCh38
rs1043127760 Chr.16: 10531454-10531454 on Build GRCh38
rs12925190 Chr.16: 10531492-10531492 on Build GRCh38
rs947817238 Chr.16: 10531396-10531396 on Build GRCh38
rs114697308 Chr.16: 10531647-10531647 on Build GRCh38
rs995073534 Chr.16: 10531751-10531752 on Build GRCh38
rs879046931 Chr.16: 10531753-10531756 on Build GRCh38
rs938990074 Chr.16: 10531378-10531378 on Build GRCh38
rs1142436 Chr.16: 10531557-10531557 on Build GRCh38
rs770114867 Chr.16: 10531786-10531787 on Build GRCh38
rs1035903638 Chr.16: 10531334-10531334 on Build GRCh38
rs138628745 Chr.16: 10531541-10531541 on Build GRCh38
rs112700344 Chr.16: 10531503-10531503 on Build GRCh38
rs947835620 Chr.16: 10531538-10531538 on Build GRCh38
rs10459782 Chr.16: 10531318-10531318 on Build GRCh38
rs530239089 Chr.16: 10531382-10531382 on Build GRCh38
rs12924510 Chr.16: 10531401-10531401 on Build GRCh38
rs144379656 Chr.16: 10531290-10531290 on Build GRCh38
rs941311973 Chr.16: 10531330-10531330 on Build GRCh38
rs151171267 Chr.16: 10531481-10531481 on Build GRCh38
rs912356263 Chr.16: 10531540-10531540 on Build GRCh38
rs540372726 Chr.16: 10531707-10531707 on Build GRCh38
rs916240763 Chr.16: 10531535-10531535 on Build GRCh38
rs879942799 Chr.16: 10531422-10531422 on Build GRCh38
rs151287012 Chr.16: 10531749-10531752 on Build GRCh38
rs371172581 Chr.16: 10531367-10531367 on Build GRCh38
rs563215996 Chr.16: 10531558-10531559 on Build GRCh38
rs1138755 Chr.16: 10531388-10531388 on Build GRCh38
rs918260237 Chr.16: 10531358-10531358 on Build GRCh38
rs12929194 Chr.16: 10531570-10531570 on Build GRCh38
rs552639369 Chr.16: 10531459-10531459 on Build GRCh38
rs779537352 Chr.16: 10531301-10531301 on Build GRCh38
rs992034910 Chr.16: 10531775-10531775 on Build GRCh38
rs12924690 Chr.16: 10531519-10531519 on Build GRCh38
rs1015647904 Chr.16: 10531361-10531361 on Build GRCh38
rs367727964 Chr.16: 10531348-10531348 on Build GRCh38
rs1001645021 Chr.16: 10531479-10531479 on Build GRCh38
rs752713386 Chr.16: 10531703-10531703 on Build GRCh38
rs962541448 Chr.16: 10531498-10531498 on Build GRCh38
rs983751053 Chr.16: 10531345-10531345 on Build GRCh38
rs12928798 Chr.16: 10531317-10531317 on Build GRCh38
rs1026645686 Chr.16: 10531522-10531522 on Build GRCh38
rs12929088 Chr.16: 10531526-10531526 on Build GRCh38
rs905894069 Chr.16: 10531651-10531651 on Build GRCh38
rs532439130 Chr.16: 10531709-10531709 on Build GRCh38
rs947327471 Chr.16: 10531603-10531603 on Build GRCh38
rs760768066 Chr.16: 10531676-10531677 on Build GRCh38
rs972585190 Chr.16: 10531331-10531331 on Build GRCh38
rs759869089 Chr.16: 10531628-10531628 on Build GRCh38
rs953619089 Chr.16: 10531488-10531488 on Build GRCh38
rs781308866 Chr.16: 10531756-10531757 on Build GRCh38
rs1017181498 Chr.16: 10531496-10531496 on Build GRCh38
rs973755327 Chr.16: 10531381-10531381 on Build GRCh38
rs953467957 Chr.16: 10531308-10531308 on Build GRCh38
rs141178854 Chr.16: 10531487-10531487 on Build GRCh38
rs913158894 Chr.16: 10531594-10531594 on Build GRCh38
rs976174367 Chr.16: 10531292-10531292 on Build GRCh38
rs985693798 Chr.16: 10531392-10531392 on Build GRCh38
rs960552631 Chr.16: 10531335-10531339 on Build GRCh38
rs1056159791 Chr.16: 10531387-10531387 on Build GRCh38
rs879124452 Chr.16: 10531753-10531760 on Build GRCh38
rs965532142 Chr.16: 10531573-10531573 on Build GRCh38
rs762735861 Chr.16: 10531784-10531790 on Build GRCh38
rs574644503 Chr.16: 10531701-10531701 on Build GRCh38
rs991817092 Chr.16: 10531531-10531531 on Build GRCh38
rs1005060961 Chr.16: 10531706-10531707 on Build GRCh38
rs916235377 Chr.16: 10531786-10531786 on Build GRCh38
rs760451570 Chr.16: 10531615-10531615 on Build GRCh38
rs925454036 Chr.16: 10531545-10531545 on Build GRCh38

View Full Product Details