Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1828351-1828850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAHD1
Entrez Gene ID
81889
Gene Name
fumarylacetoacetate hydrolase domain containing 1
Gene Aliases
C16orf36, YISKL
UniGene
Hs.513265
Gene Chromosome Location
Chr.16: 1827224-1840207 on Build GRCh38
Transcripts
NM_001018104.2 NM_001142398.1 NM_031208.3

SNP Detail

SNP ID Chromosome Location
rs1065583 Chr.16: 1828491-1828491 on Build GRCh38
rs539343917 Chr.16: 1828591-1828591 on Build GRCh38
rs544636006 Chr.16: 1828643-1828643 on Build GRCh38
rs1046433153 Chr.16: 1828521-1828521 on Build GRCh38
rs1065584 Chr.16: 1828520-1828520 on Build GRCh38
rs1032395520 Chr.16: 1828572-1828572 on Build GRCh38
rs989744001 Chr.16: 1828774-1828774 on Build GRCh38
rs540914406 Chr.16: 1828764-1828764 on Build GRCh38
rs928487267 Chr.16: 1828590-1828590 on Build GRCh38
rs148196681 Chr.16: 1828456-1828456 on Build GRCh38
rs1004257844 Chr.16: 1828464-1828464 on Build GRCh38
rs993237890 Chr.16: 1828409-1828409 on Build GRCh38
rs1065589 Chr.16: 1828800-1828800 on Build GRCh38
rs897456780 Chr.16: 1828652-1828652 on Build GRCh38
rs772190781 Chr.16: 1828655-1828655 on Build GRCh38
rs564470414 Chr.16: 1828653-1828653 on Build GRCh38
rs572976724 Chr.16: 1828597-1828597 on Build GRCh38
rs1065585 Chr.16: 1828596-1828596 on Build GRCh38
rs1065586 Chr.16: 1828640-1828640 on Build GRCh38
rs954819744 Chr.16: 1828440-1828440 on Build GRCh38
rs1008163743 Chr.16: 1828733-1828733 on Build GRCh38
rs890647068 Chr.16: 1828709-1828709 on Build GRCh38
rs1000004031 Chr.16: 1828829-1828829 on Build GRCh38
rs1065587 Chr.16: 1828739-1828739 on Build GRCh38
rs913904626 Chr.16: 1828495-1828495 on Build GRCh38
rs980856959 Chr.16: 1828539-1828539 on Build GRCh38
rs958041158 Chr.16: 1828584-1828584 on Build GRCh38
rs1030512643 Chr.16: 1828434-1828434 on Build GRCh38
rs11558503 Chr.16: 1828457-1828457 on Build GRCh38
rs895767465 Chr.16: 1828766-1828766 on Build GRCh38
rs1014117744 Chr.16: 1828502-1828502 on Build GRCh38
rs762822790 Chr.16: 1828753-1828753 on Build GRCh38
rs969501535 Chr.16: 1828535-1828535 on Build GRCh38
rs144837050 Chr.16: 1828442-1828442 on Build GRCh38
rs1048093299 Chr.16: 1828402-1828405 on Build GRCh38
rs765298470 Chr.16: 1828700-1828700 on Build GRCh38
rs775750222 Chr.16: 1828820-1828820 on Build GRCh38
rs1020911586 Chr.16: 1828797-1828797 on Build GRCh38
rs894412867 Chr.16: 1828626-1828626 on Build GRCh38
rs115144764 Chr.16: 1828816-1828816 on Build GRCh38
rs542831868 Chr.16: 1828610-1828610 on Build GRCh38
rs763585750 Chr.16: 1828788-1828788 on Build GRCh38
rs555784251 Chr.16: 1828636-1828636 on Build GRCh38

View Full Product Details