Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136231137-136231655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTFR2
Entrez Gene ID
113115
Gene Name
mitochondrial fission regulator 2
Gene Aliases
DUFD1, FAM54A
UniGene
Hs.121536
Gene Chromosome Location
Chr.6: 136231030-136253659 on Build GRCh38
Transcripts
NM_001099286.2 NM_001318738.1 NM_138419.4 XM_011535410.2 XM_011535412.2 XM_011535413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377800 c.1108G>A p.A370T Chr.6: 136231325-136231325 on Build GRCh38
5466677 c.1087G>T p.E363* Chr.6: 136231346-136231346 on Build GRCh38
3620928 c.1133C>T p.T378I Chr.6: 136231300-136231300 on Build GRCh38
5466676 c.1090G>T p.E364* Chr.6: 136231343-136231343 on Build GRCh38
4459211 c.1116C>T p.D372D Chr.6: 136231317-136231317 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs895203515 Chr.6: 136231551-136231551 on Build GRCh38
rs775348671 Chr.6: 136231255-136231255 on Build GRCh38
rs563015574 Chr.6: 136231357-136231357 on Build GRCh38
rs968867469 Chr.6: 136231197-136231241 on Build GRCh38
rs936103462 Chr.6: 136231378-136231378 on Build GRCh38
rs909610974 Chr.6: 136231212-136231212 on Build GRCh38
rs978029219 Chr.6: 136231268-136231268 on Build GRCh38
rs1032241942 Chr.6: 136231577-136231577 on Build GRCh38
rs143750902 Chr.6: 136231503-136231503 on Build GRCh38
rs765307774 Chr.6: 136231438-136231439 on Build GRCh38
rs760006454 Chr.6: 136231297-136231297 on Build GRCh38
rs1013978739 Chr.6: 136231221-136231221 on Build GRCh38
rs962445181 Chr.6: 136231227-136231227 on Build GRCh38
rs553490938 Chr.6: 136231234-136231234 on Build GRCh38
rs375131149 Chr.6: 136231262-136231262 on Build GRCh38
rs962436221 Chr.6: 136231630-136231630 on Build GRCh38
rs79645194 Chr.6: 136231354-136231354 on Build GRCh38
rs766250002 Chr.6: 136231431-136231432 on Build GRCh38
rs759795777 Chr.6: 136231432-136231433 on Build GRCh38
rs778594457 Chr.6: 136231502-136231502 on Build GRCh38
rs753706702 Chr.6: 136231337-136231337 on Build GRCh38
rs921655148 Chr.6: 136231331-136231331 on Build GRCh38
rs1032209425 Chr.6: 136231291-136231291 on Build GRCh38
rs760723145 Chr.6: 136231330-136231330 on Build GRCh38
rs898692175 Chr.6: 136231421-136231421 on Build GRCh38
rs113405027 Chr.6: 136231566-136231566 on Build GRCh38
rs369891404 Chr.6: 136231417-136231417 on Build GRCh38
rs867875886 Chr.6: 136231301-136231302 on Build GRCh38
rs112930946 Chr.6: 136231443-136231444 on Build GRCh38
rs765994353 Chr.6: 136231592-136231592 on Build GRCh38
rs539842885 Chr.6: 136231173-136231173 on Build GRCh38
rs767629082 Chr.6: 136231309-136231309 on Build GRCh38
rs780855809 Chr.6: 136231386-136231386 on Build GRCh38
rs987508763 Chr.6: 136231364-136231364 on Build GRCh38
rs181684816 Chr.6: 136231365-136231365 on Build GRCh38
rs143974258 Chr.6: 136231355-136231355 on Build GRCh38
rs764183981 Chr.6: 136231333-136231333 on Build GRCh38
rs919474868 Chr.6: 136231285-136231285 on Build GRCh38
rs956143296 Chr.6: 136231160-136231160 on Build GRCh38
rs763404708 Chr.6: 136231267-136231267 on Build GRCh38
rs746679262 Chr.6: 136231368-136231368 on Build GRCh38
rs761456823 Chr.6: 136231491-136231491 on Build GRCh38
rs371998160 Chr.6: 136231246-136231246 on Build GRCh38
rs957874057 Chr.6: 136231603-136231604 on Build GRCh38
rs114818440 Chr.6: 136231565-136231565 on Build GRCh38
rs771610930 Chr.6: 136231243-136231243 on Build GRCh38
rs773238035 Chr.6: 136231367-136231367 on Build GRCh38
rs889717874 Chr.6: 136231568-136231568 on Build GRCh38
rs775734522 Chr.6: 136231176-136231176 on Build GRCh38
rs371740838 Chr.6: 136231384-136231384 on Build GRCh38
rs891650232 Chr.6: 136231475-136231475 on Build GRCh38
rs895170959 Chr.6: 136231209-136231209 on Build GRCh38
rs199873104 Chr.6: 136231294-136231294 on Build GRCh38
rs953764633 Chr.6: 136231190-136231190 on Build GRCh38
rs931460279 Chr.6: 136231460-136231460 on Build GRCh38
rs1025432928 Chr.6: 136231564-136231564 on Build GRCh38
rs760582976 Chr.6: 136231432-136231432 on Build GRCh38
rs772063364 Chr.6: 136231247-136231247 on Build GRCh38
rs778575597 Chr.6: 136231230-136231230 on Build GRCh38
rs370674781 Chr.6: 136231258-136231258 on Build GRCh38
rs545444672 Chr.6: 136231263-136231263 on Build GRCh38
rs745555176 Chr.6: 136231567-136231567 on Build GRCh38
rs1013588694 Chr.6: 136231557-136231557 on Build GRCh38
rs752958388 Chr.6: 136231319-136231319 on Build GRCh38
rs974512974 Chr.6: 136231311-136231311 on Build GRCh38
rs115367796 Chr.6: 136231415-136231415 on Build GRCh38
rs771332998 Chr.6: 136231408-136231408 on Build GRCh38
rs1006471110 Chr.6: 136231573-136231573 on Build GRCh38
rs985104166 Chr.6: 136231207-136231207 on Build GRCh38
rs367584878 Chr.6: 136231296-136231296 on Build GRCh38
rs746483201 Chr.6: 136231256-136231256 on Build GRCh38
rs546685563 Chr.6: 136231393-136231393 on Build GRCh38
rs35740558 Chr.6: 136231412-136231412 on Build GRCh38

View Full Product Details