Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128677527-128677856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM71F2
Entrez Gene ID
346653
Gene Name
family with sequence similarity 71 member F2
Gene Aliases
FAM137B
UniGene
Hs.445236
Gene Chromosome Location
Chr.7: 128671628-128702262 on Build GRCh38
Transcripts
NM_001012454.5 NM_001128926.3 NM_001290254.1 NM_001290255.1 XM_011516128.2 XM_011516129.2 XM_011516130.2 XM_011516131.2 XM_011516132.2 XM_011516134.2 XM_011516135.2 XM_011516136.2 XM_017012147.1 XM_017012149.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1312660 c.588G>A p.K196K Chr.7: 128677786-128677786 on Build GRCh38
5494874 c.448A>T p.I150F Chr.7: 128677646-128677646 on Build GRCh38
3949915 c.522G>T p.L174L Chr.7: 128677720-128677720 on Build GRCh38
744643 c.588G>T p.K196N Chr.7: 128677786-128677786 on Build GRCh38
4004288 c.406C>T p.R136W Chr.7: 128677604-128677604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549567409 Chr.7: 128677651-128677651 on Build GRCh38
rs201240369 Chr.7: 128677582-128677582 on Build GRCh38
rs766449722 Chr.7: 128677752-128677752 on Build GRCh38
rs754104577 Chr.7: 128677753-128677753 on Build GRCh38
rs554571321 Chr.7: 128677754-128677754 on Build GRCh38
rs776255301 Chr.7: 128677613-128677613 on Build GRCh38
rs553880583 Chr.7: 128677667-128677667 on Build GRCh38
rs752098817 Chr.7: 128677618-128677618 on Build GRCh38
rs764712714 Chr.7: 128677548-128677548 on Build GRCh38
rs748624402 Chr.7: 128677767-128677767 on Build GRCh38
rs538444441 Chr.7: 128677663-128677663 on Build GRCh38
rs757102292 Chr.7: 128677648-128677648 on Build GRCh38
rs749783143 Chr.7: 128677593-128677593 on Build GRCh38
rs992589481 Chr.7: 128677660-128677660 on Build GRCh38
rs745551542 Chr.7: 128677575-128677575 on Build GRCh38
rs531576408 Chr.7: 128677614-128677614 on Build GRCh38
rs772423422 Chr.7: 128677682-128677682 on Build GRCh38
rs377622805 Chr.7: 128677771-128677771 on Build GRCh38
rs966463057 Chr.7: 128677814-128677815 on Build GRCh38
rs774424019 Chr.7: 128677726-128677726 on Build GRCh38
rs199592994 Chr.7: 128677728-128677728 on Build GRCh38
rs780040728 Chr.7: 128677586-128677586 on Build GRCh38
rs773614211 Chr.7: 128677685-128677685 on Build GRCh38
rs976543004 Chr.7: 128677820-128677820 on Build GRCh38
rs7797745 Chr.7: 128677828-128677828 on Build GRCh38
rs771339781 Chr.7: 128677816-128677816 on Build GRCh38
rs181109862 Chr.7: 128677599-128677599 on Build GRCh38
rs79425518 Chr.7: 128677772-128677772 on Build GRCh38
rs769910752 Chr.7: 128677583-128677583 on Build GRCh38
rs914063241 Chr.7: 128677773-128677773 on Build GRCh38
rs945491450 Chr.7: 128677784-128677784 on Build GRCh38
rs750138953 Chr.7: 128677735-128677735 on Build GRCh38
rs764781019 Chr.7: 128677616-128677616 on Build GRCh38
rs751358688 Chr.7: 128677573-128677573 on Build GRCh38
rs747269571 Chr.7: 128677798-128677798 on Build GRCh38
rs757118745 Chr.7: 128677578-128677578 on Build GRCh38
rs201533178 Chr.7: 128677743-128677743 on Build GRCh38
rs892515978 Chr.7: 128677554-128677554 on Build GRCh38
rs913661005 Chr.7: 128677563-128677563 on Build GRCh38
rs200938500 Chr.7: 128677640-128677640 on Build GRCh38
rs762556424 Chr.7: 128677635-128677635 on Build GRCh38
rs1014236879 Chr.7: 128677645-128677645 on Build GRCh38
rs1020657862 Chr.7: 128677775-128677775 on Build GRCh38
rs779032793 Chr.7: 128677761-128677761 on Build GRCh38
rs969739917 Chr.7: 128677736-128677736 on Build GRCh38
rs778282553 Chr.7: 128677797-128677797 on Build GRCh38
rs571067980 Chr.7: 128677662-128677662 on Build GRCh38
rs763750831 Chr.7: 128677717-128677717 on Build GRCh38
rs749032788 Chr.7: 128677831-128677831 on Build GRCh38
rs6467210 Chr.7: 128677604-128677604 on Build GRCh38
rs138918252 Chr.7: 128677639-128677639 on Build GRCh38
rs769291032 Chr.7: 128677694-128677694 on Build GRCh38
rs777788075 Chr.7: 128677570-128677570 on Build GRCh38
rs762308334 Chr.7: 128677708-128677708 on Build GRCh38
rs186458943 Chr.7: 128677605-128677605 on Build GRCh38
rs377562102 Chr.7: 128677704-128677704 on Build GRCh38
rs768787544 Chr.7: 128677666-128677666 on Build GRCh38
rs749291100 Chr.7: 128677655-128677655 on Build GRCh38
rs752647419 Chr.7: 128677552-128677552 on Build GRCh38
rs748714129 Chr.7: 128677672-128677672 on Build GRCh38
rs761122140 Chr.7: 128677690-128677690 on Build GRCh38
rs1052231589 Chr.7: 128677592-128677592 on Build GRCh38
rs781197315 Chr.7: 128677650-128677650 on Build GRCh38
rs372599605 Chr.7: 128677653-128677653 on Build GRCh38
rs758314723 Chr.7: 128677556-128677556 on Build GRCh38
rs1046174394 Chr.7: 128677646-128677646 on Build GRCh38

View Full Product Details