Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45233964-45234382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL1
Entrez Gene ID
752
Gene Name
formin like 1
Gene Aliases
C17orf1, C17orf1B, FHOD4, FMNL, KW-13
UniGene
Hs.100217
Gene Chromosome Location
Chr.17: 45221728-45247322 on Build GRCh38
Transcripts
NM_005892.3 XM_006722062.3 XM_006722063.3 XM_006722064.3 XM_006722065.3 XM_006722066.3 XM_006722069.2 XM_006722070.3 XM_011525179.2 XM_011525180.2 XM_011525182.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6665508 c.585C>T p.S195S Chr.17: 45234171-45234171 on Build GRCh38
4551366 c.526G>C p.E176Q Chr.17: 45234112-45234112 on Build GRCh38
1734486 c.586G>A p.V196M Chr.17: 45234172-45234172 on Build GRCh38
4924361 c.584C>A p.S195Y Chr.17: 45234170-45234170 on Build GRCh38
1207292 c.614+1G>A p.? Chr.17: 45234201-45234201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552137406 Chr.17: 45234331-45234331 on Build GRCh38
rs927565227 Chr.17: 45234054-45234054 on Build GRCh38
rs765181550 Chr.17: 45234235-45234235 on Build GRCh38
rs777337969 Chr.17: 45234184-45234184 on Build GRCh38
rs758324241 Chr.17: 45234338-45234338 on Build GRCh38
rs369158547 Chr.17: 45234195-45234195 on Build GRCh38
rs142930756 Chr.17: 45234072-45234072 on Build GRCh38
rs917533888 Chr.17: 45234323-45234323 on Build GRCh38
rs748591833 Chr.17: 45234288-45234288 on Build GRCh38
rs775915127 Chr.17: 45234081-45234081 on Build GRCh38
rs188584942 Chr.17: 45234252-45234252 on Build GRCh38
rs778765351 Chr.17: 45234172-45234172 on Build GRCh38
rs756207299 Chr.17: 45234153-45234153 on Build GRCh38
rs762110167 Chr.17: 45234135-45234135 on Build GRCh38
rs776749286 Chr.17: 45234311-45234311 on Build GRCh38
rs963498619 Chr.17: 45234260-45234260 on Build GRCh38
rs753932162 Chr.17: 45234226-45234226 on Build GRCh38
rs760976906 Chr.17: 45234220-45234220 on Build GRCh38
rs773681950 Chr.17: 45234298-45234298 on Build GRCh38
rs768010499 Chr.17: 45234214-45234214 on Build GRCh38
rs777529271 Chr.17: 45234248-45234248 on Build GRCh38
rs372127527 Chr.17: 45234089-45234089 on Build GRCh38
rs200640751 Chr.17: 45234083-45234083 on Build GRCh38
rs750681404 Chr.17: 45234147-45234147 on Build GRCh38
rs61742620 Chr.17: 45234052-45234052 on Build GRCh38
rs745347714 Chr.17: 45234268-45234268 on Build GRCh38
rs754038484 Chr.17: 45234163-45234163 on Build GRCh38
rs1042358370 Chr.17: 45234356-45234356 on Build GRCh38
rs374780853 Chr.17: 45234032-45234032 on Build GRCh38
rs12944148 Chr.17: 45234287-45234287 on Build GRCh38
rs1001625522 Chr.17: 45233990-45233990 on Build GRCh38
rs202135542 Chr.17: 45234215-45234215 on Build GRCh38
rs973588803 Chr.17: 45234278-45234279 on Build GRCh38
rs923075102 Chr.17: 45234165-45234165 on Build GRCh38
rs964396284 Chr.17: 45234126-45234126 on Build GRCh38
rs749253370 Chr.17: 45234021-45234021 on Build GRCh38
rs771793020 Chr.17: 45234179-45234179 on Build GRCh38
rs771623262 Chr.17: 45234263-45234263 on Build GRCh38
rs745701308 Chr.17: 45234178-45234178 on Build GRCh38
rs559226610 Chr.17: 45234223-45234223 on Build GRCh38
rs530551198 Chr.17: 45234278-45234278 on Build GRCh38
rs369927345 Chr.17: 45234097-45234097 on Build GRCh38
rs767742597 Chr.17: 45234146-45234146 on Build GRCh38
rs375374786 Chr.17: 45234091-45234091 on Build GRCh38
rs9898221 Chr.17: 45234253-45234253 on Build GRCh38
rs770429242 Chr.17: 45234080-45234080 on Build GRCh38
rs1031970859 Chr.17: 45234125-45234125 on Build GRCh38
rs937586270 Chr.17: 45234283-45234283 on Build GRCh38
rs747927505 Chr.17: 45234174-45234174 on Build GRCh38
rs966340815 Chr.17: 45234078-45234078 on Build GRCh38
rs761172187 Chr.17: 45234300-45234300 on Build GRCh38
rs769161697 Chr.17: 45234199-45234199 on Build GRCh38
rs747933926 Chr.17: 45234044-45234044 on Build GRCh38
rs778831957 Chr.17: 45234092-45234094 on Build GRCh38
rs1041781192 Chr.17: 45234341-45234341 on Build GRCh38
rs376075736 Chr.17: 45234243-45234243 on Build GRCh38
rs896002266 Chr.17: 45234003-45234003 on Build GRCh38
rs780849007 Chr.17: 45234259-45234259 on Build GRCh38
rs373132848 Chr.17: 45234185-45234185 on Build GRCh38
rs778766041 Chr.17: 45234037-45234037 on Build GRCh38
rs780269020 Chr.17: 45234158-45234158 on Build GRCh38
rs1037668783 Chr.17: 45234222-45234222 on Build GRCh38
rs141085778 Chr.17: 45234171-45234171 on Build GRCh38
rs143814916 Chr.17: 45234337-45234337 on Build GRCh38
rs373352953 Chr.17: 45234208-45234208 on Build GRCh38
rs772775462 Chr.17: 45234296-45234296 on Build GRCh38
rs569386802 Chr.17: 45234262-45234262 on Build GRCh38
rs774756543 Chr.17: 45234205-45234205 on Build GRCh38
rs772872778 Chr.17: 45234056-45234056 on Build GRCh38
rs759591900 Chr.17: 45234327-45234327 on Build GRCh38
rs766659793 Chr.17: 45234309-45234309 on Build GRCh38
rs774866768 Chr.17: 45234279-45234279 on Build GRCh38
rs755034225 Chr.17: 45234231-45234231 on Build GRCh38
rs572623597 Chr.17: 45234001-45234001 on Build GRCh38
rs7209538 Chr.17: 45234096-45234096 on Build GRCh38
rs974533292 Chr.17: 45234160-45234160 on Build GRCh38
rs370344494 Chr.17: 45234247-45234247 on Build GRCh38

View Full Product Details