Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 71758006-71758533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GC
Entrez Gene ID
2638
Gene Name
GC, vitamin D binding protein
Gene Aliases
DBP, DBP/GC, GRD3, Gc-MAF, GcMAF, HEL-S-51, VDBG, VDBP
UniGene
Hs.418497
Gene Chromosome Location
Chr.4: 71741693-71805520 on Build GRCh38
Transcripts
NM_000583.3 NM_001204306.1 NM_001204307.1 XM_006714177.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6901101 c.879G>T p.M293I Chr.4: 71758051-71758051 on Build GRCh38
4392731 c.723A>G p.Q241Q Chr.4: 71758150-71758150 on Build GRCh38
1430856 c.761T>A p.L254Q Chr.4: 71758112-71758112 on Build GRCh38
7192338 c.831+6A>G p.? Chr.4: 71758036-71758036 on Build GRCh38
5457132 c.787C>T p.P263S Chr.4: 71758143-71758143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575990488 Chr.4: 71758378-71758378 on Build GRCh38
rs535186997 Chr.4: 71758309-71758309 on Build GRCh38
rs113588583 Chr.4: 71758174-71758174 on Build GRCh38
rs566149557 Chr.4: 71758237-71758237 on Build GRCh38
rs866493183 Chr.4: 71758346-71758346 on Build GRCh38
rs769811509 Chr.4: 71758163-71758163 on Build GRCh38
rs915432644 Chr.4: 71758438-71758438 on Build GRCh38
rs113964594 Chr.4: 71758041-71758041 on Build GRCh38
rs749016057 Chr.4: 71758191-71758191 on Build GRCh38
rs767191138 Chr.4: 71758210-71758210 on Build GRCh38
rs762031035 Chr.4: 71758508-71758508 on Build GRCh38
rs558191611 Chr.4: 71758328-71758328 on Build GRCh38
rs1048476781 Chr.4: 71758279-71758279 on Build GRCh38
rs747394970 Chr.4: 71758209-71758209 on Build GRCh38
rs761456392 Chr.4: 71758062-71758062 on Build GRCh38
rs768938415 Chr.4: 71758176-71758176 on Build GRCh38
rs143184570 Chr.4: 71758122-71758122 on Build GRCh38
rs890285416 Chr.4: 71758240-71758240 on Build GRCh38
rs371686563 Chr.4: 71758035-71758035 on Build GRCh38
rs749933780 Chr.4: 71758068-71758068 on Build GRCh38
rs540700701 Chr.4: 71758505-71758505 on Build GRCh38
rs530881904 Chr.4: 71758209-71758209 on Build GRCh38
rs930028567 Chr.4: 71758258-71758258 on Build GRCh38
rs745927733 Chr.4: 71758159-71758159 on Build GRCh38
rs1032753124 Chr.4: 71758403-71758403 on Build GRCh38
rs921590889 Chr.4: 71758449-71758449 on Build GRCh38
rs749825631 Chr.4: 71758172-71758172 on Build GRCh38
rs760212860 Chr.4: 71758223-71758223 on Build GRCh38
rs112895006 Chr.4: 71758212-71758212 on Build GRCh38
rs903352043 Chr.4: 71758382-71758382 on Build GRCh38
rs565721085 Chr.4: 71758296-71758296 on Build GRCh38
rs1042721874 Chr.4: 71758232-71758232 on Build GRCh38
rs776658446 Chr.4: 71758499-71758499 on Build GRCh38
rs746711586 Chr.4: 71758409-71758409 on Build GRCh38
rs113444205 Chr.4: 71758238-71758238 on Build GRCh38
rs374914389 Chr.4: 71758048-71758048 on Build GRCh38
rs909839961 Chr.4: 71758285-71758285 on Build GRCh38
rs781329202 Chr.4: 71758146-71758146 on Build GRCh38
rs770672042 Chr.4: 71758192-71758192 on Build GRCh38
rs1018596545 Chr.4: 71758448-71758448 on Build GRCh38
rs557739883 Chr.4: 71758312-71758312 on Build GRCh38
rs776015810 Chr.4: 71758169-71758169 on Build GRCh38
rs773949276 Chr.4: 71758060-71758060 on Build GRCh38
rs182805109 Chr.4: 71758112-71758112 on Build GRCh38
rs547640798 Chr.4: 71758259-71758259 on Build GRCh38
rs953054775 Chr.4: 71758190-71758190 on Build GRCh38
rs766312099 Chr.4: 71758084-71758084 on Build GRCh38
rs10488854 Chr.4: 71758418-71758418 on Build GRCh38
rs757515784 Chr.4: 71758142-71758142 on Build GRCh38
rs369732946 Chr.4: 71758228-71758228 on Build GRCh38
rs745518127 Chr.4: 71758101-71758101 on Build GRCh38
rs755649458 Chr.4: 71758071-71758071 on Build GRCh38
rs771823776 Chr.4: 71758125-71758125 on Build GRCh38
rs766876824 Chr.4: 71758067-71758067 on Build GRCh38
rs536606734 Chr.4: 71758394-71758394 on Build GRCh38
rs746304661 Chr.4: 71758037-71758037 on Build GRCh38
rs369308032 Chr.4: 71758188-71758188 on Build GRCh38
rs969447990 Chr.4: 71758313-71758313 on Build GRCh38
rs1004682843 Chr.4: 71758270-71758270 on Build GRCh38
rs911615516 Chr.4: 71758506-71758506 on Build GRCh38
rs529043158 Chr.4: 71758134-71758136 on Build GRCh38
rs763281635 Chr.4: 71758049-71758049 on Build GRCh38
rs747162006 Chr.4: 71758137-71758137 on Build GRCh38
rs573445895 Chr.4: 71758469-71758469 on Build GRCh38
rs967501167 Chr.4: 71758036-71758036 on Build GRCh38
rs928114730 Chr.4: 71758063-71758063 on Build GRCh38
rs369309193 Chr.4: 71758034-71758034 on Build GRCh38
rs1007162743 Chr.4: 71758419-71758419 on Build GRCh38
rs778664416 Chr.4: 71758201-71758202 on Build GRCh38
rs112922527 Chr.4: 71758229-71758229 on Build GRCh38
rs770328076 Chr.4: 71758042-71758042 on Build GRCh38
rs1029754492 Chr.4: 71758133-71758134 on Build GRCh38
rs764464348 Chr.4: 71758055-71758055 on Build GRCh38

View Full Product Details