Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 230907928-230908478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR55
Entrez Gene ID
9290
Gene Name
G protein-coupled receptor 55
Gene Aliases
LPIR1
UniGene
Hs.114545
Gene Chromosome Location
Chr.2: 230907318-230961269 on Build GRCh38
Transcripts
NM_005683.3 XM_005246952.3 XM_011512175.2 XM_011512176.2

SNP Detail

SNP ID Chromosome Location
rs937532601 Chr.2: 230908066-230908066 on Build GRCh38
rs762704732 Chr.2: 230908055-230908056 on Build GRCh38
rs1010340068 Chr.2: 230908203-230908203 on Build GRCh38
rs765897643 Chr.2: 230908075-230908075 on Build GRCh38
rs548138548 Chr.2: 230908453-230908453 on Build GRCh38
rs1030809515 Chr.2: 230908091-230908091 on Build GRCh38
rs751799549 Chr.2: 230908353-230908353 on Build GRCh38
rs927571751 Chr.2: 230908278-230908278 on Build GRCh38
rs971386696 Chr.2: 230908426-230908426 on Build GRCh38
rs760038173 Chr.2: 230907963-230907963 on Build GRCh38
rs997374048 Chr.2: 230908145-230908145 on Build GRCh38
rs758199258 Chr.2: 230908294-230908294 on Build GRCh38
rs954802013 Chr.2: 230908375-230908375 on Build GRCh38
rs750060140 Chr.2: 230908220-230908224 on Build GRCh38
rs1034351226 Chr.2: 230908443-230908443 on Build GRCh38
rs570646845 Chr.2: 230908000-230908000 on Build GRCh38
rs1020406277 Chr.2: 230908413-230908413 on Build GRCh38
rs1002911630 Chr.2: 230908442-230908442 on Build GRCh38
rs1013370220 Chr.2: 230908333-230908333 on Build GRCh38
rs73992968 Chr.2: 230908190-230908190 on Build GRCh38
rs184053115 Chr.2: 230907955-230907955 on Build GRCh38
rs541853658 Chr.2: 230908239-230908239 on Build GRCh38
rs957645782 Chr.2: 230908202-230908202 on Build GRCh38
rs566666602 Chr.2: 230907982-230907982 on Build GRCh38
rs751428261 Chr.2: 230908213-230908213 on Build GRCh38
rs1013419192 Chr.2: 230908116-230908116 on Build GRCh38
rs957385762 Chr.2: 230907974-230907974 on Build GRCh38
rs574326884 Chr.2: 230908192-230908192 on Build GRCh38
rs901527861 Chr.2: 230908138-230908138 on Build GRCh38
rs537714669 Chr.2: 230908052-230908052 on Build GRCh38
rs572128792 Chr.2: 230908281-230908281 on Build GRCh38
rs1035961331 Chr.2: 230908134-230908134 on Build GRCh38
rs962361711 Chr.2: 230908358-230908358 on Build GRCh38
rs559579119 Chr.2: 230908391-230908391 on Build GRCh38
rs34670968 Chr.2: 230908058-230908059 on Build GRCh38
rs150432172 Chr.2: 230908305-230908305 on Build GRCh38
rs72539509 Chr.2: 230908056-230908057 on Build GRCh38
rs889407347 Chr.2: 230908016-230908016 on Build GRCh38
rs753310353 Chr.2: 230908056-230908056 on Build GRCh38
rs1001140543 Chr.2: 230908177-230908177 on Build GRCh38
rs999693712 Chr.2: 230908063-230908063 on Build GRCh38
rs947648255 Chr.2: 230908250-230908250 on Build GRCh38
rs908919004 Chr.2: 230908111-230908111 on Build GRCh38
rs553892992 Chr.2: 230908280-230908280 on Build GRCh38
rs909232789 Chr.2: 230908153-230908153 on Build GRCh38
rs1023499985 Chr.2: 230908130-230908130 on Build GRCh38
rs888951702 Chr.2: 230908165-230908165 on Build GRCh38
rs1050326718 Chr.2: 230908154-230908154 on Build GRCh38
rs146670040 Chr.2: 230908178-230908179 on Build GRCh38
rs200477178 Chr.2: 230908056-230908056 on Build GRCh38
rs765190646 Chr.2: 230908271-230908271 on Build GRCh38
rs1023447537 Chr.2: 230908347-230908347 on Build GRCh38
rs910791548 Chr.2: 230908228-230908228 on Build GRCh38
rs563242790 Chr.2: 230908386-230908386 on Build GRCh38
rs892313046 Chr.2: 230908387-230908387 on Build GRCh38
rs534038759 Chr.2: 230907993-230907993 on Build GRCh38
rs558283223 Chr.2: 230907996-230907996 on Build GRCh38
rs574186854 Chr.2: 230908284-230908284 on Build GRCh38
rs58928368 Chr.2: 230908374-230908374 on Build GRCh38
rs76190894 Chr.2: 230908356-230908356 on Build GRCh38
rs11414001 Chr.2: 230908057-230908058 on Build GRCh38
rs905104623 Chr.2: 230908352-230908352 on Build GRCh38
rs981674116 Chr.2: 230908276-230908276 on Build GRCh38
rs1006561650 Chr.2: 230908046-230908046 on Build GRCh38
rs888997912 Chr.2: 230908357-230908357 on Build GRCh38
rs971417692 Chr.2: 230908240-230908240 on Build GRCh38
rs913239034 Chr.2: 230907995-230907995 on Build GRCh38
rs940452851 Chr.2: 230908113-230908113 on Build GRCh38
rs752616165 Chr.2: 230908385-230908385 on Build GRCh38
rs777787604 Chr.2: 230908312-230908312 on Build GRCh38
rs922921959 Chr.2: 230908314-230908314 on Build GRCh38

View Full Product Details