Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73252366-73252869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC9-AS1
Entrez Gene ID
414245
Gene Name
DNAJC9 antisense RNA 1
Gene Aliases
C10orf103, bA537A6.3
UniGene
Hs.568860
Gene Chromosome Location
Chr.10: 73247367-73276984 on Build GRCh38
Transcripts
NR_038373.1 NR_134458.1 NR_134459.1 NR_134460.1 NM_016065.3 XM_017016305.1

Gene Symbol
MRPS16
Entrez Gene ID
51021
Gene Name
mitochondrial ribosomal protein S16
Gene Aliases
CGI-132, COXPD2, MRP-S16, RPMS16
UniGene
Hs.180312
Gene Chromosome Location
Chr.10: 73248843-73252693 on Build GRCh38
Transcripts
NR_038373.1 NR_134458.1 NR_134459.1 NR_134460.1 NM_016065.3 XM_017016305.1

SNP Detail

SNP ID Chromosome Location
rs556708182 Chr.10: 73252662-73252662 on Build GRCh38
rs1030350921 Chr.10: 73252728-73252728 on Build GRCh38
rs987964735 Chr.10: 73252617-73252617 on Build GRCh38
rs917596683 Chr.10: 73252813-73252813 on Build GRCh38
rs112170947 Chr.10: 73252589-73252589 on Build GRCh38
rs1012915949 Chr.10: 73252780-73252780 on Build GRCh38
rs778384037 Chr.10: 73252420-73252420 on Build GRCh38
rs767776570 Chr.10: 73252476-73252476 on Build GRCh38
rs774116549 Chr.10: 73252569-73252569 on Build GRCh38
rs929815086 Chr.10: 73252644-73252644 on Build GRCh38
rs866946627 Chr.10: 73252592-73252592 on Build GRCh38
rs956126967 Chr.10: 73252730-73252730 on Build GRCh38
rs560675235 Chr.10: 73252778-73252778 on Build GRCh38
rs374969072 Chr.10: 73252492-73252492 on Build GRCh38
rs1026910616 Chr.10: 73252568-73252568 on Build GRCh38
rs752831037 Chr.10: 73252481-73252481 on Build GRCh38
rs926286061 Chr.10: 73252639-73252639 on Build GRCh38
rs16930477 Chr.10: 73252708-73252708 on Build GRCh38
rs540074971 Chr.10: 73252817-73252817 on Build GRCh38
rs1046863246 Chr.10: 73252558-73252558 on Build GRCh38
rs929064428 Chr.10: 73252665-73252665 on Build GRCh38
rs539276417 Chr.10: 73252661-73252661 on Build GRCh38
rs974355126 Chr.10: 73252839-73252839 on Build GRCh38
rs549463907 Chr.10: 73252616-73252616 on Build GRCh38
rs960048650 Chr.10: 73252412-73252412 on Build GRCh38
rs916499884 Chr.10: 73252625-73252625 on Build GRCh38
rs1047871823 Chr.10: 73252691-73252691 on Build GRCh38
rs553595766 Chr.10: 73252640-73252640 on Build GRCh38
rs551492853 Chr.10: 73252554-73252554 on Build GRCh38
rs111672256 Chr.10: 73252577-73252577 on Build GRCh38
rs759778656 Chr.10: 73252475-73252475 on Build GRCh38
rs775444654 Chr.10: 73252440-73252440 on Build GRCh38
rs912346194 Chr.10: 73252655-73252655 on Build GRCh38
rs201102344 Chr.10: 73252484-73252484 on Build GRCh38
rs370675578 Chr.10: 73252818-73252818 on Build GRCh38
rs776697170 Chr.10: 73252450-73252450 on Build GRCh38
rs979348815 Chr.10: 73252635-73252635 on Build GRCh38
rs150104167 Chr.10: 73252714-73252714 on Build GRCh38
rs887970953 Chr.10: 73252692-73252692 on Build GRCh38
rs746866136 Chr.10: 73252449-73252449 on Build GRCh38
rs757774630 Chr.10: 73252503-73252503 on Build GRCh38
rs1043376293 Chr.10: 73252499-73252499 on Build GRCh38
rs1023420313 Chr.10: 73252803-73252803 on Build GRCh38
rs560766660 Chr.10: 73252458-73252458 on Build GRCh38
rs572599463 Chr.10: 73252783-73252783 on Build GRCh38
rs993644712 Chr.10: 73252547-73252547 on Build GRCh38
rs959293527 Chr.10: 73252795-73252795 on Build GRCh38
rs886047203 Chr.10: 73252665-73252665 on Build GRCh38
rs542289315 Chr.10: 73252401-73252401 on Build GRCh38
rs919731727 Chr.10: 73252643-73252643 on Build GRCh38
rs778594862 Chr.10: 73252504-73252504 on Build GRCh38
rs929056636 Chr.10: 73252820-73252820 on Build GRCh38
rs779888299 Chr.10: 73252508-73252508 on Build GRCh38
rs1017909075 Chr.10: 73252707-73252707 on Build GRCh38
rs991759381 Chr.10: 73252802-73252802 on Build GRCh38
rs755601767 Chr.10: 73252808-73252808 on Build GRCh38
rs370218765 Chr.10: 73252770-73252770 on Build GRCh38
rs761641903 Chr.10: 73252453-73252453 on Build GRCh38
rs886047201 Chr.10: 73252564-73252565 on Build GRCh38
rs369318577 Chr.10: 73252531-73252531 on Build GRCh38
rs755097246 Chr.10: 73252658-73252658 on Build GRCh38
rs904893713 Chr.10: 73252501-73252501 on Build GRCh38
rs1010761483 Chr.10: 73252737-73252737 on Build GRCh38
rs762908842 Chr.10: 73252482-73252482 on Build GRCh38
rs886047202 Chr.10: 73252629-73252629 on Build GRCh38
rs757956436 Chr.10: 73252507-73252507 on Build GRCh38
rs375981670 Chr.10: 73252422-73252422 on Build GRCh38
rs946637595 Chr.10: 73252629-73252629 on Build GRCh38
rs900120742 Chr.10: 73252576-73252576 on Build GRCh38
rs780790838 Chr.10: 73252534-73252534 on Build GRCh38
rs768281195 Chr.10: 73252506-73252515 on Build GRCh38
rs564880289 Chr.10: 73252521-73252521 on Build GRCh38
rs1035739116 Chr.10: 73252607-73252607 on Build GRCh38
rs1056178124 Chr.10: 73252527-73252527 on Build GRCh38
rs574159820 Chr.10: 73252600-73252602 on Build GRCh38
rs549047626 Chr.10: 73252563-73252563 on Build GRCh38
rs768552774 Chr.10: 73252533-73252533 on Build GRCh38
rs900076095 Chr.10: 73252664-73252664 on Build GRCh38
rs182975011 Chr.10: 73252620-73252620 on Build GRCh38
rs968322646 Chr.10: 73252761-73252761 on Build GRCh38
rs745523347 Chr.10: 73252429-73252429 on Build GRCh38
rs530527250 Chr.10: 73252384-73252384 on Build GRCh38
rs928610632 Chr.10: 73252404-73252404 on Build GRCh38
rs997484375 Chr.10: 73252713-73252713 on Build GRCh38
rs771808248 Chr.10: 73252436-73252436 on Build GRCh38
rs890773975 Chr.10: 73252564-73252565 on Build GRCh38
rs979317883 Chr.10: 73252776-73252776 on Build GRCh38
rs1039946659 Chr.10: 73252570-73252570 on Build GRCh38
rs578195824 Chr.10: 73252650-73252650 on Build GRCh38
rs367600544 Chr.10: 73252551-73252551 on Build GRCh38
rs865835072 Chr.10: 73252586-73252586 on Build GRCh38
rs1006464814 Chr.10: 73252693-73252693 on Build GRCh38
rs1020775258 Chr.10: 73252747-73252747 on Build GRCh38
rs113958603 Chr.10: 73252609-73252609 on Build GRCh38
rs552975155 Chr.10: 73252594-73252594 on Build GRCh38
rs372523400 Chr.10: 73252633-73252633 on Build GRCh38
rs984778749 Chr.10: 73252439-73252439 on Build GRCh38
rs11538567 Chr.10: 73252585-73252585 on Build GRCh38
rs1006908007 Chr.10: 73252579-73252579 on Build GRCh38
rs778511220 Chr.10: 73252423-73252423 on Build GRCh38
rs2271909 Chr.10: 73252490-73252490 on Build GRCh38
rs749900739 Chr.10: 73252506-73252506 on Build GRCh38

View Full Product Details