Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 64579706-64580230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
-
UniGene
Hs.443891
Gene Chromosome Location
Chr.1: 64470793-64693058 on Build GRCh38
Transcripts
NM_001293274.1 NM_020925.2 XM_011541862.1 XM_011541863.1 NR_039957.1

Gene Symbol
MIR4794
Entrez Gene ID
100616338
Gene Name
microRNA 4794
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.1: 64579847-64579923 on Build GRCh38
Transcripts
NM_001293274.1 NM_020925.2 XM_011541862.1 XM_011541863.1 NR_039957.1

SNP Detail

SNP ID Chromosome Location
rs148540562 Chr.1: 64579831-64579831 on Build GRCh38
rs1057151869 Chr.1: 64580128-64580128 on Build GRCh38
rs776596003 Chr.1: 64580021-64580021 on Build GRCh38
rs973810258 Chr.1: 64580049-64580049 on Build GRCh38
rs5774708 Chr.1: 64579920-64579920 on Build GRCh38
rs1001603453 Chr.1: 64579830-64579830 on Build GRCh38
rs1029961688 Chr.1: 64579773-64579773 on Build GRCh38
rs747932215 Chr.1: 64579920-64579921 on Build GRCh38
rs909759404 Chr.1: 64579740-64579740 on Build GRCh38
rs1029600947 Chr.1: 64579987-64579987 on Build GRCh38
rs397808274 Chr.1: 64579931-64579931 on Build GRCh38
rs945565778 Chr.1: 64579873-64579873 on Build GRCh38
rs945618097 Chr.1: 64580116-64580116 on Build GRCh38
rs763693323 Chr.1: 64580031-64580031 on Build GRCh38
rs937320474 Chr.1: 64580124-64580124 on Build GRCh38
rs201914776 Chr.1: 64579920-64579920 on Build GRCh38
rs1042504150 Chr.1: 64579879-64579879 on Build GRCh38
rs922306561 Chr.1: 64580057-64580057 on Build GRCh38
rs140412351 Chr.1: 64580060-64580060 on Build GRCh38
rs59458339 Chr.1: 64579836-64579836 on Build GRCh38
rs561908925 Chr.1: 64579828-64579828 on Build GRCh38
rs554943377 Chr.1: 64579898-64579898 on Build GRCh38
rs767205047 Chr.1: 64580178-64580178 on Build GRCh38
rs543526824 Chr.1: 64579869-64579869 on Build GRCh38
rs982642250 Chr.1: 64580029-64580029 on Build GRCh38
rs927311206 Chr.1: 64580118-64580118 on Build GRCh38
rs142886593 Chr.1: 64579976-64579976 on Build GRCh38
rs909716348 Chr.1: 64580099-64580099 on Build GRCh38
rs951255938 Chr.1: 64580081-64580081 on Build GRCh38
rs1000460372 Chr.1: 64580193-64580193 on Build GRCh38
rs759213973 Chr.1: 64580107-64580107 on Build GRCh38
rs941298841 Chr.1: 64580100-64580100 on Build GRCh38
rs17126710 Chr.1: 64579891-64579891 on Build GRCh38
rs1037705573 Chr.1: 64579779-64579779 on Build GRCh38
rs762087948 Chr.1: 64580150-64580150 on Build GRCh38
rs537409046 Chr.1: 64580005-64580005 on Build GRCh38
rs997889099 Chr.1: 64579983-64579983 on Build GRCh38
rs941282740 Chr.1: 64579744-64579744 on Build GRCh38
rs774219880 Chr.1: 64580095-64580095 on Build GRCh38
rs1017980867 Chr.1: 64580011-64580011 on Build GRCh38
rs1010317358 Chr.1: 64580144-64580144 on Build GRCh38
rs896690832 Chr.1: 64579837-64579837 on Build GRCh38
rs77782458 Chr.1: 64580033-64580033 on Build GRCh38
rs955441416 Chr.1: 64579992-64579992 on Build GRCh38
rs941075675 Chr.1: 64579808-64579808 on Build GRCh38
rs904674399 Chr.1: 64580176-64580176 on Build GRCh38
rs908306138 Chr.1: 64579791-64579791 on Build GRCh38
rs962493175 Chr.1: 64580034-64580034 on Build GRCh38
rs1000884513 Chr.1: 64579909-64579909 on Build GRCh38
rs895922943 Chr.1: 64580138-64580138 on Build GRCh38
rs530401370 Chr.1: 64579775-64579775 on Build GRCh38
rs536890836 Chr.1: 64579892-64579892 on Build GRCh38
rs982589833 Chr.1: 64579790-64579790 on Build GRCh38
rs969415574 Chr.1: 64579926-64579926 on Build GRCh38
rs191156964 Chr.1: 64579868-64579868 on Build GRCh38

View Full Product Details