Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 10934834-10935332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF2
Entrez Gene ID
10659
Gene Name
CUGBP, Elav-like family member 2
Gene Aliases
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
UniGene
Hs.309288
Gene Chromosome Location
Chr.10: 10462515-11336675 on Build GRCh38
Transcripts
NM_001326317.1 NM_001326318.1 NM_001326319.1 NM_001326320.1 NM_001326321.1 NM_001326323.1 NM_001326324.1 NM_001326325.1 NM_001326326.1 NM_001326327.1 NM_001326328.1 XM_017015557.1 XM_017015568.1 NR_015413.1 NR_024410.1 NR_024411.1

Gene Symbol
LINC00710
Entrez Gene ID
254312
Gene Name
long intergenic non-protein coding RNA 710
Gene Aliases
-
UniGene
Hs.498620
Gene Chromosome Location
Chr.10: 10934940-10952163 on Build GRCh38
Transcripts
NM_001326317.1 NM_001326318.1 NM_001326319.1 NM_001326320.1 NM_001326321.1 NM_001326323.1 NM_001326324.1 NM_001326325.1 NM_001326326.1 NM_001326327.1 NM_001326328.1 XM_017015557.1 XM_017015568.1 NR_015413.1 NR_024410.1 NR_024411.1

SNP Detail

SNP ID Chromosome Location
rs770424763 Chr.10: 10935087-10935087 on Build GRCh38
rs372450100 Chr.10: 10934902-10934903 on Build GRCh38
rs1034199660 Chr.10: 10934923-10934923 on Build GRCh38
rs750939312 Chr.10: 10935020-10935020 on Build GRCh38
rs567248603 Chr.10: 10935229-10935229 on Build GRCh38
rs530396052 Chr.10: 10934885-10934885 on Build GRCh38
rs751906418 Chr.10: 10935164-10935164 on Build GRCh38
rs1054204094 Chr.10: 10935071-10935071 on Build GRCh38
rs536135911 Chr.10: 10935264-10935264 on Build GRCh38
rs1026215786 Chr.10: 10935086-10935086 on Build GRCh38
rs973460766 Chr.10: 10935181-10935181 on Build GRCh38
rs115624736 Chr.10: 10935215-10935215 on Build GRCh38
rs1001842529 Chr.10: 10935145-10935145 on Build GRCh38
rs369679120 Chr.10: 10934984-10934984 on Build GRCh38
rs763469832 Chr.10: 10935024-10935024 on Build GRCh38
rs989933623 Chr.10: 10935179-10935179 on Build GRCh38
rs1010197317 Chr.10: 10934910-10934910 on Build GRCh38
rs959700788 Chr.10: 10934899-10934900 on Build GRCh38
rs945715643 Chr.10: 10935308-10935308 on Build GRCh38
rs766883761 Chr.10: 10935142-10935142 on Build GRCh38
rs898904652 Chr.10: 10935283-10935283 on Build GRCh38
rs527296242 Chr.10: 10935202-10935202 on Build GRCh38
rs912959035 Chr.10: 10935271-10935271 on Build GRCh38
rs970598090 Chr.10: 10935111-10935111 on Build GRCh38
rs537008040 Chr.10: 10935057-10935057 on Build GRCh38
rs763627140 Chr.10: 10935001-10935001 on Build GRCh38
rs760168904 Chr.10: 10934991-10934991 on Build GRCh38
rs979932610 Chr.10: 10934914-10934914 on Build GRCh38
rs555979103 Chr.10: 10935273-10935273 on Build GRCh38
rs61744549 Chr.10: 10935005-10935005 on Build GRCh38
rs975392706 Chr.10: 10935244-10935244 on Build GRCh38
rs375432356 Chr.10: 10934992-10934992 on Build GRCh38
rs959510840 Chr.10: 10934947-10934947 on Build GRCh38
rs1009873848 Chr.10: 10935103-10935103 on Build GRCh38
rs993280829 Chr.10: 10935075-10935075 on Build GRCh38
rs34083809 Chr.10: 10934869-10934870 on Build GRCh38
rs936116334 Chr.10: 10935255-10935255 on Build GRCh38
rs552242123 Chr.10: 10935146-10935146 on Build GRCh38
rs971647757 Chr.10: 10934912-10934912 on Build GRCh38
rs945005316 Chr.10: 10935234-10935234 on Build GRCh38
rs186774181 Chr.10: 10935099-10935099 on Build GRCh38
rs777238469 Chr.10: 10935106-10935106 on Build GRCh38
rs1024656839 Chr.10: 10935135-10935135 on Build GRCh38
rs771651347 Chr.10: 10934875-10934875 on Build GRCh38
rs915327508 Chr.10: 10935196-10935196 on Build GRCh38
rs959494357 Chr.10: 10935153-10935153 on Build GRCh38
rs146803327 Chr.10: 10935061-10935061 on Build GRCh38
rs962857575 Chr.10: 10935178-10935178 on Build GRCh38
rs571225085 Chr.10: 10935141-10935141 on Build GRCh38
rs796690745 Chr.10: 10935010-10935010 on Build GRCh38
rs934202501 Chr.10: 10935218-10935218 on Build GRCh38
rs755265663 Chr.10: 10935200-10935200 on Build GRCh38
rs922162960 Chr.10: 10935252-10935252 on Build GRCh38
rs775109420 Chr.10: 10934911-10934911 on Build GRCh38

View Full Product Details