Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 10944072-10944579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF2
Entrez Gene ID
10659
Gene Name
CUGBP, Elav-like family member 2
Gene Aliases
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
UniGene
Hs.309288
Gene Chromosome Location
Chr.10: 10462515-11336675 on Build GRCh38
Transcripts
NM_001326317.1 NM_001326318.1 NM_001326319.1 NM_001326320.1 NM_001326321.1 NM_001326323.1 NM_001326324.1 NM_001326325.1 NM_001326326.1 NM_001326327.1 NM_001326328.1 XM_017015557.1 XM_017015568.1 NR_015413.1 NR_024410.1 NR_024411.1

Gene Symbol
LINC00710
Entrez Gene ID
254312
Gene Name
long intergenic non-protein coding RNA 710
Gene Aliases
-
UniGene
Hs.498620
Gene Chromosome Location
Chr.10: 10934940-10952163 on Build GRCh38
Transcripts
NM_001326317.1 NM_001326318.1 NM_001326319.1 NM_001326320.1 NM_001326321.1 NM_001326323.1 NM_001326324.1 NM_001326325.1 NM_001326326.1 NM_001326327.1 NM_001326328.1 XM_017015557.1 XM_017015568.1 NR_015413.1 NR_024410.1 NR_024411.1

SNP Detail

SNP ID Chromosome Location
rs56006313 Chr.10: 10944488-10944488 on Build GRCh38
rs994384158 Chr.10: 10944408-10944408 on Build GRCh38
rs552999452 Chr.10: 10944336-10944336 on Build GRCh38
rs575418023 Chr.10: 10944513-10944513 on Build GRCh38
rs746402557 Chr.10: 10944118-10944118 on Build GRCh38
rs1009245260 Chr.10: 10944481-10944481 on Build GRCh38
rs979778964 Chr.10: 10944162-10944162 on Build GRCh38
rs923877489 Chr.10: 10944215-10944215 on Build GRCh38
rs968176561 Chr.10: 10944494-10944494 on Build GRCh38
rs1007078485 Chr.10: 10944464-10944464 on Build GRCh38
rs1000488300 Chr.10: 10944114-10944114 on Build GRCh38
rs780293592 Chr.10: 10944349-10944349 on Build GRCh38
rs539467018 Chr.10: 10944304-10944304 on Build GRCh38
rs188958448 Chr.10: 10944407-10944407 on Build GRCh38
rs957752479 Chr.10: 10944181-10944181 on Build GRCh38
rs979080474 Chr.10: 10944496-10944496 on Build GRCh38
rs956624691 Chr.10: 10944521-10944521 on Build GRCh38
rs1005570124 Chr.10: 10944345-10944345 on Build GRCh38
rs768777544 Chr.10: 10944412-10944412 on Build GRCh38
rs988190665 Chr.10: 10944218-10944218 on Build GRCh38
rs758626374 Chr.10: 10944197-10944197 on Build GRCh38
rs747143441 Chr.10: 10944402-10944402 on Build GRCh38
rs1028009274 Chr.10: 10944527-10944527 on Build GRCh38
rs992031412 Chr.10: 10944523-10944523 on Build GRCh38
rs965067077 Chr.10: 10944250-10944250 on Build GRCh38
rs895906268 Chr.10: 10944400-10944400 on Build GRCh38
rs769501596 Chr.10: 10944534-10944534 on Build GRCh38
rs950213863 Chr.10: 10944288-10944288 on Build GRCh38
rs181716761 Chr.10: 10944553-10944553 on Build GRCh38
rs1056234669 Chr.10: 10944272-10944272 on Build GRCh38
rs917708674 Chr.10: 10944275-10944275 on Build GRCh38
rs940048534 Chr.10: 10944352-10944352 on Build GRCh38
rs114760796 Chr.10: 10944341-10944341 on Build GRCh38
rs773022480 Chr.10: 10944549-10944549 on Build GRCh38
rs113322533 Chr.10: 10944144-10944144 on Build GRCh38
rs1038352232 Chr.10: 10944354-10944354 on Build GRCh38
rs1036999592 Chr.10: 10944397-10944397 on Build GRCh38
rs1000754671 Chr.10: 10944425-10944425 on Build GRCh38
rs780118038 Chr.10: 10944461-10944461 on Build GRCh38
rs550709027 Chr.10: 10944186-10944186 on Build GRCh38
rs935219874 Chr.10: 10944222-10944222 on Build GRCh38
rs570542935 Chr.10: 10944262-10944262 on Build GRCh38
rs1029877208 Chr.10: 10944422-10944422 on Build GRCh38

View Full Product Details