Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 10947337-10947833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF2
Entrez Gene ID
10659
Gene Name
CUGBP, Elav-like family member 2
Gene Aliases
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
UniGene
Hs.309288
Gene Chromosome Location
Chr.10: 10462515-11336675 on Build GRCh38
Transcripts
NM_001326317.1 NM_001326318.1 NM_001326319.1 NM_001326320.1 NM_001326321.1 NM_001326323.1 NM_001326324.1 NM_001326325.1 NM_001326326.1 NM_001326327.1 NM_001326328.1 XM_017015557.1 XM_017015568.1 NR_015413.1 NR_024410.1 NR_024411.1

Gene Symbol
LINC00710
Entrez Gene ID
254312
Gene Name
long intergenic non-protein coding RNA 710
Gene Aliases
-
UniGene
Hs.498620
Gene Chromosome Location
Chr.10: 10934940-10952163 on Build GRCh38
Transcripts
NM_001326317.1 NM_001326318.1 NM_001326319.1 NM_001326320.1 NM_001326321.1 NM_001326323.1 NM_001326324.1 NM_001326325.1 NM_001326326.1 NM_001326327.1 NM_001326328.1 XM_017015557.1 XM_017015568.1 NR_015413.1 NR_024410.1 NR_024411.1

SNP Detail

SNP ID Chromosome Location
rs529882192 Chr.10: 10947591-10947591 on Build GRCh38
rs967360991 Chr.10: 10947598-10947598 on Build GRCh38
rs892863287 Chr.10: 10947523-10947523 on Build GRCh38
rs1041013117 Chr.10: 10947689-10947689 on Build GRCh38
rs563641543 Chr.10: 10947752-10947753 on Build GRCh38
rs924107193 Chr.10: 10947749-10947752 on Build GRCh38
rs923485656 Chr.10: 10947715-10947715 on Build GRCh38
rs781224419 Chr.10: 10947577-10947577 on Build GRCh38
rs983359506 Chr.10: 10947619-10947619 on Build GRCh38
rs940898311 Chr.10: 10947460-10947460 on Build GRCh38
rs745509965 Chr.10: 10947412-10947412 on Build GRCh38
rs1035558621 Chr.10: 10947609-10947609 on Build GRCh38
rs182411235 Chr.10: 10947688-10947688 on Build GRCh38
rs201066 Chr.10: 10947738-10947738 on Build GRCh38
rs979528454 Chr.10: 10947739-10947739 on Build GRCh38
rs768249817 Chr.10: 10947670-10947670 on Build GRCh38
rs944360622 Chr.10: 10947782-10947782 on Build GRCh38
rs901999887 Chr.10: 10947492-10947492 on Build GRCh38
rs1043966826 Chr.10: 10947503-10947503 on Build GRCh38
rs549926290 Chr.10: 10947599-10947599 on Build GRCh38
rs796709955 Chr.10: 10947574-10947574 on Build GRCh38
rs939464962 Chr.10: 10947400-10947400 on Build GRCh38
rs900210288 Chr.10: 10947810-10947810 on Build GRCh38
rs532663567 Chr.10: 10947659-10947659 on Build GRCh38
rs990915620 Chr.10: 10947431-10947431 on Build GRCh38
rs147476579 Chr.10: 10947541-10947541 on Build GRCh38
rs545263178 Chr.10: 10947629-10947629 on Build GRCh38
rs371963103 Chr.10: 10947552-10947552 on Build GRCh38
rs537296113 Chr.10: 10947799-10947799 on Build GRCh38
rs994657622 Chr.10: 10947594-10947594 on Build GRCh38
rs928041470 Chr.10: 10947377-10947386 on Build GRCh38
rs192374675 Chr.10: 10947669-10947669 on Build GRCh38
rs774991669 Chr.10: 10947499-10947499 on Build GRCh38
rs868058576 Chr.10: 10947375-10947375 on Build GRCh38
rs139985428 Chr.10: 10947769-10947769 on Build GRCh38
rs908134344 Chr.10: 10947456-10947456 on Build GRCh38
rs950579855 Chr.10: 10947603-10947603 on Build GRCh38
rs1050115612 Chr.10: 10947522-10947522 on Build GRCh38
rs902747541 Chr.10: 10947507-10947507 on Build GRCh38
rs952077388 Chr.10: 10947651-10947651 on Build GRCh38
rs201065 Chr.10: 10947641-10947641 on Build GRCh38
rs958766902 Chr.10: 10947621-10947621 on Build GRCh38
rs568718248 Chr.10: 10947790-10947790 on Build GRCh38
rs374814274 Chr.10: 10947469-10947469 on Build GRCh38
rs930650836 Chr.10: 10947422-10947422 on Build GRCh38
rs892895944 Chr.10: 10947772-10947772 on Build GRCh38
rs934339090 Chr.10: 10947519-10947519 on Build GRCh38
rs35371619 Chr.10: 10947522-10947523 on Build GRCh38
rs543246170 Chr.10: 10947766-10947766 on Build GRCh38
rs1011241782 Chr.10: 10947792-10947792 on Build GRCh38

View Full Product Details