Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 11075187-11075694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF2
Entrez Gene ID
10659
Gene Name
CUGBP, Elav-like family member 2
Gene Aliases
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
UniGene
Hs.309288
Gene Chromosome Location
Chr.10: 10462515-11336675 on Build GRCh38
Transcripts
NM_001025077.2 NM_001326317.1 NM_001326318.1 NM_001326319.1 NM_001326320.1 NM_001326321.1 NM_001326323.1 NM_001326324.1 NM_001326325.1 NM_001326326.1 NM_001326327.1 NM_001326328.1 NM_001326329.1 NM_001326330.1 NM_001326331.1 NM_001326332.1 NM_001326333.1 NM_001326334.1 NM_001326335.1 NM_001326336.1 NM_001326337.1 NM_001326338.1 NM_001326339.1 NM_001326340.1 NM_001326341.1 NM_001326342.1 NM_001326343.1 NM_001326344.1 NM_001326345.1 NM_006561.3 XM_005252354.4 XM_005252357.4 XM_011519294.2 XM_017015543.1 XM_017015544.1 XM_017015545.1 XM_017015546.1 XM_017015547.1 XM_017015548.1 XM_017015549.1 XM_017015550.1 XM_017015551.1 XM_017015557.1 XM_017015558.1 XM_017015560.1 XM_017015561.1 XM_017015563.1 XM_017015565.1 XM_017015567.1 XM_017015568.1 XM_017015570.1 XM_017015572.1 NR_046108.1

Gene Symbol
CELF2-AS2
Entrez Gene ID
439950
Gene Name
CELF2 antisense RNA 2
Gene Aliases
-
UniGene
Hs.568157
Gene Chromosome Location
Chr.10: 11071541-11105488 on Build GRCh38
Transcripts
NM_001025077.2 NM_001326317.1 NM_001326318.1 NM_001326319.1 NM_001326320.1 NM_001326321.1 NM_001326323.1 NM_001326324.1 NM_001326325.1 NM_001326326.1 NM_001326327.1 NM_001326328.1 NM_001326329.1 NM_001326330.1 NM_001326331.1 NM_001326332.1 NM_001326333.1 NM_001326334.1 NM_001326335.1 NM_001326336.1 NM_001326337.1 NM_001326338.1 NM_001326339.1 NM_001326340.1 NM_001326341.1 NM_001326342.1 NM_001326343.1 NM_001326344.1 NM_001326345.1 NM_006561.3 XM_005252354.4 XM_005252357.4 XM_011519294.2 XM_017015543.1 XM_017015544.1 XM_017015545.1 XM_017015546.1 XM_017015547.1 XM_017015548.1 XM_017015549.1 XM_017015550.1 XM_017015551.1 XM_017015557.1 XM_017015558.1 XM_017015560.1 XM_017015561.1 XM_017015563.1 XM_017015565.1 XM_017015567.1 XM_017015568.1 XM_017015570.1 XM_017015572.1 NR_046108.1

SNP Detail

SNP ID Chromosome Location
rs998389158 Chr.10: 11075493-11075493 on Build GRCh38
rs1028562208 Chr.10: 11075294-11075294 on Build GRCh38
rs886152914 Chr.10: 11075438-11075438 on Build GRCh38
rs565960425 Chr.10: 11075593-11075593 on Build GRCh38
rs142327534 Chr.10: 11075516-11075516 on Build GRCh38
rs954028273 Chr.10: 11075308-11075308 on Build GRCh38
rs749556793 Chr.10: 11075445-11075445 on Build GRCh38
rs939252709 Chr.10: 11075385-11075385 on Build GRCh38
rs1035175387 Chr.10: 11075345-11075345 on Build GRCh38
rs1048768978 Chr.10: 11075428-11075428 on Build GRCh38
rs148635114 Chr.10: 11075604-11075604 on Build GRCh38
rs989478501 Chr.10: 11075312-11075312 on Build GRCh38
rs927803443 Chr.10: 11075373-11075373 on Build GRCh38
rs925692234 Chr.10: 11075227-11075227 on Build GRCh38
rs966793778 Chr.10: 11075548-11075548 on Build GRCh38
rs779282858 Chr.10: 11075551-11075551 on Build GRCh38
rs138157600 Chr.10: 11075458-11075458 on Build GRCh38
rs915614541 Chr.10: 11075389-11075389 on Build GRCh38
rs2773497 Chr.10: 11075643-11075643 on Build GRCh38
rs757714724 Chr.10: 11075416-11075416 on Build GRCh38
rs2762540 Chr.10: 11075599-11075599 on Build GRCh38
rs554998446 Chr.10: 11075454-11075454 on Build GRCh38
rs965635145 Chr.10: 11075288-11075288 on Build GRCh38
rs1028154077 Chr.10: 11075323-11075323 on Build GRCh38
rs895104825 Chr.10: 11075422-11075422 on Build GRCh38
rs145953357 Chr.10: 11075583-11075583 on Build GRCh38
rs142091867 Chr.10: 11075644-11075644 on Build GRCh38
rs114503938 Chr.10: 11075502-11075502 on Build GRCh38
rs1005058712 Chr.10: 11075453-11075453 on Build GRCh38
rs554086629 Chr.10: 11075508-11075508 on Build GRCh38
rs2762539 Chr.10: 11075217-11075217 on Build GRCh38
rs978592299 Chr.10: 11075550-11075550 on Build GRCh38
rs911722451 Chr.10: 11075459-11075459 on Build GRCh38
rs1039048849 Chr.10: 11075284-11075284 on Build GRCh38
rs901325963 Chr.10: 11075457-11075457 on Build GRCh38
rs1055031698 Chr.10: 11075414-11075414 on Build GRCh38
rs901633169 Chr.10: 11075668-11075668 on Build GRCh38
rs531711858 Chr.10: 11075669-11075669 on Build GRCh38
rs991069467 Chr.10: 11075366-11075366 on Build GRCh38
rs1016182208 Chr.10: 11075273-11075273 on Build GRCh38
rs185611529 Chr.10: 11075595-11075595 on Build GRCh38
rs954327911 Chr.10: 11075503-11075503 on Build GRCh38
rs978412256 Chr.10: 11075426-11075426 on Build GRCh38
rs189900390 Chr.10: 11075277-11075277 on Build GRCh38
rs930289410 Chr.10: 11075423-11075423 on Build GRCh38
rs977911137 Chr.10: 11075357-11075357 on Build GRCh38
rs1022255363 Chr.10: 11075532-11075532 on Build GRCh38
rs890400745 Chr.10: 11075243-11075243 on Build GRCh38
rs535102961 Chr.10: 11075394-11075394 on Build GRCh38
rs375247732 Chr.10: 11075240-11075240 on Build GRCh38
rs1040510374 Chr.10: 11075653-11075653 on Build GRCh38
rs995262333 Chr.10: 11075542-11075542 on Build GRCh38
rs900375647 Chr.10: 11075528-11075528 on Build GRCh38
rs182457873 Chr.10: 11075292-11075292 on Build GRCh38
rs950809572 Chr.10: 11075328-11075328 on Build GRCh38
rs945460383 Chr.10: 11075354-11075354 on Build GRCh38
rs76432107 Chr.10: 11075329-11075329 on Build GRCh38
rs886225241 Chr.10: 11075232-11075232 on Build GRCh38
rs928098289 Chr.10: 11075561-11075561 on Build GRCh38
rs960802474 Chr.10: 11075568-11075568 on Build GRCh38

View Full Product Details