Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 11850406-11850937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROSER2
Entrez Gene ID
254427
Gene Name
proline and serine rich 2
Gene Aliases
C10orf47
UniGene
Hs.435775
Gene Chromosome Location
Chr.10: 11823341-11872277 on Build GRCh38
Transcripts
NM_153256.3 XM_011519438.2 XM_017016028.1 NR_038222.1

Gene Symbol
PROSER2-AS1
Entrez Gene ID
219731
Gene Name
PROSER2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.576787
Gene Chromosome Location
Chr.10: 11849608-11894710 on Build GRCh38
Transcripts
NM_153256.3 XM_011519438.2 XM_017016028.1 NR_038222.1

SNP Detail

SNP ID Chromosome Location
rs1053850277 Chr.10: 11850494-11850494 on Build GRCh38
rs535637126 Chr.10: 11850633-11850633 on Build GRCh38
rs371904254 Chr.10: 11850622-11850622 on Build GRCh38
rs945664607 Chr.10: 11850600-11850600 on Build GRCh38
rs1042578478 Chr.10: 11850834-11850834 on Build GRCh38
rs954265155 Chr.10: 11850620-11850620 on Build GRCh38
rs1041423728 Chr.10: 11850604-11850604 on Build GRCh38
rs147920719 Chr.10: 11850754-11850754 on Build GRCh38
rs1017392650 Chr.10: 11850448-11850448 on Build GRCh38
rs192487919 Chr.10: 11850642-11850642 on Build GRCh38
rs1021173395 Chr.10: 11850510-11850510 on Build GRCh38
rs1001992568 Chr.10: 11850489-11850489 on Build GRCh38
rs965482197 Chr.10: 11850541-11850541 on Build GRCh38
rs954262924 Chr.10: 11850896-11850897 on Build GRCh38
rs1009748777 Chr.10: 11850501-11850501 on Build GRCh38
rs904937181 Chr.10: 11850456-11850456 on Build GRCh38
rs893442389 Chr.10: 11850805-11850805 on Build GRCh38
rs1046649492 Chr.10: 11850766-11850766 on Build GRCh38
rs539885688 Chr.10: 11850820-11850820 on Build GRCh38
rs1028377396 Chr.10: 11850797-11850797 on Build GRCh38
rs975974825 Chr.10: 11850521-11850521 on Build GRCh38
rs539697165 Chr.10: 11850512-11850512 on Build GRCh38
rs914164430 Chr.10: 11850595-11850595 on Build GRCh38
rs975501877 Chr.10: 11850789-11850789 on Build GRCh38
rs112545015 Chr.10: 11850730-11850730 on Build GRCh38
rs1016966947 Chr.10: 11850742-11850742 on Build GRCh38
rs986984625 Chr.10: 11850572-11850572 on Build GRCh38
rs949159933 Chr.10: 11850763-11850763 on Build GRCh38
rs577225623 Chr.10: 11850774-11850774 on Build GRCh38
rs986933235 Chr.10: 11850815-11850815 on Build GRCh38
rs934182136 Chr.10: 11850571-11850571 on Build GRCh38
rs894290251 Chr.10: 11850643-11850643 on Build GRCh38
rs10906027 Chr.10: 11850431-11850431 on Build GRCh38
rs145534214 Chr.10: 11850605-11850605 on Build GRCh38
rs945681605 Chr.10: 11850844-11850844 on Build GRCh38
rs887025044 Chr.10: 11850709-11850709 on Build GRCh38
rs776602886 Chr.10: 11850562-11850562 on Build GRCh38
rs148425470 Chr.10: 11850660-11850660 on Build GRCh38
rs925664137 Chr.10: 11850895-11850895 on Build GRCh38
rs982809089 Chr.10: 11850437-11850437 on Build GRCh38
rs1053558010 Chr.10: 11850802-11850802 on Build GRCh38
rs977478766 Chr.10: 11850845-11850845 on Build GRCh38
rs769620737 Chr.10: 11850653-11850653 on Build GRCh38
rs935792631 Chr.10: 11850606-11850606 on Build GRCh38
rs574981553 Chr.10: 11850716-11850716 on Build GRCh38
rs551763645 Chr.10: 11850447-11850447 on Build GRCh38
rs909857919 Chr.10: 11850658-11850658 on Build GRCh38
rs75150568 Chr.10: 11850547-11850547 on Build GRCh38
rs867493085 Chr.10: 11850808-11850808 on Build GRCh38
rs376763851 Chr.10: 11850861-11850861 on Build GRCh38
rs1009193900 Chr.10: 11850812-11850812 on Build GRCh38
rs1028898845 Chr.10: 11850896-11850896 on Build GRCh38
rs571690260 Chr.10: 11850498-11850498 on Build GRCh38

View Full Product Details