Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88992484-88993022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAS
Entrez Gene ID
355
Gene Name
Fas cell surface death receptor
Gene Aliases
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
UniGene
Hs.244139
Gene Chromosome Location
Chr.10: 88968429-89017061 on Build GRCh38
Transcripts
NM_000043.5 NM_001320619.1 NM_152871.3 NM_152872.3 NR_028033.3 NR_028034.3 NR_028035.3 NR_028036.3 NR_135313.1 NR_135314.1 NR_135315.1 XM_006717819.3 XM_011539764.2 XM_011539765.2 XM_011539766.2 XM_011539767.2 NR_028371.1

Gene Symbol
FAS-AS1
Entrez Gene ID
100302740
Gene Name
FAS antisense RNA 1
Gene Aliases
FAS-AS, FASAS, SAF
UniGene
Hs.244139
Gene Chromosome Location
Chr.10: 88991422-88992975 on Build GRCh38
Transcripts
NM_000043.5 NM_001320619.1 NM_152871.3 NM_152872.3 NR_028033.3 NR_028034.3 NR_028035.3 NR_028036.3 NR_135313.1 NR_135314.1 NR_135315.1 XM_006717819.3 XM_011539764.2 XM_011539765.2 XM_011539766.2 XM_011539767.2 NR_028371.1

SNP Detail

SNP ID Chromosome Location
rs917988794 Chr.10: 88992507-88992507 on Build GRCh38
rs988588867 Chr.10: 88992828-88992828 on Build GRCh38
rs181378579 Chr.10: 88992914-88992914 on Build GRCh38
rs890865194 Chr.10: 88992625-88992625 on Build GRCh38
rs964004662 Chr.10: 88992915-88992915 on Build GRCh38
rs975856757 Chr.10: 88992928-88992928 on Build GRCh38
rs1045057815 Chr.10: 88992539-88992542 on Build GRCh38
rs760170390 Chr.10: 88992878-88992878 on Build GRCh38
rs538207455 Chr.10: 88992653-88992653 on Build GRCh38
rs922629309 Chr.10: 88992984-88992984 on Build GRCh38
rs992326105 Chr.10: 88992730-88992733 on Build GRCh38
rs980855319 Chr.10: 88992777-88992777 on Build GRCh38
rs1006611223 Chr.10: 88992641-88992641 on Build GRCh38
rs755588877 Chr.10: 88992738-88992738 on Build GRCh38
rs1023881975 Chr.10: 88992813-88992813 on Build GRCh38
rs942724855 Chr.10: 88992645-88992645 on Build GRCh38
rs922542189 Chr.10: 88992654-88992654 on Build GRCh38
rs547515624 Chr.10: 88992700-88992700 on Build GRCh38
rs538469186 Chr.10: 88992837-88992837 on Build GRCh38
rs767743492 Chr.10: 88992880-88992880 on Build GRCh38
rs894128255 Chr.10: 88992799-88992799 on Build GRCh38
rs906819023 Chr.10: 88992816-88992816 on Build GRCh38
rs1051395418 Chr.10: 88992620-88992620 on Build GRCh38
rs539587358 Chr.10: 88992919-88992919 on Build GRCh38
rs12571917 Chr.10: 88992773-88992773 on Build GRCh38
rs1025576820 Chr.10: 88992758-88992758 on Build GRCh38
rs3824729 Chr.10: 88992859-88992859 on Build GRCh38
rs73368853 Chr.10: 88992551-88992551 on Build GRCh38
rs955433716 Chr.10: 88992999-88992999 on Build GRCh38
rs1031082292 Chr.10: 88992510-88992510 on Build GRCh38
rs3781204 Chr.10: 88992586-88992586 on Build GRCh38
rs925293026 Chr.10: 88992545-88992545 on Build GRCh38
rs933835759 Chr.10: 88992681-88992681 on Build GRCh38
rs890959022 Chr.10: 88992964-88992964 on Build GRCh38
rs945986173 Chr.10: 88992860-88992860 on Build GRCh38
rs1047293779 Chr.10: 88992718-88992718 on Build GRCh38
rs747598958 Chr.10: 88992598-88992598 on Build GRCh38
rs1010924533 Chr.10: 88992900-88992900 on Build GRCh38
rs956750836 Chr.10: 88992550-88992550 on Build GRCh38
rs1054388363 Chr.10: 88992585-88992586 on Build GRCh38
rs951740237 Chr.10: 88992725-88992725 on Build GRCh38
rs1038143874 Chr.10: 88992760-88992760 on Build GRCh38
rs761912209 Chr.10: 88992603-88992604 on Build GRCh38
rs555800740 Chr.10: 88992755-88992755 on Build GRCh38
rs574161354 Chr.10: 88992823-88992823 on Build GRCh38
rs886947977 Chr.10: 88992729-88992729 on Build GRCh38
rs565544568 Chr.10: 88992585-88992585 on Build GRCh38
rs1013027435 Chr.10: 88992812-88992812 on Build GRCh38

View Full Product Details