Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 117544467-117544966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMX2
Entrez Gene ID
2018
Gene Name
empty spiracles homeobox 2
Gene Aliases
-
UniGene
Hs.202095
Gene Chromosome Location
Chr.10: 117542445-117549546 on Build GRCh38
Transcripts
NM_001165924.1 NM_004098.3 NR_002791.2

Gene Symbol
EMX2OS
Entrez Gene ID
196047
Gene Name
EMX2 opposite strand/antisense RNA
Gene Aliases
EMX2-AS1, NCRNA00045
UniGene
Hs.312592
Gene Chromosome Location
Chr.10: 117484293-117545068 on Build GRCh38
Transcripts
NM_001165924.1 NM_004098.3 NR_002791.2

SNP Detail

SNP ID Chromosome Location
rs1007985984 Chr.10: 117544662-117544662 on Build GRCh38
rs144615230 Chr.10: 117544774-117544774 on Build GRCh38
rs779609690 Chr.10: 117544655-117544655 on Build GRCh38
rs991171163 Chr.10: 117544731-117544731 on Build GRCh38
rs778741263 Chr.10: 117544709-117544709 on Build GRCh38
rs1003408745 Chr.10: 117544581-117544581 on Build GRCh38
rs1003785313 Chr.10: 117544788-117544788 on Build GRCh38
rs77117305 Chr.10: 117544696-117544696 on Build GRCh38
rs1054952428 Chr.10: 117544658-117544658 on Build GRCh38
rs893885466 Chr.10: 117544542-117544542 on Build GRCh38
rs1042854196 Chr.10: 117544725-117544725 on Build GRCh38
rs855769 Chr.10: 117544585-117544585 on Build GRCh38
rs568516949 Chr.10: 117544573-117544573 on Build GRCh38
rs1028367164 Chr.10: 117544495-117544495 on Build GRCh38
rs907631915 Chr.10: 117544671-117544671 on Build GRCh38
rs934831914 Chr.10: 117544803-117544803 on Build GRCh38
rs915602856 Chr.10: 117544740-117544740 on Build GRCh38
rs1038999736 Chr.10: 117544818-117544818 on Build GRCh38
rs542380156 Chr.10: 117544781-117544781 on Build GRCh38
rs951719644 Chr.10: 117544664-117544664 on Build GRCh38
rs998054118 Chr.10: 117544879-117544879 on Build GRCh38
rs573078784 Chr.10: 117544724-117544724 on Build GRCh38
rs144153936 Chr.10: 117544915-117544915 on Build GRCh38
rs893772434 Chr.10: 117544716-117544716 on Build GRCh38
rs909545958 Chr.10: 117544507-117544507 on Build GRCh38
rs1010929028 Chr.10: 117544721-117544721 on Build GRCh38
rs1035332373 Chr.10: 117544582-117544582 on Build GRCh38
rs543152640 Chr.10: 117544831-117544831 on Build GRCh38
rs779545172 Chr.10: 117544881-117544881 on Build GRCh38
rs755054903 Chr.10: 117544890-117544890 on Build GRCh38
rs385209 Chr.10: 117544545-117544545 on Build GRCh38
rs181158316 Chr.10: 117544892-117544892 on Build GRCh38
rs534693550 Chr.10: 117544591-117544591 on Build GRCh38
rs1029536289 Chr.10: 117544880-117544880 on Build GRCh38
rs1007668982 Chr.10: 117544817-117544817 on Build GRCh38
rs769560576 Chr.10: 117544872-117544872 on Build GRCh38
rs951687555 Chr.10: 117544868-117544868 on Build GRCh38
rs1021050981 Chr.10: 117544546-117544546 on Build GRCh38
rs1046054258 Chr.10: 117544759-117544759 on Build GRCh38
rs1012082432 Chr.10: 117544849-117544849 on Build GRCh38
rs559714904 Chr.10: 117544926-117544926 on Build GRCh38
rs200373841 Chr.10: 117544689-117544690 on Build GRCh38
rs983141634 Chr.10: 117544666-117544666 on Build GRCh38
rs1027548384 Chr.10: 117544852-117544852 on Build GRCh38
rs753687427 Chr.10: 117544698-117544698 on Build GRCh38
rs190329342 Chr.10: 117544699-117544699 on Build GRCh38
rs370838318 Chr.10: 117544707-117544707 on Build GRCh38
rs1029441651 Chr.10: 117544690-117544690 on Build GRCh38
rs902621070 Chr.10: 117544741-117544741 on Build GRCh38
rs950267193 Chr.10: 117544756-117544756 on Build GRCh38
rs365446 Chr.10: 117544804-117544804 on Build GRCh38
rs139266889 Chr.10: 117544813-117544813 on Build GRCh38
rs576952616 Chr.10: 117544904-117544904 on Build GRCh38
rs752356685 Chr.10: 117544493-117544493 on Build GRCh38
rs903379647 Chr.10: 117544638-117544638 on Build GRCh38
rs920230331 Chr.10: 117544613-117544613 on Build GRCh38
rs116998855 Chr.10: 117544677-117544677 on Build GRCh38
rs977315158 Chr.10: 117544902-117544902 on Build GRCh38
rs893103395 Chr.10: 117544659-117544659 on Build GRCh38

View Full Product Details