Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 117544749-117545268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMX2
Entrez Gene ID
2018
Gene Name
empty spiracles homeobox 2
Gene Aliases
-
UniGene
Hs.202095
Gene Chromosome Location
Chr.10: 117542445-117549546 on Build GRCh38
Transcripts
NM_001165924.1 NM_004098.3 NR_002791.2

Gene Symbol
EMX2OS
Entrez Gene ID
196047
Gene Name
EMX2 opposite strand/antisense RNA
Gene Aliases
EMX2-AS1, NCRNA00045
UniGene
Hs.312592
Gene Chromosome Location
Chr.10: 117484293-117545068 on Build GRCh38
Transcripts
NM_001165924.1 NM_004098.3 NR_002791.2

SNP Detail

SNP ID Chromosome Location
rs528400800 Chr.10: 117544975-117544975 on Build GRCh38
rs144615230 Chr.10: 117544774-117544774 on Build GRCh38
rs561742447 Chr.10: 117545156-117545156 on Build GRCh38
rs974763822 Chr.10: 117545241-117545241 on Build GRCh38
rs1049980613 Chr.10: 117544964-117544964 on Build GRCh38
rs981123515 Chr.10: 117545154-117545154 on Build GRCh38
rs1056524915 Chr.10: 117545242-117545242 on Build GRCh38
rs954164056 Chr.10: 117545171-117545171 on Build GRCh38
rs975818255 Chr.10: 117545158-117545158 on Build GRCh38
rs1014623230 Chr.10: 117545114-117545114 on Build GRCh38
rs924870542 Chr.10: 117544964-117544966 on Build GRCh38
rs1003785313 Chr.10: 117544788-117544788 on Build GRCh38
rs933329036 Chr.10: 117545022-117545022 on Build GRCh38
rs915494979 Chr.10: 117545197-117545197 on Build GRCh38
rs1042430752 Chr.10: 117545026-117545026 on Build GRCh38
rs902588563 Chr.10: 117545042-117545042 on Build GRCh38
rs934831914 Chr.10: 117544803-117544803 on Build GRCh38
rs923928179 Chr.10: 117545213-117545213 on Build GRCh38
rs1054529072 Chr.10: 117545082-117545082 on Build GRCh38
rs1038999736 Chr.10: 117544818-117544818 on Build GRCh38
rs990706962 Chr.10: 117544976-117544976 on Build GRCh38
rs542380156 Chr.10: 117544781-117544781 on Build GRCh38
rs998054118 Chr.10: 117544879-117544879 on Build GRCh38
rs144153936 Chr.10: 117544915-117544915 on Build GRCh38
rs914777619 Chr.10: 117544990-117544990 on Build GRCh38
rs894961412 Chr.10: 117545072-117545072 on Build GRCh38
rs1004951076 Chr.10: 117545106-117545106 on Build GRCh38
rs939368495 Chr.10: 117545216-117545216 on Build GRCh38
rs879041716 Chr.10: 117545069-117545069 on Build GRCh38
rs781475452 Chr.10: 117544963-117544964 on Build GRCh38
rs932808639 Chr.10: 117544953-117544953 on Build GRCh38
rs543152640 Chr.10: 117544831-117544831 on Build GRCh38
rs971022691 Chr.10: 117545145-117545145 on Build GRCh38
rs779545172 Chr.10: 117544881-117544881 on Build GRCh38
rs755054903 Chr.10: 117544890-117544890 on Build GRCh38
rs181158316 Chr.10: 117544892-117544892 on Build GRCh38
rs117965495 Chr.10: 117545044-117545044 on Build GRCh38
rs575712997 Chr.10: 117544969-117544969 on Build GRCh38
rs946682583 Chr.10: 117545017-117545017 on Build GRCh38
rs1029536289 Chr.10: 117544880-117544880 on Build GRCh38
rs768179477 Chr.10: 117545245-117545245 on Build GRCh38
rs946987108 Chr.10: 117545208-117545208 on Build GRCh38
rs1007668982 Chr.10: 117544817-117544817 on Build GRCh38
rs769560576 Chr.10: 117544872-117544872 on Build GRCh38
rs966107719 Chr.10: 117545147-117545147 on Build GRCh38
rs951687555 Chr.10: 117544868-117544868 on Build GRCh38
rs1012049740 Chr.10: 117545077-117545077 on Build GRCh38
rs914724120 Chr.10: 117545217-117545217 on Build GRCh38
rs1012082432 Chr.10: 117544849-117544849 on Build GRCh38
rs559714904 Chr.10: 117544926-117544926 on Build GRCh38
rs915508182 Chr.10: 117545004-117545004 on Build GRCh38
rs1027548384 Chr.10: 117544852-117544852 on Build GRCh38
rs887718105 Chr.10: 117545091-117545091 on Build GRCh38
rs1039135350 Chr.10: 117545019-117545019 on Build GRCh38
rs1022629552 Chr.10: 117544949-117544949 on Build GRCh38
rs365446 Chr.10: 117544804-117544804 on Build GRCh38
rs1029176748 Chr.10: 117545163-117545163 on Build GRCh38
rs139266889 Chr.10: 117544813-117544813 on Build GRCh38
rs1056956346 Chr.10: 117545071-117545071 on Build GRCh38
rs576952616 Chr.10: 117544904-117544904 on Build GRCh38
rs985651812 Chr.10: 117545184-117545184 on Build GRCh38
rs1022729690 Chr.10: 117545121-117545121 on Build GRCh38
rs901810812 Chr.10: 117545022-117545022 on Build GRCh38
rs943540990 Chr.10: 117545231-117545231 on Build GRCh38
rs532655092 Chr.10: 117545209-117545209 on Build GRCh38
rs977315158 Chr.10: 117544902-117544902 on Build GRCh38

View Full Product Details