Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 118207516-118208062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC2
Entrez Gene ID
255082
Gene Name
cancer susceptibility candidate 2 (non-protein coding)
Gene Aliases
C10orf5
UniGene
Hs.89387
Gene Chromosome Location
Chr.10: 118046821-118210153 on Build GRCh38
Transcripts
NR_026939.1 NR_026940.1

SNP Detail

SNP ID Chromosome Location
rs923426390 Chr.10: 118207868-118207868 on Build GRCh38
rs1038294433 Chr.10: 118207816-118207816 on Build GRCh38
rs114286764 Chr.10: 118207694-118207694 on Build GRCh38
rs990733534 Chr.10: 118207546-118207546 on Build GRCh38
rs956895826 Chr.10: 118207898-118207898 on Build GRCh38
rs952074961 Chr.10: 118207568-118207568 on Build GRCh38
rs1035078930 Chr.10: 118207889-118207889 on Build GRCh38
rs528867529 Chr.10: 118207906-118207906 on Build GRCh38
rs952025925 Chr.10: 118207767-118207767 on Build GRCh38
rs564286310 Chr.10: 118207763-118207763 on Build GRCh38
rs376768199 Chr.10: 118207761-118207761 on Build GRCh38
rs982298866 Chr.10: 118207684-118207684 on Build GRCh38
rs963431409 Chr.10: 118207593-118207593 on Build GRCh38
rs1006140637 Chr.10: 118208036-118208036 on Build GRCh38
rs748051669 Chr.10: 118207790-118207790 on Build GRCh38
rs548719828 Chr.10: 118207922-118207922 on Build GRCh38
rs562013465 Chr.10: 118207970-118207973 on Build GRCh38
rs772698787 Chr.10: 118207858-118207858 on Build GRCh38
rs853569 Chr.10: 118207725-118207725 on Build GRCh38
rs907985717 Chr.10: 118207589-118207589 on Build GRCh38
rs542197715 Chr.10: 118207670-118207670 on Build GRCh38
rs1043415045 Chr.10: 118207930-118207930 on Build GRCh38
rs1043466843 Chr.10: 118207700-118207700 on Build GRCh38
rs146281077 Chr.10: 118207738-118207738 on Build GRCh38
rs1023620595 Chr.10: 118207832-118207832 on Build GRCh38
rs781607982 Chr.10: 118207606-118207606 on Build GRCh38
rs923670335 Chr.10: 118207960-118207960 on Build GRCh38
rs116538927 Chr.10: 118207831-118207831 on Build GRCh38
rs73423502 Chr.10: 118207827-118207827 on Build GRCh38
rs534230864 Chr.10: 118207632-118207632 on Build GRCh38
rs11540434 Chr.10: 118207757-118207757 on Build GRCh38
rs976232442 Chr.10: 118207856-118207856 on Build GRCh38
rs560217636 Chr.10: 118207900-118207900 on Build GRCh38
rs935065282 Chr.10: 118207993-118207993 on Build GRCh38
rs1369 Chr.10: 118207731-118207731 on Build GRCh38
rs1003641961 Chr.10: 118207880-118207880 on Build GRCh38
rs976666744 Chr.10: 118207613-118207613 on Build GRCh38
rs887705288 Chr.10: 118208035-118208035 on Build GRCh38
rs975072720 Chr.10: 118208029-118208029 on Build GRCh38
rs1023581920 Chr.10: 118207565-118207565 on Build GRCh38
rs923508569 Chr.10: 118207631-118207631 on Build GRCh38
rs988428680 Chr.10: 118207727-118207727 on Build GRCh38
rs986363798 Chr.10: 118207910-118207910 on Build GRCh38
rs896351712 Chr.10: 118207744-118207744 on Build GRCh38
rs1012103794 Chr.10: 118207752-118207752 on Build GRCh38
rs889443294 Chr.10: 118207686-118207686 on Build GRCh38
rs969457114 Chr.10: 118207668-118207668 on Build GRCh38
rs1004432295 Chr.10: 118207777-118207777 on Build GRCh38
rs73423500 Chr.10: 118207645-118207645 on Build GRCh38
rs1039337690 Chr.10: 118207740-118207740 on Build GRCh38
rs546262976 Chr.10: 118208005-118208005 on Build GRCh38
rs1012054642 Chr.10: 118208022-118208022 on Build GRCh38
rs914838987 Chr.10: 118207916-118207916 on Build GRCh38
rs867846620 Chr.10: 118207659-118207659 on Build GRCh38
rs757533528 Chr.10: 118207582-118207582 on Build GRCh38
rs1044934050 Chr.10: 118208023-118208023 on Build GRCh38
rs747149729 Chr.10: 118207576-118207576 on Build GRCh38
rs960910990 Chr.10: 118207544-118207544 on Build GRCh38
rs568585345 Chr.10: 118207996-118207996 on Build GRCh38
rs886961611 Chr.10: 118207773-118207773 on Build GRCh38

View Full Product Details