Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8194527-8195034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM66C
Entrez Gene ID
440078
Gene Name
family with sequence similarity 66 member C
Gene Aliases
-
UniGene
Hs.512257
Gene Chromosome Location
Chr.12: 8180209-8201000 on Build GRCh38
Transcripts
NR_026788.1

SNP Detail

SNP ID Chromosome Location
rs78481554 Chr.12: 8194568-8194568 on Build GRCh38
rs887252224 Chr.12: 8194984-8194984 on Build GRCh38
rs773476387 Chr.12: 8194761-8194761 on Build GRCh38
rs368069935 Chr.12: 8194608-8194608 on Build GRCh38
rs1056855231 Chr.12: 8194646-8194646 on Build GRCh38
rs10770308 Chr.12: 8194762-8194762 on Build GRCh38
rs144121911 Chr.12: 8194987-8194987 on Build GRCh38
rs947182282 Chr.12: 8194603-8194603 on Build GRCh38
rs948003583 Chr.12: 8194935-8194935 on Build GRCh38
rs565212318 Chr.12: 8194695-8194695 on Build GRCh38
rs975272057 Chr.12: 8194582-8194582 on Build GRCh38
rs1056531272 Chr.12: 8194910-8194910 on Build GRCh38
rs75932809 Chr.12: 8194551-8194551 on Build GRCh38
rs75621498 Chr.12: 8194685-8194685 on Build GRCh38
rs867046742 Chr.12: 8194640-8194640 on Build GRCh38
rs963657298 Chr.12: 8194800-8194800 on Build GRCh38
rs75843697 Chr.12: 8194648-8194648 on Build GRCh38
rs143454960 Chr.12: 8194833-8194833 on Build GRCh38
rs1047659046 Chr.12: 8194968-8194968 on Build GRCh38
rs951185784 Chr.12: 8194781-8194781 on Build GRCh38
rs77268408 Chr.12: 8194689-8194689 on Build GRCh38
rs77826260 Chr.12: 8194570-8194570 on Build GRCh38
rs773902001 Chr.12: 8194628-8194628 on Build GRCh38
rs779253683 Chr.12: 8194738-8194738 on Build GRCh38
rs996459443 Chr.12: 8194999-8194999 on Build GRCh38
rs80112877 Chr.12: 8194900-8194900 on Build GRCh38
rs112171088 Chr.12: 8194566-8194566 on Build GRCh38
rs1019321531 Chr.12: 8194989-8194989 on Build GRCh38
rs567126848 Chr.12: 8194955-8194955 on Build GRCh38
rs766883091 Chr.12: 8194817-8194817 on Build GRCh38
rs924078758 Chr.12: 8194878-8194878 on Build GRCh38
rs968563385 Chr.12: 8194839-8194839 on Build GRCh38
rs1045304646 Chr.12: 8194719-8194719 on Build GRCh38
rs77636294 Chr.12: 8194899-8194899 on Build GRCh38
rs902595012 Chr.12: 8194624-8194624 on Build GRCh38
rs1019711347 Chr.12: 8194796-8194796 on Build GRCh38
rs181650959 Chr.12: 8194951-8194951 on Build GRCh38
rs977207124 Chr.12: 8194863-8194863 on Build GRCh38
rs750923382 Chr.12: 8194886-8194886 on Build GRCh38
rs1043796947 Chr.12: 8194605-8194605 on Build GRCh38
rs1007921175 Chr.12: 8194783-8194783 on Build GRCh38
rs995696985 Chr.12: 8194956-8194956 on Build GRCh38
rs184733532 Chr.12: 8194634-8194634 on Build GRCh38
rs914424361 Chr.12: 8194594-8194594 on Build GRCh38
rs189498680 Chr.12: 8194932-8194932 on Build GRCh38
rs935426596 Chr.12: 8194904-8194904 on Build GRCh38
rs1012523897 Chr.12: 8194703-8194703 on Build GRCh38

View Full Product Details