Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 127898228-127898741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ37505
Entrez Gene ID
400087
Gene Name
uncharacterized LOC400087
Gene Aliases
-
UniGene
Hs.524846
Gene Chromosome Location
Chr.12: 127881617-127898639 on Build GRCh38
Transcripts
NR_033987.1 NR_126452.2

Gene Symbol
LINC00508
Entrez Gene ID
104472718
Gene Name
long intergenic non-protein coding RNA 508
Gene Aliases
-
UniGene
Hs.628661
Gene Chromosome Location
Chr.12: 127883989-127983891 on Build GRCh38
Transcripts
NR_033987.1 NR_126452.2

SNP Detail

SNP ID Chromosome Location
rs973278825 Chr.12: 127898580-127898580 on Build GRCh38
rs1037550114 Chr.12: 127898556-127898556 on Build GRCh38
rs890321506 Chr.12: 127898492-127898492 on Build GRCh38
rs983444806 Chr.12: 127898515-127898515 on Build GRCh38
rs532868869 Chr.12: 127898267-127898267 on Build GRCh38
rs755783447 Chr.12: 127898516-127898516 on Build GRCh38
rs1037528722 Chr.12: 127898290-127898290 on Build GRCh38
rs937087444 Chr.12: 127898573-127898573 on Build GRCh38
rs941910738 Chr.12: 127898536-127898536 on Build GRCh38
rs947055783 Chr.12: 127898404-127898404 on Build GRCh38
rs186306595 Chr.12: 127898309-127898309 on Build GRCh38
rs536304774 Chr.12: 127898692-127898692 on Build GRCh38
rs1032449107 Chr.12: 127898435-127898435 on Build GRCh38
rs974018975 Chr.12: 127898322-127898322 on Build GRCh38
rs920021122 Chr.12: 127898328-127898328 on Build GRCh38
rs1009885813 Chr.12: 127898647-127898647 on Build GRCh38
rs781046732 Chr.12: 127898454-127898454 on Build GRCh38
rs182970430 Chr.12: 127898648-127898648 on Build GRCh38
rs951380357 Chr.12: 127898693-127898693 on Build GRCh38
rs1030081617 Chr.12: 127898459-127898459 on Build GRCh38
rs998631584 Chr.12: 127898455-127898455 on Build GRCh38
rs982868044 Chr.12: 127898349-127898349 on Build GRCh38
rs369344338 Chr.12: 127898630-127898630 on Build GRCh38
rs1056920903 Chr.12: 127898608-127898608 on Build GRCh38
rs928835664 Chr.12: 127898356-127898356 on Build GRCh38
rs895442473 Chr.12: 127898344-127898344 on Build GRCh38
rs986494817 Chr.12: 127898254-127898254 on Build GRCh38
rs534250025 Chr.12: 127898668-127898668 on Build GRCh38
rs139323668 Chr.12: 127898252-127898252 on Build GRCh38
rs113939500 Chr.12: 127898397-127898397 on Build GRCh38
rs941879438 Chr.12: 127898289-127898289 on Build GRCh38
rs930088433 Chr.12: 127898345-127898345 on Build GRCh38
rs779515340 Chr.12: 127898663-127898663 on Build GRCh38
rs1019888981 Chr.12: 127898651-127898651 on Build GRCh38
rs1463668 Chr.12: 127898382-127898382 on Build GRCh38
rs1001260604 Chr.12: 127898317-127898317 on Build GRCh38
rs575276956 Chr.12: 127898719-127898719 on Build GRCh38
rs886354699 Chr.12: 127898286-127898286 on Build GRCh38
rs78974693 Chr.12: 127898572-127898572 on Build GRCh38
rs895546464 Chr.12: 127898612-127898612 on Build GRCh38
rs142857789 Chr.12: 127898451-127898451 on Build GRCh38
rs112544132 Chr.12: 127898260-127898260 on Build GRCh38
rs190325578 Chr.12: 127898529-127898529 on Build GRCh38
rs1035769366 Chr.12: 127898339-127898339 on Build GRCh38
rs933377644 Chr.12: 127898504-127898504 on Build GRCh38
rs954552881 Chr.12: 127898253-127898253 on Build GRCh38
rs2730775 Chr.12: 127898360-127898360 on Build GRCh38
rs2699095 Chr.12: 127898437-127898437 on Build GRCh38
rs745483314 Chr.12: 127898496-127898496 on Build GRCh38

View Full Product Details