Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85282751-85283297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSE1
Entrez Gene ID
23199
Gene Name
Gse1 coiled-coil protein
Gene Aliases
CRHSP24, KIAA0182
UniGene
Hs.461647
Gene Chromosome Location
Chr.16: 85169512-85676206 on Build GRCh38
Transcripts
XM_005255859.4 XM_005255860.3 XM_005255861.4 NR_038859.1

Gene Symbol
LINC00311
Entrez Gene ID
197196
Gene Name
long intergenic non-protein coding RNA 311
Gene Aliases
NCRNA00311, TMEM148
UniGene
Hs.679002
Gene Chromosome Location
Chr.16: 85282958-85288082 on Build GRCh38
Transcripts
XM_005255859.4 XM_005255860.3 XM_005255861.4 NR_038859.1

SNP Detail

SNP ID Chromosome Location
rs749998204 Chr.16: 85282865-85282865 on Build GRCh38
rs193204586 Chr.16: 85283044-85283044 on Build GRCh38
rs547762543 Chr.16: 85283238-85283238 on Build GRCh38
rs995390554 Chr.16: 85283106-85283106 on Build GRCh38
rs187872931 Chr.16: 85282916-85282916 on Build GRCh38
rs948651266 Chr.16: 85283006-85283006 on Build GRCh38
rs968211281 Chr.16: 85283209-85283209 on Build GRCh38
rs184373979 Chr.16: 85283227-85283227 on Build GRCh38
rs879791758 Chr.16: 85283077-85283077 on Build GRCh38
rs1013141372 Chr.16: 85282785-85282785 on Build GRCh38
rs959310134 Chr.16: 85283170-85283170 on Build GRCh38
rs761676450 Chr.16: 85282861-85282861 on Build GRCh38
rs978164322 Chr.16: 85282948-85282948 on Build GRCh38
rs540562978 Chr.16: 85282968-85282968 on Build GRCh38
rs555689906 Chr.16: 85282820-85282820 on Build GRCh38
rs1054655390 Chr.16: 85283245-85283245 on Build GRCh38
rs923999803 Chr.16: 85283218-85283218 on Build GRCh38
rs1057090718 Chr.16: 85282997-85282997 on Build GRCh38
rs961751191 Chr.16: 85283086-85283086 on Build GRCh38
rs374103568 Chr.16: 85283054-85283054 on Build GRCh38
rs1029829246 Chr.16: 85282851-85282851 on Build GRCh38
rs55872904 Chr.16: 85283041-85283041 on Build GRCh38
rs1044311873 Chr.16: 85283256-85283256 on Build GRCh38
rs932780868 Chr.16: 85283264-85283264 on Build GRCh38
rs140547181 Chr.16: 85282824-85282824 on Build GRCh38
rs954467630 Chr.16: 85283089-85283089 on Build GRCh38
rs917570914 Chr.16: 85283012-85283012 on Build GRCh38
rs1015685187 Chr.16: 85283083-85283083 on Build GRCh38
rs1029273780 Chr.16: 85283065-85283065 on Build GRCh38
rs544662638 Chr.16: 85282829-85282829 on Build GRCh38
rs758890972 Chr.16: 85283057-85283057 on Build GRCh38
rs141893018 Chr.16: 85283194-85283194 on Build GRCh38
rs765635366 Chr.16: 85282927-85282927 on Build GRCh38
rs778140057 Chr.16: 85283205-85283205 on Build GRCh38
rs923945929 Chr.16: 85282951-85282951 on Build GRCh38
rs895706703 Chr.16: 85283005-85283005 on Build GRCh38
rs961018721 Chr.16: 85282833-85282833 on Build GRCh38
rs767162799 Chr.16: 85283051-85283051 on Build GRCh38
rs961237075 Chr.16: 85282915-85282915 on Build GRCh38
rs949028651 Chr.16: 85283015-85283015 on Build GRCh38
rs893361492 Chr.16: 85282796-85282796 on Build GRCh38
rs1016069147 Chr.16: 85282867-85282867 on Build GRCh38
rs1042157766 Chr.16: 85283020-85283020 on Build GRCh38
rs948683877 Chr.16: 85283248-85283248 on Build GRCh38
rs369369161 Chr.16: 85283029-85283029 on Build GRCh38
rs954251539 Chr.16: 85282884-85282884 on Build GRCh38
rs939295540 Chr.16: 85282969-85282969 on Build GRCh38
rs544299154 Chr.16: 85283257-85283257 on Build GRCh38
rs8046524 Chr.16: 85283211-85283211 on Build GRCh38
rs992188266 Chr.16: 85283239-85283239 on Build GRCh38
rs990710605 Chr.16: 85283175-85283175 on Build GRCh38
rs1044644464 Chr.16: 85283014-85283014 on Build GRCh38
rs145686364 Chr.16: 85282838-85282838 on Build GRCh38
rs1026756870 Chr.16: 85283132-85283132 on Build GRCh38
rs535848901 Chr.16: 85282813-85282813 on Build GRCh38
rs937927230 Chr.16: 85282949-85282949 on Build GRCh38
rs1028327704 Chr.16: 85282816-85282816 on Build GRCh38
rs946832797 Chr.16: 85282940-85282940 on Build GRCh38
rs559973909 Chr.16: 85283058-85283058 on Build GRCh38
rs916566803 Chr.16: 85283243-85283243 on Build GRCh38
rs562470693 Chr.16: 85283050-85283050 on Build GRCh38
rs970370617 Chr.16: 85283210-85283210 on Build GRCh38
rs371156738 Chr.16: 85283240-85283240 on Build GRCh38
rs915396131 Chr.16: 85282934-85282934 on Build GRCh38
rs2966857 Chr.16: 85283033-85283033 on Build GRCh38
rs150656362 Chr.16: 85283270-85283270 on Build GRCh38
rs923646799 Chr.16: 85283026-85283026 on Build GRCh38
rs531327739 Chr.16: 85283142-85283142 on Build GRCh38
rs761222734 Chr.16: 85282828-85282828 on Build GRCh38
rs934855522 Chr.16: 85283032-85283032 on Build GRCh38

View Full Product Details