Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43917110-43917614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM215A
Entrez Gene ID
23591
Gene Name
family with sequence similarity 215 member A (non-protein coding)
Gene Aliases
APR-2, C17orf88, LINC00530
UniGene
Hs.721804
Gene Chromosome Location
Chr.17: 43917208-43917987 on Build GRCh38
Transcripts
NR_026770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5957987 c.162C>T p.H54H Chr.17: 43917472-43917472 on Build GRCh38
6456334 c.144G>A p.A48A Chr.17: 43917454-43917454 on Build GRCh38
5423234 c.194G>A p.R65H Chr.17: 43917504-43917504 on Build GRCh38
5957794 c.70G>T p.G24C Chr.17: 43917380-43917380 on Build GRCh38
3387926 c.221G>A p.G74E Chr.17: 43917531-43917531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905046288 Chr.17: 43917460-43917460 on Build GRCh38
rs552852273 Chr.17: 43917442-43917442 on Build GRCh38
rs763824568 Chr.17: 43917533-43917533 on Build GRCh38
rs747482557 Chr.17: 43917278-43917278 on Build GRCh38
rs1032675795 Chr.17: 43917468-43917468 on Build GRCh38
rs931310868 Chr.17: 43917430-43917430 on Build GRCh38
rs773827253 Chr.17: 43917137-43917137 on Build GRCh38
rs559556788 Chr.17: 43917356-43917356 on Build GRCh38
rs1013465602 Chr.17: 43917230-43917230 on Build GRCh38
rs954452504 Chr.17: 43917304-43917304 on Build GRCh38
rs568040297 Chr.17: 43917558-43917558 on Build GRCh38
rs762449205 Chr.17: 43917525-43917525 on Build GRCh38
rs61455159 Chr.17: 43917328-43917328 on Build GRCh38
rs1002008675 Chr.17: 43917464-43917464 on Build GRCh38
rs539286879 Chr.17: 43917176-43917176 on Build GRCh38
rs1017282239 Chr.17: 43917552-43917552 on Build GRCh38
rs775099660 Chr.17: 43917488-43917488 on Build GRCh38
rs1002103233 Chr.17: 43917255-43917255 on Build GRCh38
rs766840445 Chr.17: 43917584-43917584 on Build GRCh38
rs1009678865 Chr.17: 43917494-43917494 on Build GRCh38
rs975571281 Chr.17: 43917577-43917577 on Build GRCh38
rs949435722 Chr.17: 43917456-43917456 on Build GRCh38
rs557524836 Chr.17: 43917268-43917268 on Build GRCh38
rs574800509 Chr.17: 43917418-43917418 on Build GRCh38
rs767784483 Chr.17: 43917203-43917203 on Build GRCh38
rs564673001 Chr.17: 43917487-43917487 on Build GRCh38
rs777064946 Chr.17: 43917343-43917343 on Build GRCh38
rs111396420 Chr.17: 43917365-43917365 on Build GRCh38
rs1008618490 Chr.17: 43917542-43917542 on Build GRCh38
rs887225202 Chr.17: 43917454-43917454 on Build GRCh38
rs528637129 Chr.17: 43917538-43917538 on Build GRCh38
rs113545721 Chr.17: 43917330-43917330 on Build GRCh38
rs772824017 Chr.17: 43917401-43917401 on Build GRCh38
rs572652542 Chr.17: 43917311-43917311 on Build GRCh38
rs1024807168 Chr.17: 43917243-43917243 on Build GRCh38
rs887162241 Chr.17: 43917225-43917226 on Build GRCh38
rs531222871 Chr.17: 43917439-43917439 on Build GRCh38
rs957950646 Chr.17: 43917266-43917266 on Build GRCh38
rs575176628 Chr.17: 43917162-43917162 on Build GRCh38
rs905139494 Chr.17: 43917252-43917252 on Build GRCh38
rs964190129 Chr.17: 43917554-43917554 on Build GRCh38
rs231458 Chr.17: 43917426-43917426 on Build GRCh38
rs1047599470 Chr.17: 43917207-43917207 on Build GRCh38
rs1032231587 Chr.17: 43917257-43917257 on Build GRCh38
rs746857297 Chr.17: 43917402-43917402 on Build GRCh38
rs764600446 Chr.17: 43917469-43917469 on Build GRCh38
rs1046288152 Chr.17: 43917458-43917458 on Build GRCh38
rs865998387 Chr.17: 43917355-43917355 on Build GRCh38
rs1047057242 Chr.17: 43917431-43917431 on Build GRCh38
rs770984547 Chr.17: 43917422-43917422 on Build GRCh38
rs527276788 Chr.17: 43917364-43917365 on Build GRCh38
rs1020603423 Chr.17: 43917512-43917512 on Build GRCh38
rs754388703 Chr.17: 43917447-43917447 on Build GRCh38
rs372600002 Chr.17: 43917541-43917541 on Build GRCh38
rs867029853 Chr.17: 43917587-43917587 on Build GRCh38
rs566549881 Chr.17: 43917292-43917292 on Build GRCh38
rs371125145 Chr.17: 43917341-43917341 on Build GRCh38
rs61231498 Chr.17: 43917424-43917424 on Build GRCh38

View Full Product Details