Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48724229-48724731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR3185
Entrez Gene ID
100422978
Gene Name
microRNA 3185
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.17: 48724408-48724475 on Build GRCh38
Transcripts
NR_036150.1 NM_001282275.1 NM_001282276.1 XM_006721865.3 XM_011524747.2 XM_011524748.1

Gene Symbol
PRAC2
Entrez Gene ID
360205
Gene Name
prostate cancer susceptibility candidate 2
Gene Aliases
C17orf93, HOXB-AS5, HOXB13-AS1, NCRNA00253
UniGene
Hs.236557
Gene Chromosome Location
Chr.17: 48720802-48724758 on Build GRCh38
Transcripts
NR_036150.1 NM_001282275.1 NM_001282276.1 XM_006721865.3 XM_011524747.2 XM_011524748.1

SNP Detail

SNP ID Chromosome Location
rs1018215451 Chr.17: 48724671-48724672 on Build GRCh38
rs544838647 Chr.17: 48724390-48724390 on Build GRCh38
rs1006547151 Chr.17: 48724664-48724664 on Build GRCh38
rs773085292 Chr.17: 48724445-48724445 on Build GRCh38
rs1056191395 Chr.17: 48724648-48724648 on Build GRCh38
rs551938680 Chr.17: 48724354-48724355 on Build GRCh38
rs774322029 Chr.17: 48724304-48724304 on Build GRCh38
rs550126263 Chr.17: 48724461-48724463 on Build GRCh38
rs909926409 Chr.17: 48724483-48724483 on Build GRCh38
rs976776672 Chr.17: 48724555-48724555 on Build GRCh38
rs968780923 Chr.17: 48724382-48724382 on Build GRCh38
rs56290537 Chr.17: 48724636-48724636 on Build GRCh38
rs926251338 Chr.17: 48724640-48724640 on Build GRCh38
rs1052576030 Chr.17: 48724630-48724630 on Build GRCh38
rs948343441 Chr.17: 48724359-48724359 on Build GRCh38
rs765341927 Chr.17: 48724545-48724545 on Build GRCh38
rs983019138 Chr.17: 48724576-48724576 on Build GRCh38
rs746067606 Chr.17: 48724355-48724356 on Build GRCh38
rs200748701 Chr.17: 48724399-48724399 on Build GRCh38
rs3110599 Chr.17: 48724362-48724362 on Build GRCh38
rs1018184442 Chr.17: 48724393-48724393 on Build GRCh38
rs941073665 Chr.17: 48724365-48724365 on Build GRCh38
rs895988170 Chr.17: 48724653-48724653 on Build GRCh38
rs781039600 Chr.17: 48724463-48724463 on Build GRCh38
rs761591102 Chr.17: 48724316-48724316 on Build GRCh38
rs199607933 Chr.17: 48724360-48724361 on Build GRCh38
rs1045118150 Chr.17: 48724366-48724366 on Build GRCh38
rs2271891 Chr.17: 48724646-48724646 on Build GRCh38
rs1010439597 Chr.17: 48724661-48724661 on Build GRCh38
rs556446830 Chr.17: 48724328-48724328 on Build GRCh38
rs941262668 Chr.17: 48724619-48724619 on Build GRCh38
rs1003273521 Chr.17: 48724421-48724421 on Build GRCh38
rs1020575828 Chr.17: 48724458-48724458 on Build GRCh38
rs1000231825 Chr.17: 48724469-48724469 on Build GRCh38
rs543910385 Chr.17: 48724325-48724325 on Build GRCh38
rs922448551 Chr.17: 48724562-48724562 on Build GRCh38
rs560532978 Chr.17: 48724609-48724609 on Build GRCh38
rs1010498767 Chr.17: 48724449-48724449 on Build GRCh38
rs575018161 Chr.17: 48724290-48724290 on Build GRCh38
rs527565410 Chr.17: 48724480-48724480 on Build GRCh38
rs1045288858 Chr.17: 48724360-48724360 on Build GRCh38
rs951179846 Chr.17: 48724567-48724567 on Build GRCh38
rs745592280 Chr.17: 48724369-48724370 on Build GRCh38
rs768519132 Chr.17: 48724260-48724260 on Build GRCh38
rs1042266957 Chr.17: 48724688-48724688 on Build GRCh38
rs757069417 Chr.17: 48724462-48724462 on Build GRCh38
rs1010319713 Chr.17: 48724369-48724369 on Build GRCh38
rs866683266 Chr.17: 48724448-48724448 on Build GRCh38
rs775484364 Chr.17: 48724435-48724435 on Build GRCh38
rs372079370 Chr.17: 48724362-48724362 on Build GRCh38
rs185214256 Chr.17: 48724493-48724493 on Build GRCh38
rs769491789 Chr.17: 48724516-48724516 on Build GRCh38
rs1046942638 Chr.17: 48724354-48724354 on Build GRCh38
rs369888513 Chr.17: 48724362-48724363 on Build GRCh38
rs942777600 Chr.17: 48724283-48724283 on Build GRCh38
rs374896479 Chr.17: 48724363-48724364 on Build GRCh38
rs1057337259 Chr.17: 48724441-48724441 on Build GRCh38
rs376980892 Chr.17: 48724459-48724459 on Build GRCh38
rs1045320287 Chr.17: 48724527-48724527 on Build GRCh38

View Full Product Details