Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26933204-26933647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP4-AS1
Entrez Gene ID
147429
Gene Name
AQP4 antisense RNA 1
Gene Aliases
C18orf16, CHST9-AS1
UniGene
Hs.559280
Gene Chromosome Location
Chr.18: 26865308-26935946 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_017026033.1 XM_017026034.1

Gene Symbol
CHST9
Entrez Gene ID
83539
Gene Name
carbohydrate sulfotransferase 9
Gene Aliases
GALNAC4ST-2, GalNAc4ST2
UniGene
Hs.44584
Gene Chromosome Location
Chr.18: 26908754-27185364 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_017026033.1 XM_017026034.1

SNP Detail

SNP ID Chromosome Location
rs571728240 Chr.18: 26933523-26933523 on Build GRCh38
rs557654986 Chr.18: 26933561-26933561 on Build GRCh38
rs948951645 Chr.18: 26933409-26933409 on Build GRCh38
rs925789213 Chr.18: 26933538-26933538 on Build GRCh38
rs539377260 Chr.18: 26933527-26933527 on Build GRCh38
rs190178733 Chr.18: 26933414-26933414 on Build GRCh38
rs934395541 Chr.18: 26933241-26933241 on Build GRCh38
rs440554 Chr.18: 26933615-26933615 on Build GRCh38
rs186783991 Chr.18: 26933608-26933608 on Build GRCh38
rs185804399 Chr.18: 26933247-26933247 on Build GRCh38
rs35376920 Chr.18: 26933377-26933377 on Build GRCh38
rs1029713610 Chr.18: 26933475-26933475 on Build GRCh38
rs1044634347 Chr.18: 26933270-26933270 on Build GRCh38
rs576636726 Chr.18: 26933597-26933597 on Build GRCh38
rs955367860 Chr.18: 26933484-26933484 on Build GRCh38
rs985452371 Chr.18: 26933489-26933489 on Build GRCh38
rs964607949 Chr.18: 26933524-26933524 on Build GRCh38
rs118177881 Chr.18: 26933244-26933244 on Build GRCh38
rs1005756963 Chr.18: 26933352-26933352 on Build GRCh38
rs949681124 Chr.18: 26933260-26933260 on Build GRCh38
rs534912785 Chr.18: 26933450-26933450 on Build GRCh38
rs551238279 Chr.18: 26933261-26933261 on Build GRCh38
rs182450686 Chr.18: 26933487-26933487 on Build GRCh38
rs978648485 Chr.18: 26933532-26933532 on Build GRCh38
rs1017575102 Chr.18: 26933520-26933520 on Build GRCh38
rs897266483 Chr.18: 26933447-26933447 on Build GRCh38
rs906115941 Chr.18: 26933338-26933338 on Build GRCh38
rs566188743 Chr.18: 26933292-26933292 on Build GRCh38
rs549087357 Chr.18: 26933391-26933391 on Build GRCh38
rs760362529 Chr.18: 26933305-26933305 on Build GRCh38
rs576131920 Chr.18: 26933577-26933577 on Build GRCh38
rs374751441 Chr.18: 26933623-26933623 on Build GRCh38
rs537012147 Chr.18: 26933336-26933336 on Build GRCh38
rs569748621 Chr.18: 26933294-26933294 on Build GRCh38
rs994678337 Chr.18: 26933452-26933452 on Build GRCh38

View Full Product Details