Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3150697-3151233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD5
Entrez Gene ID
60493
Gene Name
FAST kinase domains 5
Gene Aliases
dJ1187M17.5
UniGene
Hs.741168
Gene Chromosome Location
Chr.20: 3146519-3159886 on Build GRCh38
Transcripts
NM_021826.4 NM_001267584.1 NM_014948.3 NM_199415.2 NR_038395.1

Gene Symbol
UBOX5
Entrez Gene ID
22888
Gene Name
U-box domain containing 5
Gene Aliases
RNF37, UBCE7IP5, UIP5, hUIP5
UniGene
Hs.654646
Gene Chromosome Location
Chr.20: 3107573-3159910 on Build GRCh38
Transcripts
NM_021826.4 NM_001267584.1 NM_014948.3 NM_199415.2 NR_038395.1

Gene Symbol
UBOX5-AS1
Entrez Gene ID
100134015
Gene Name
UBOX5 antisense RNA 1
Gene Aliases
-
UniGene
Hs.129448
Gene Chromosome Location
Chr.20: 3106911-3150867 on Build GRCh38
Transcripts
NM_021826.4 NM_001267584.1 NM_014948.3 NM_199415.2 NR_038395.1

SNP Detail

SNP ID Chromosome Location
rs1031086132 Chr.20: 3151019-3151019 on Build GRCh38
rs1005605535 Chr.20: 3151118-3151118 on Build GRCh38
rs202236949 Chr.20: 3150878-3150878 on Build GRCh38
rs1028724944 Chr.20: 3151102-3151102 on Build GRCh38
rs191609790 Chr.20: 3150826-3150826 on Build GRCh38
rs41310183 Chr.20: 3150985-3150985 on Build GRCh38
rs372608459 Chr.20: 3151055-3151056 on Build GRCh38
rs941110505 Chr.20: 3150760-3150760 on Build GRCh38
rs985354839 Chr.20: 3151065-3151065 on Build GRCh38
rs113875306 Chr.20: 3150841-3150842 on Build GRCh38
rs550816229 Chr.20: 3151068-3151069 on Build GRCh38
rs1020256393 Chr.20: 3150756-3150756 on Build GRCh38
rs1041301810 Chr.20: 3151069-3151072 on Build GRCh38
rs190242647 Chr.20: 3151068-3151068 on Build GRCh38
rs532035083 Chr.20: 3151022-3151022 on Build GRCh38
rs73608130 Chr.20: 3150923-3150923 on Build GRCh38
rs965425242 Chr.20: 3150761-3150761 on Build GRCh38
rs573891086 Chr.20: 3150736-3150736 on Build GRCh38
rs1044444142 Chr.20: 3151034-3151034 on Build GRCh38
rs757321660 Chr.20: 3151208-3151208 on Build GRCh38
rs1038121545 Chr.20: 3151111-3151111 on Build GRCh38
rs896889701 Chr.20: 3151153-3151153 on Build GRCh38
rs866700365 Chr.20: 3150745-3150745 on Build GRCh38
rs911054386 Chr.20: 3151074-3151074 on Build GRCh38
rs151138009 Chr.20: 3151085-3151085 on Build GRCh38
rs1053984220 Chr.20: 3151057-3151060 on Build GRCh38
rs148795630 Chr.20: 3150947-3150947 on Build GRCh38
rs923319314 Chr.20: 3150896-3150896 on Build GRCh38
rs901386631 Chr.20: 3151079-3151079 on Build GRCh38
rs114562672 Chr.20: 3151057-3151057 on Build GRCh38
rs764287257 Chr.20: 3151051-3151051 on Build GRCh38
rs12625307 Chr.20: 3151187-3151187 on Build GRCh38
rs929671363 Chr.20: 3151158-3151158 on Build GRCh38
rs955254798 Chr.20: 3150903-3150903 on Build GRCh38
rs1012469064 Chr.20: 3150929-3150929 on Build GRCh38
rs979935215 Chr.20: 3151010-3151010 on Build GRCh38
rs948423467 Chr.20: 3150987-3150987 on Build GRCh38
rs559841352 Chr.20: 3150926-3150926 on Build GRCh38
rs144364047 Chr.20: 3150939-3150939 on Build GRCh38
rs1035256527 Chr.20: 3150843-3150843 on Build GRCh38
rs993037422 Chr.20: 3151182-3151182 on Build GRCh38
rs956732367 Chr.20: 3151040-3151040 on Build GRCh38
rs556144640 Chr.20: 3150790-3150790 on Build GRCh38
rs2422858 Chr.20: 3150842-3150842 on Build GRCh38
rs756089437 Chr.20: 3150805-3150805 on Build GRCh38
rs142154039 Chr.20: 3151069-3151070 on Build GRCh38
rs976192059 Chr.20: 3150882-3150882 on Build GRCh38
rs772959206 Chr.20: 3151070-3151071 on Build GRCh38
rs933863695 Chr.20: 3150728-3150728 on Build GRCh38
rs6107248 Chr.20: 3151161-3151161 on Build GRCh38
rs1028443126 Chr.20: 3150802-3150802 on Build GRCh38
rs751614338 Chr.20: 3151174-3151174 on Build GRCh38
rs889599102 Chr.20: 3150757-3150757 on Build GRCh38
rs933915192 Chr.20: 3151058-3151058 on Build GRCh38
rs952828027 Chr.20: 3151109-3151109 on Build GRCh38
rs748857440 Chr.20: 3150884-3150884 on Build GRCh38
rs543816444 Chr.20: 3150778-3150778 on Build GRCh38

View Full Product Details