Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 47166629-47167128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYA2
Entrez Gene ID
2139
Gene Name
EYA transcriptional coactivator and phosphatase 2
Gene Aliases
EAB1
UniGene
Hs.472877
Gene Chromosome Location
Chr.20: 46894624-47188844 on Build GRCh38
Transcripts
NM_005244.4 NM_172110.3 XM_005260327.2 XM_017027721.1 NR_037410.1

Gene Symbol
MIR3616
Entrez Gene ID
100500814
Gene Name
microRNA 3616
Gene Aliases
mir-3616
UniGene
Gene Chromosome Location
Chr.20: 47166967-47167058 on Build GRCh38
Transcripts
NM_005244.4 NM_172110.3 XM_005260327.2 XM_017027721.1 NR_037410.1

SNP Detail

SNP ID Chromosome Location
rs1050461193 Chr.20: 47166754-47166754 on Build GRCh38
rs919621523 Chr.20: 47166687-47166687 on Build GRCh38
rs184904477 Chr.20: 47167065-47167065 on Build GRCh38
rs921129436 Chr.20: 47166933-47166933 on Build GRCh38
rs948075172 Chr.20: 47166668-47166668 on Build GRCh38
rs932482686 Chr.20: 47166936-47166936 on Build GRCh38
rs1005886100 Chr.20: 47167053-47167053 on Build GRCh38
rs751652021 Chr.20: 47167048-47167048 on Build GRCh38
rs59858403 Chr.20: 47166657-47166657 on Build GRCh38
rs374378744 Chr.20: 47166775-47166775 on Build GRCh38
rs908178636 Chr.20: 47166913-47166913 on Build GRCh38
rs932095258 Chr.20: 47166743-47166743 on Build GRCh38
rs992793670 Chr.20: 47167071-47167071 on Build GRCh38
rs897058249 Chr.20: 47166935-47166935 on Build GRCh38
rs60011920 Chr.20: 47167082-47167082 on Build GRCh38
rs1041293993 Chr.20: 47166728-47166728 on Build GRCh38
rs760379897 Chr.20: 47166890-47166890 on Build GRCh38
rs1046675950 Chr.20: 47167005-47167005 on Build GRCh38
rs376097316 Chr.20: 47167066-47167066 on Build GRCh38
rs138356203 Chr.20: 47166922-47166922 on Build GRCh38
rs140512735 Chr.20: 47166722-47166722 on Build GRCh38
rs897026909 Chr.20: 47166730-47166730 on Build GRCh38
rs115089277 Chr.20: 47167105-47167105 on Build GRCh38
rs980552167 Chr.20: 47166875-47166875 on Build GRCh38
rs979720862 Chr.20: 47166854-47166854 on Build GRCh38
rs35800994 Chr.20: 47166721-47166721 on Build GRCh38
rs953927914 Chr.20: 47167106-47167106 on Build GRCh38
rs994001309 Chr.20: 47166738-47166738 on Build GRCh38
rs192179398 Chr.20: 47166976-47166976 on Build GRCh38
rs1883885 Chr.20: 47166814-47166814 on Build GRCh38
rs78302267 Chr.20: 47166886-47166886 on Build GRCh38
rs899656570 Chr.20: 47166733-47166733 on Build GRCh38
rs969534048 Chr.20: 47166869-47166869 on Build GRCh38
rs765803807 Chr.20: 47167004-47167004 on Build GRCh38
rs75118991 Chr.20: 47166882-47166882 on Build GRCh38
rs570230378 Chr.20: 47166701-47166701 on Build GRCh38
rs1007022566 Chr.20: 47166774-47166774 on Build GRCh38
rs753430018 Chr.20: 47166982-47166982 on Build GRCh38
rs34935499 Chr.20: 47166694-47166694 on Build GRCh38
rs1042829153 Chr.20: 47166963-47166963 on Build GRCh38
rs924183034 Chr.20: 47166855-47166855 on Build GRCh38
rs930119990 Chr.20: 47166706-47166706 on Build GRCh38
rs750749182 Chr.20: 47167046-47167046 on Build GRCh38
rs34703594 Chr.20: 47166710-47166710 on Build GRCh38
rs749145098 Chr.20: 47166781-47166781 on Build GRCh38
rs532889629 Chr.20: 47167049-47167049 on Build GRCh38
rs78308190 Chr.20: 47167068-47167068 on Build GRCh38
rs115633564 Chr.20: 47166877-47166877 on Build GRCh38
rs980863042 Chr.20: 47167103-47167103 on Build GRCh38
rs757122944 Chr.20: 47167097-47167097 on Build GRCh38
rs145622462 Chr.20: 47166795-47166795 on Build GRCh38
rs992322925 Chr.20: 47166868-47166868 on Build GRCh38
rs956987853 Chr.20: 47167069-47167069 on Build GRCh38
rs1037746878 Chr.20: 47167061-47167061 on Build GRCh38
rs546005671 Chr.20: 47166824-47166824 on Build GRCh38
rs757683299 Chr.20: 47166998-47166998 on Build GRCh38
rs943976212 Chr.20: 47166918-47166918 on Build GRCh38
rs957123323 Chr.20: 47166865-47166865 on Build GRCh38
rs3091782 Chr.20: 47167102-47167102 on Build GRCh38
rs79480878 Chr.20: 47166831-47166831 on Build GRCh38
rs576379844 Chr.20: 47166997-47166997 on Build GRCh38
rs896557548 Chr.20: 47166667-47166667 on Build GRCh38
rs35757245 Chr.20: 47166773-47166773 on Build GRCh38
rs557286910 Chr.20: 47167020-47167020 on Build GRCh38

View Full Product Details