Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19047349-19047854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR11
Entrez Gene ID
25786
Gene Name
DiGeorge syndrome critical region gene 11 (non-protein coding)
Gene Aliases
DGS-D
UniGene
Hs.720269
Gene Chromosome Location
Chr.22: 19046162-19048375 on Build GRCh38
Transcripts
NR_024157.1 NM_001173533.1 NM_001173534.1 NM_001184781.1 NM_005137.2 NR_033674.1

Gene Symbol
DGCR2
Entrez Gene ID
9993
Gene Name
DiGeorge syndrome critical region gene 2
Gene Aliases
DGS-C, IDD, LAN, SEZ-12
UniGene
Hs.517357
Gene Chromosome Location
Chr.22: 19036282-19122454 on Build GRCh38
Transcripts
NR_024157.1 NM_001173533.1 NM_001173534.1 NM_001184781.1 NM_005137.2 NR_033674.1

SNP Detail

SNP ID Chromosome Location
rs201824496 Chr.22: 19047548-19047548 on Build GRCh38
rs775336253 Chr.22: 19047550-19047550 on Build GRCh38
rs748138512 Chr.22: 19047676-19047676 on Build GRCh38
rs989874475 Chr.22: 19047779-19047779 on Build GRCh38
rs999705973 Chr.22: 19047740-19047740 on Build GRCh38
rs576901697 Chr.22: 19047661-19047661 on Build GRCh38
rs372803623 Chr.22: 19047652-19047652 on Build GRCh38
rs911946538 Chr.22: 19047557-19047557 on Build GRCh38
rs760954089 Chr.22: 19047470-19047470 on Build GRCh38
rs948188551 Chr.22: 19047518-19047518 on Build GRCh38
rs545768794 Chr.22: 19047687-19047687 on Build GRCh38
rs774542940 Chr.22: 19047554-19047554 on Build GRCh38
rs1023314046 Chr.22: 19047491-19047491 on Build GRCh38
rs930901943 Chr.22: 19047676-19047676 on Build GRCh38
rs190196439 Chr.22: 19047822-19047822 on Build GRCh38
rs904475269 Chr.22: 19047829-19047829 on Build GRCh38
rs1031233046 Chr.22: 19047765-19047765 on Build GRCh38
rs968625976 Chr.22: 19047493-19047493 on Build GRCh38
rs34515606 Chr.22: 19047715-19047716 on Build GRCh38
rs982344103 Chr.22: 19047526-19047526 on Build GRCh38
rs17810649 Chr.22: 19047559-19047559 on Build GRCh38
rs200733342 Chr.22: 19047626-19047627 on Build GRCh38
rs927839175 Chr.22: 19047533-19047533 on Build GRCh38
rs527453573 Chr.22: 19047534-19047534 on Build GRCh38
rs535756456 Chr.22: 19047412-19047412 on Build GRCh38
rs62221693 Chr.22: 19047473-19047473 on Build GRCh38
rs115917550 Chr.22: 19047556-19047556 on Build GRCh38
rs1013168250 Chr.22: 19047785-19047785 on Build GRCh38
rs775534814 Chr.22: 19047376-19047376 on Build GRCh38
rs939645540 Chr.22: 19047760-19047760 on Build GRCh38
rs754955224 Chr.22: 19047796-19047796 on Build GRCh38
rs1039398106 Chr.22: 19047635-19047635 on Build GRCh38
rs1022797402 Chr.22: 19047821-19047821 on Build GRCh38
rs890593711 Chr.22: 19047702-19047702 on Build GRCh38
rs958297265 Chr.22: 19047778-19047778 on Build GRCh38
rs569854016 Chr.22: 19047622-19047622 on Build GRCh38
rs760828752 Chr.22: 19047641-19047641 on Build GRCh38
rs556204485 Chr.22: 19047783-19047783 on Build GRCh38
rs979210719 Chr.22: 19047514-19047514 on Build GRCh38
rs916633353 Chr.22: 19047482-19047482 on Build GRCh38
rs941001366 Chr.22: 19047597-19047597 on Build GRCh38
rs766233399 Chr.22: 19047604-19047604 on Build GRCh38
rs569970496 Chr.22: 19047404-19047404 on Build GRCh38
rs899569551 Chr.22: 19047646-19047646 on Build GRCh38
rs1035544751 Chr.22: 19047437-19047437 on Build GRCh38
rs752675567 Chr.22: 19047576-19047576 on Build GRCh38
rs1048117958 Chr.22: 19047696-19047696 on Build GRCh38

View Full Product Details