Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19047862-19048364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR11
Entrez Gene ID
25786
Gene Name
DiGeorge syndrome critical region gene 11 (non-protein coding)
Gene Aliases
DGS-D
UniGene
Hs.720269
Gene Chromosome Location
Chr.22: 19046162-19048375 on Build GRCh38
Transcripts
NR_024157.1 NM_001173533.1 NM_001173534.1 NM_001184781.1 NM_005137.2 NR_033674.1

Gene Symbol
DGCR2
Entrez Gene ID
9993
Gene Name
DiGeorge syndrome critical region gene 2
Gene Aliases
DGS-C, IDD, LAN, SEZ-12
UniGene
Hs.517357
Gene Chromosome Location
Chr.22: 19036282-19122454 on Build GRCh38
Transcripts
NR_024157.1 NM_001173533.1 NM_001173534.1 NM_001184781.1 NM_005137.2 NR_033674.1

SNP Detail

SNP ID Chromosome Location
rs777666438 Chr.22: 19048227-19048227 on Build GRCh38
rs558333737 Chr.22: 19048020-19048020 on Build GRCh38
rs549903481 Chr.22: 19047940-19047940 on Build GRCh38
rs948500897 Chr.22: 19048330-19048330 on Build GRCh38
rs1039771311 Chr.22: 19047923-19047923 on Build GRCh38
rs935837832 Chr.22: 19048335-19048335 on Build GRCh38
rs41277590 Chr.22: 19047920-19047920 on Build GRCh38
rs969380885 Chr.22: 19048030-19048030 on Build GRCh38
rs529287149 Chr.22: 19048251-19048251 on Build GRCh38
rs576135991 Chr.22: 19048066-19048066 on Build GRCh38
rs891672896 Chr.22: 19048155-19048155 on Build GRCh38
rs758085307 Chr.22: 19048182-19048182 on Build GRCh38
rs774733434 Chr.22: 19048339-19048339 on Build GRCh38
rs571889839 Chr.22: 19048329-19048329 on Build GRCh38
rs962947116 Chr.22: 19048136-19048136 on Build GRCh38
rs549443230 Chr.22: 19047975-19047975 on Build GRCh38
rs538484169 Chr.22: 19048125-19048126 on Build GRCh38
rs528791293 Chr.22: 19048311-19048311 on Build GRCh38
rs148800476 Chr.22: 19048319-19048319 on Build GRCh38
rs541505326 Chr.22: 19047887-19047887 on Build GRCh38
rs904338880 Chr.22: 19048079-19048079 on Build GRCh38
rs138189232 Chr.22: 19047963-19047963 on Build GRCh38
rs1003513156 Chr.22: 19048056-19048056 on Build GRCh38
rs113010545 Chr.22: 19048156-19048156 on Build GRCh38
rs982569222 Chr.22: 19048115-19048115 on Build GRCh38
rs572202999 Chr.22: 19048107-19048107 on Build GRCh38
rs111958116 Chr.22: 19048011-19048011 on Build GRCh38
rs909755261 Chr.22: 19048124-19048124 on Build GRCh38
rs539553276 Chr.22: 19048043-19048043 on Build GRCh38
rs577683751 Chr.22: 19048201-19048201 on Build GRCh38
rs902951476 Chr.22: 19047976-19047976 on Build GRCh38
rs933646072 Chr.22: 19048194-19048194 on Build GRCh38
rs930913255 Chr.22: 19047945-19047945 on Build GRCh38
rs533257244 Chr.22: 19047917-19047917 on Build GRCh38
rs190711268 Chr.22: 19048261-19048261 on Build GRCh38
rs149567882 Chr.22: 19048022-19048022 on Build GRCh38
rs893934549 Chr.22: 19048328-19048328 on Build GRCh38
rs114683635 Chr.22: 19048003-19048003 on Build GRCh38
rs62221712 Chr.22: 19048210-19048210 on Build GRCh38
rs564470110 Chr.22: 19047894-19047894 on Build GRCh38
rs1018938538 Chr.22: 19048178-19048178 on Build GRCh38
rs768215331 Chr.22: 19048325-19048325 on Build GRCh38
rs929784045 Chr.22: 19047896-19047896 on Build GRCh38
rs1011196424 Chr.22: 19048021-19048021 on Build GRCh38
rs1056778890 Chr.22: 19048008-19048008 on Build GRCh38
rs529597142 Chr.22: 19048068-19048068 on Build GRCh38
rs141599309 Chr.22: 19048331-19048331 on Build GRCh38
rs529519320 Chr.22: 19047971-19047971 on Build GRCh38
rs556165870 Chr.22: 19048065-19048065 on Build GRCh38
rs144197578 Chr.22: 19048101-19048101 on Build GRCh38
rs1008890194 Chr.22: 19048158-19048158 on Build GRCh38
rs563325640 Chr.22: 19048213-19048213 on Build GRCh38
rs2240102 Chr.22: 19048184-19048184 on Build GRCh38
rs999673525 Chr.22: 19047983-19047983 on Build GRCh38
rs899537668 Chr.22: 19047956-19047956 on Build GRCh38
rs375779172 Chr.22: 19048157-19048157 on Build GRCh38
rs73384864 Chr.22: 19048333-19048333 on Build GRCh38
rs112310120 Chr.22: 19048106-19048106 on Build GRCh38
rs559747170 Chr.22: 19048263-19048263 on Build GRCh38
rs753882876 Chr.22: 19047919-19047919 on Build GRCh38

View Full Product Details