Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24429747-24430258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA2A
Entrez Gene ID
135
Gene Name
adenosine A2a receptor
Gene Aliases
A2aR, ADORA2, RDC8
UniGene
Hs.197029
Gene Chromosome Location
Chr.22: 24423597-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

Gene Symbol
ADORA2A-AS1
Entrez Gene ID
646023
Gene Name
ADORA2A antisense RNA 1
Gene Aliases
C22orf45
UniGene
Hs.632761
Gene Chromosome Location
Chr.22: 24429206-24495074 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

SNP Detail

SNP ID Chromosome Location
rs1050471062 Chr.22: 24429812-24429812 on Build GRCh38
rs954592040 Chr.22: 24429888-24429888 on Build GRCh38
rs908710799 Chr.22: 24430140-24430140 on Build GRCh38
rs5760414 Chr.22: 24430117-24430117 on Build GRCh38
rs1002849009 Chr.22: 24429826-24429826 on Build GRCh38
rs923201912 Chr.22: 24430214-24430214 on Build GRCh38
rs758338639 Chr.22: 24430136-24430136 on Build GRCh38
rs1035952106 Chr.22: 24430098-24430098 on Build GRCh38
rs554335769 Chr.22: 24429909-24429909 on Build GRCh38
rs527618057 Chr.22: 24430076-24430076 on Build GRCh38
rs764578616 Chr.22: 24429852-24429852 on Build GRCh38
rs78431322 Chr.22: 24430118-24430118 on Build GRCh38
rs542891593 Chr.22: 24430092-24430092 on Build GRCh38
rs532636169 Chr.22: 24430160-24430160 on Build GRCh38
rs991874953 Chr.22: 24429896-24429896 on Build GRCh38
rs878903204 Chr.22: 24429842-24429842 on Build GRCh38
rs539710373 Chr.22: 24430093-24430093 on Build GRCh38
rs931461103 Chr.22: 24429780-24429780 on Build GRCh38
rs960198282 Chr.22: 24429889-24429889 on Build GRCh38
rs904582877 Chr.22: 24430013-24430013 on Build GRCh38
rs549827257 Chr.22: 24429820-24429820 on Build GRCh38
rs968963606 Chr.22: 24429863-24429863 on Build GRCh38
rs538817565 Chr.22: 24429907-24429907 on Build GRCh38
rs969529804 Chr.22: 24430121-24430121 on Build GRCh38
rs1057322663 Chr.22: 24430154-24430154 on Build GRCh38
rs78784622 Chr.22: 24429777-24429777 on Build GRCh38
rs1047771767 Chr.22: 24429783-24429783 on Build GRCh38
rs889215638 Chr.22: 24429971-24429971 on Build GRCh38
rs368967970 Chr.22: 24429925-24429925 on Build GRCh38
rs964383946 Chr.22: 24430096-24430096 on Build GRCh38
rs987982065 Chr.22: 24430213-24430213 on Build GRCh38
rs761957244 Chr.22: 24429995-24429996 on Build GRCh38
rs191944535 Chr.22: 24429932-24429932 on Build GRCh38
rs556249179 Chr.22: 24429947-24429947 on Build GRCh38
rs187042606 Chr.22: 24429928-24429928 on Build GRCh38
rs752116557 Chr.22: 24429893-24429893 on Build GRCh38
rs1006646311 Chr.22: 24429846-24429846 on Build GRCh38
rs555439096 Chr.22: 24429922-24429922 on Build GRCh38
rs1035860134 Chr.22: 24429859-24429859 on Build GRCh38
rs979238771 Chr.22: 24430172-24430172 on Build GRCh38
rs190339124 Chr.22: 24430041-24430041 on Build GRCh38
rs920325621 Chr.22: 24429902-24429902 on Build GRCh38
rs572697380 Chr.22: 24429916-24429916 on Build GRCh38
rs938810908 Chr.22: 24430149-24430153 on Build GRCh38
rs557070190 Chr.22: 24429857-24429857 on Build GRCh38
rs767886086 Chr.22: 24430086-24430087 on Build GRCh38
rs567487708 Chr.22: 24430085-24430085 on Build GRCh38
rs962596767 Chr.22: 24429853-24429853 on Build GRCh38
rs755307187 Chr.22: 24430199-24430199 on Build GRCh38
rs897867509 Chr.22: 24430077-24430077 on Build GRCh38
rs149170930 Chr.22: 24429788-24429788 on Build GRCh38
rs973065112 Chr.22: 24430112-24430112 on Build GRCh38
rs895960158 Chr.22: 24430108-24430108 on Build GRCh38
rs143265985 Chr.22: 24429806-24429806 on Build GRCh38
rs577919109 Chr.22: 24429970-24429970 on Build GRCh38
rs547703343 Chr.22: 24430177-24430177 on Build GRCh38
rs762320152 Chr.22: 24429908-24429908 on Build GRCh38
rs185254367 Chr.22: 24430040-24430040 on Build GRCh38
rs3761420 Chr.22: 24430187-24430187 on Build GRCh38
rs1050439937 Chr.22: 24429960-24429960 on Build GRCh38
rs973566339 Chr.22: 24429895-24429895 on Build GRCh38
rs977251998 Chr.22: 24429866-24429866 on Build GRCh38
rs750543742 Chr.22: 24430066-24430066 on Build GRCh38
rs571528247 Chr.22: 24429855-24429855 on Build GRCh38
rs536929793 Chr.22: 24429921-24429921 on Build GRCh38
rs950496754 Chr.22: 24430046-24430046 on Build GRCh38
rs1032024124 Chr.22: 24429882-24429882 on Build GRCh38
rs560961776 Chr.22: 24430111-24430111 on Build GRCh38
rs1035639533 Chr.22: 24429854-24429854 on Build GRCh38
rs780276327 Chr.22: 24430153-24430153 on Build GRCh38

View Full Product Details