Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49624156-49624653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf34
Entrez Gene ID
348645
Gene Name
chromosome 22 open reading frame 34
Gene Aliases
-
UniGene
Hs.133159
Gene Chromosome Location
Chr.22: 49414524-49657542 on Build GRCh38
Transcripts
NM_001289922.2 NR_110522.1 NR_110523.1 XM_011530167.2 XM_017028796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5498435 c.449C>T p.A150V Chr.22: 49624364-49624364 on Build GRCh38
5550518 c.228G>A p.A76A Chr.22: 49624585-49624585 on Build GRCh38
4137516 c.405G>A p.R135R Chr.22: 49624408-49624408 on Build GRCh38
5601148 c.167G>A p.R56Q Chr.22: 49624646-49624646 on Build GRCh38
249000 c.303G>T p.R101S Chr.22: 49624510-49624510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764225330 Chr.22: 49624530-49624530 on Build GRCh38
rs529011765 Chr.22: 49624479-49624479 on Build GRCh38
rs191345855 Chr.22: 49624477-49624477 on Build GRCh38
rs573128517 Chr.22: 49624253-49624253 on Build GRCh38
rs771194102 Chr.22: 49624474-49624474 on Build GRCh38
rs988292222 Chr.22: 49624622-49624622 on Build GRCh38
rs911418998 Chr.22: 49624411-49624411 on Build GRCh38
rs978927225 Chr.22: 49624322-49624322 on Build GRCh38
rs770690001 Chr.22: 49624438-49624438 on Build GRCh38
rs139374240 Chr.22: 49624394-49624394 on Build GRCh38
rs779461186 Chr.22: 49624309-49624317 on Build GRCh38
rs1007350196 Chr.22: 49624598-49624598 on Build GRCh38
rs777349224 Chr.22: 49624413-49624413 on Build GRCh38
rs781021421 Chr.22: 49624392-49624392 on Build GRCh38
rs754982060 Chr.22: 49624379-49624379 on Build GRCh38
rs1005382540 Chr.22: 49624507-49624507 on Build GRCh38
rs776028129 Chr.22: 49624319-49624319 on Build GRCh38
rs1023392206 Chr.22: 49624190-49624190 on Build GRCh38
rs529475708 Chr.22: 49624543-49624543 on Build GRCh38
rs942874043 Chr.22: 49624436-49624436 on Build GRCh38
rs746645198 Chr.22: 49624542-49624542 on Build GRCh38
rs1034938104 Chr.22: 49624248-49624248 on Build GRCh38
rs960951411 Chr.22: 49624210-49624210 on Build GRCh38
rs767511696 Chr.22: 49624615-49624615 on Build GRCh38
rs1005023499 Chr.22: 49624202-49624202 on Build GRCh38
rs200839218 Chr.22: 49624185-49624185 on Build GRCh38
rs1009242796 Chr.22: 49624583-49624583 on Build GRCh38
rs1003911564 Chr.22: 49624222-49624222 on Build GRCh38
rs2011097 Chr.22: 49624421-49624421 on Build GRCh38
rs1015517241 Chr.22: 49624207-49624207 on Build GRCh38
rs147509947 Chr.22: 49624211-49624211 on Build GRCh38
rs550387633 Chr.22: 49624539-49624539 on Build GRCh38
rs555237638 Chr.22: 49624340-49624340 on Build GRCh38
rs774794208 Chr.22: 49624268-49624268 on Build GRCh38
rs144373525 Chr.22: 49624443-49624443 on Build GRCh38
rs760174138 Chr.22: 49624282-49624282 on Build GRCh38
rs1022065315 Chr.22: 49624283-49624283 on Build GRCh38
rs1047022612 Chr.22: 49624463-49624463 on Build GRCh38
rs544361453 Chr.22: 49624363-49624363 on Build GRCh38
rs969644949 Chr.22: 49624586-49624586 on Build GRCh38
rs568847702 Chr.22: 49624197-49624197 on Build GRCh38
rs1000502813 Chr.22: 49624588-49624588 on Build GRCh38
rs766220311 Chr.22: 49624358-49624358 on Build GRCh38
rs558082455 Chr.22: 49624244-49624244 on Build GRCh38
rs922860839 Chr.22: 49624446-49624446 on Build GRCh38
rs892353628 Chr.22: 49624564-49624564 on Build GRCh38
rs747572264 Chr.22: 49624189-49624189 on Build GRCh38
rs769109198 Chr.22: 49624176-49624213 on Build GRCh38
rs1023929815 Chr.22: 49624213-49624213 on Build GRCh38
rs551357078 Chr.22: 49624630-49624630 on Build GRCh38
rs754814576 Chr.22: 49624175-49624175 on Build GRCh38
rs776679239 Chr.22: 49624538-49624538 on Build GRCh38
rs568910293 Chr.22: 49624590-49624590 on Build GRCh38
rs759815227 Chr.22: 49624364-49624364 on Build GRCh38
rs150058296 Chr.22: 49624439-49624439 on Build GRCh38
rs1023872848 Chr.22: 49624585-49624585 on Build GRCh38
rs909980020 Chr.22: 49624465-49624465 on Build GRCh38
rs574282843 Chr.22: 49624614-49624614 on Build GRCh38
rs969028105 Chr.22: 49624228-49624228 on Build GRCh38
rs373882447 Chr.22: 49624295-49624295 on Build GRCh38
rs533986973 Chr.22: 49624320-49624320 on Build GRCh38
rs186878080 Chr.22: 49624344-49624344 on Build GRCh38
rs987779393 Chr.22: 49624274-49624274 on Build GRCh38
rs929998553 Chr.22: 49624461-49624461 on Build GRCh38
rs967755130 Chr.22: 49624311-49624311 on Build GRCh38
rs767882880 Chr.22: 49624310-49624310 on Build GRCh38
rs761179325 Chr.22: 49624511-49624511 on Build GRCh38
rs763128 Chr.22: 49624397-49624397 on Build GRCh38
rs545339071 Chr.22: 49624404-49624404 on Build GRCh38

View Full Product Details