Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185717827-185718332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2BP2
Entrez Gene ID
10644
Gene Name
insulin like growth factor 2 mRNA binding protein 2
Gene Aliases
IMP-2, IMP2, VICKZ2
UniGene
Hs.35354
Gene Chromosome Location
Chr.3: 185643739-185825051 on Build GRCh38
Transcripts
NM_001007225.1 NM_001291869.1 NM_001291872.1 NM_001291873.1 NM_001291874.1 NM_001291875.1 NM_006548.4 XM_017005557.1 XM_017005558.1 XM_017005559.1 XM_017005560.1 XM_017005561.1 XM_017005562.1 XM_017005563.1 XM_017005564.1 NR_126326.1

Gene Symbol
IGF2BP2-AS1
Entrez Gene ID
646600
Gene Name
IGF2BP2 antisense RNA 1
Gene Aliases
C3orf65
UniGene
Hs.647949
Gene Chromosome Location
Chr.3: 185713252-185729787 on Build GRCh38
Transcripts
NM_001007225.1 NM_001291869.1 NM_001291872.1 NM_001291873.1 NM_001291874.1 NM_001291875.1 NM_006548.4 XM_017005557.1 XM_017005558.1 XM_017005559.1 XM_017005560.1 XM_017005561.1 XM_017005562.1 XM_017005563.1 XM_017005564.1 NR_126326.1

SNP Detail

SNP ID Chromosome Location
rs954477222 Chr.3: 185718063-185718063 on Build GRCh38
rs1056452639 Chr.3: 185718126-185718126 on Build GRCh38
rs1056986496 Chr.3: 185717945-185717945 on Build GRCh38
rs967545468 Chr.3: 185718302-185718302 on Build GRCh38
rs1038605754 Chr.3: 185717908-185717908 on Build GRCh38
rs985877944 Chr.3: 185718274-185718274 on Build GRCh38
rs1005539250 Chr.3: 185718004-185718004 on Build GRCh38
rs572829070 Chr.3: 185718122-185718122 on Build GRCh38
rs574996030 Chr.3: 185718088-185718088 on Build GRCh38
rs1049903197 Chr.3: 185717880-185717880 on Build GRCh38
rs897149131 Chr.3: 185717911-185717911 on Build GRCh38
rs570266555 Chr.3: 185718129-185718129 on Build GRCh38
rs941604033 Chr.3: 185717900-185717900 on Build GRCh38
rs1012301614 Chr.3: 185718158-185718158 on Build GRCh38
rs142420514 Chr.3: 185718009-185718009 on Build GRCh38
rs1031374565 Chr.3: 185718208-185718208 on Build GRCh38
rs539824175 Chr.3: 185718160-185718160 on Build GRCh38
rs923035932 Chr.3: 185718075-185718075 on Build GRCh38
rs896594258 Chr.3: 185718128-185718128 on Build GRCh38
rs956583001 Chr.3: 185718050-185718050 on Build GRCh38
rs547308018 Chr.3: 185718001-185718001 on Build GRCh38
rs377255775 Chr.3: 185717893-185717893 on Build GRCh38
rs941468234 Chr.3: 185718108-185718108 on Build GRCh38
rs911505298 Chr.3: 185717882-185717882 on Build GRCh38
rs896568782 Chr.3: 185717983-185717983 on Build GRCh38
rs369240004 Chr.3: 185717974-185717974 on Build GRCh38
rs985848450 Chr.3: 185718105-185718105 on Build GRCh38
rs903699409 Chr.3: 185718182-185718182 on Build GRCh38
rs557557570 Chr.3: 185718114-185718114 on Build GRCh38
rs929777112 Chr.3: 185718155-185718155 on Build GRCh38
rs1038639074 Chr.3: 185718110-185718110 on Build GRCh38
rs150257180 Chr.3: 185718057-185718057 on Build GRCh38
rs773475102 Chr.3: 185718095-185718095 on Build GRCh38
rs1006004252 Chr.3: 185718228-185718228 on Build GRCh38
rs1000756828 Chr.3: 185718044-185718044 on Build GRCh38
rs952998792 Chr.3: 185718269-185718269 on Build GRCh38
rs977789475 Chr.3: 185718077-185718077 on Build GRCh38
rs964324131 Chr.3: 185718016-185718016 on Build GRCh38
rs761866890 Chr.3: 185717934-185717934 on Build GRCh38
rs1015985536 Chr.3: 185718286-185718286 on Build GRCh38
rs956630439 Chr.3: 185718215-185718215 on Build GRCh38
rs149015607 Chr.3: 185718115-185718115 on Build GRCh38
rs975769517 Chr.3: 185718071-185718071 on Build GRCh38
rs754915108 Chr.3: 185718284-185718285 on Build GRCh38
rs535676841 Chr.3: 185718049-185718049 on Build GRCh38
rs776530920 Chr.3: 185718010-185718010 on Build GRCh38
rs962904529 Chr.3: 185718295-185718295 on Build GRCh38
rs539566184 Chr.3: 185718106-185718106 on Build GRCh38
rs867819390 Chr.3: 185718180-185718180 on Build GRCh38
rs1045104637 Chr.3: 185718164-185718164 on Build GRCh38
rs143799522 Chr.3: 185718267-185718267 on Build GRCh38
rs888469792 Chr.3: 185717973-185717973 on Build GRCh38
rs565386894 Chr.3: 185718230-185718231 on Build GRCh38
rs112941145 Chr.3: 185717891-185717891 on Build GRCh38
rs573948771 Chr.3: 185718216-185718216 on Build GRCh38
rs561984530 Chr.3: 185718197-185718197 on Build GRCh38
rs184477102 Chr.3: 185717857-185717857 on Build GRCh38
rs1012061082 Chr.3: 185717995-185717995 on Build GRCh38
rs562626494 Chr.3: 185718297-185718297 on Build GRCh38

View Full Product Details