Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 112161337-112161883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L4A
Entrez Gene ID
64097
Gene Name
erythrocyte membrane protein band 4.1 like 4A
Gene Aliases
EPB41L4, NBL4
UniGene
Hs.584954
Gene Chromosome Location
Chr.5: 112141829-112419490 on Build GRCh38
Transcripts
XM_011543530.1 XM_011543531.2 XM_011543532.1 XM_011543533.1 NR_015370.2 NR_002922.1

Gene Symbol
EPB41L4A-AS1
Entrez Gene ID
114915
Gene Name
EPB41L4A antisense RNA 1
Gene Aliases
C5orf26, NCRNA00219, TIGA1
UniGene
Hs.12082
Gene Chromosome Location
Chr.5: 112160526-112162501 on Build GRCh38
Transcripts
XM_011543530.1 XM_011543531.2 XM_011543532.1 XM_011543533.1 NR_015370.2 NR_002922.1

Gene Symbol
SNORA13
Entrez Gene ID
654322
Gene Name
small nucleolar RNA, H/ACA box 13
Gene Aliases
ACA13
UniGene
Hs.689709
Gene Chromosome Location
Chr.5: 112161485-112161617 on Build GRCh38
Transcripts
XM_011543530.1 XM_011543531.2 XM_011543532.1 XM_011543533.1 NR_015370.2 NR_002922.1

SNP Detail

SNP ID Chromosome Location
rs747336661 Chr.5: 112161483-112161502 on Build GRCh38
rs151252280 Chr.5: 112161444-112161444 on Build GRCh38
rs201112618 Chr.5: 112161611-112161611 on Build GRCh38
rs1018158248 Chr.5: 112161371-112161371 on Build GRCh38
rs532067110 Chr.5: 112161516-112161516 on Build GRCh38
rs772986918 Chr.5: 112161593-112161593 on Build GRCh38
rs751656380 Chr.5: 112161392-112161392 on Build GRCh38
rs1054950606 Chr.5: 112161800-112161800 on Build GRCh38
rs747201350 Chr.5: 112161451-112161451 on Build GRCh38
rs903191521 Chr.5: 112161812-112161813 on Build GRCh38
rs768998969 Chr.5: 112161555-112161555 on Build GRCh38
rs2290991 Chr.5: 112161436-112161436 on Build GRCh38
rs373134452 Chr.5: 112161613-112161613 on Build GRCh38
rs1001803100 Chr.5: 112161792-112161792 on Build GRCh38
rs778614914 Chr.5: 112161379-112161379 on Build GRCh38
rs764510397 Chr.5: 112161478-112161478 on Build GRCh38
rs749768738 Chr.5: 112161551-112161551 on Build GRCh38
rs776377126 Chr.5: 112161657-112161657 on Build GRCh38
rs1032561431 Chr.5: 112161798-112161798 on Build GRCh38
rs762947043 Chr.5: 112161720-112161720 on Build GRCh38
rs73787784 Chr.5: 112161428-112161428 on Build GRCh38
rs1038133372 Chr.5: 112161776-112161776 on Build GRCh38
rs751642591 Chr.5: 112161803-112161803 on Build GRCh38
rs925479747 Chr.5: 112161421-112161421 on Build GRCh38
rs773753835 Chr.5: 112161647-112161647 on Build GRCh38
rs765119249 Chr.5: 112161607-112161607 on Build GRCh38
rs898311764 Chr.5: 112161378-112161378 on Build GRCh38
rs2290990 Chr.5: 112161633-112161633 on Build GRCh38
rs889117074 Chr.5: 112161466-112161466 on Build GRCh38
rs145533460 Chr.5: 112161409-112161409 on Build GRCh38
rs776003873 Chr.5: 112161471-112161471 on Build GRCh38
rs557703090 Chr.5: 112161769-112161769 on Build GRCh38
rs904871792 Chr.5: 112161784-112161784 on Build GRCh38
rs746358260 Chr.5: 112161595-112161596 on Build GRCh38
rs763425410 Chr.5: 112161476-112161476 on Build GRCh38
rs957508069 Chr.5: 112161847-112161847 on Build GRCh38
rs958251493 Chr.5: 112161802-112161802 on Build GRCh38
rs754912738 Chr.5: 112161509-112161509 on Build GRCh38
rs73787785 Chr.5: 112161631-112161631 on Build GRCh38
rs765865149 Chr.5: 112161502-112161502 on Build GRCh38
rs995314989 Chr.5: 112161585-112161585 on Build GRCh38
rs778656642 Chr.5: 112161512-112161512 on Build GRCh38
rs755197891 Chr.5: 112161401-112161401 on Build GRCh38
rs76805610 Chr.5: 112161641-112161641 on Build GRCh38
rs886943722 Chr.5: 112161729-112161729 on Build GRCh38
rs775495245 Chr.5: 112161538-112161538 on Build GRCh38
rs1039626960 Chr.5: 112161517-112161517 on Build GRCh38
rs776861349 Chr.5: 112161457-112161457 on Build GRCh38
rs374445440 Chr.5: 112161636-112161636 on Build GRCh38
rs747314456 Chr.5: 112161785-112161786 on Build GRCh38
rs958220527 Chr.5: 112161643-112161643 on Build GRCh38
rs75565668 Chr.5: 112161843-112161843 on Build GRCh38
rs758129875 Chr.5: 112161616-112161616 on Build GRCh38
rs770440072 Chr.5: 112161654-112161654 on Build GRCh38
rs386691151 Chr.5: 112161631-112161633 on Build GRCh38
rs898391435 Chr.5: 112161363-112161363 on Build GRCh38
rs781721166 Chr.5: 112161574-112161578 on Build GRCh38
rs762204552 Chr.5: 112161567-112161567 on Build GRCh38
rs752008611 Chr.5: 112161479-112161479 on Build GRCh38
rs746088234 Chr.5: 112161536-112161536 on Build GRCh38
rs771198389 Chr.5: 112161524-112161526 on Build GRCh38
rs756510184 Chr.5: 112161492-112161492 on Build GRCh38
rs755613268 Chr.5: 112161440-112161440 on Build GRCh38
rs936197863 Chr.5: 112161441-112161441 on Build GRCh38
rs766312047 Chr.5: 112161757-112161757 on Build GRCh38
rs17266115 Chr.5: 112161610-112161610 on Build GRCh38
rs760353665 Chr.5: 112161651-112161651 on Build GRCh38
rs370160695 Chr.5: 112161691-112161691 on Build GRCh38
rs748725554 Chr.5: 112161645-112161645 on Build GRCh38
rs528338788 Chr.5: 112161659-112161659 on Build GRCh38
rs777859035 Chr.5: 112161455-112161456 on Build GRCh38
rs748617444 Chr.5: 112161808-112161808 on Build GRCh38
rs769902565 Chr.5: 112161537-112161537 on Build GRCh38
rs753811480 Chr.5: 112161504-112161504 on Build GRCh38
rs535212117 Chr.5: 112161688-112161688 on Build GRCh38
rs768010587 Chr.5: 112161591-112161591 on Build GRCh38
rs1009770026 Chr.5: 112161809-112161809 on Build GRCh38
rs201019931 Chr.5: 112161670-112161670 on Build GRCh38
rs987972550 Chr.5: 112161834-112161834 on Build GRCh38
rs761626679 Chr.5: 112161646-112161646 on Build GRCh38
rs189682034 Chr.5: 112161638-112161638 on Build GRCh38
rs185473047 Chr.5: 112161461-112161461 on Build GRCh38
rs908221145 Chr.5: 112161722-112161722 on Build GRCh38
rs937867031 Chr.5: 112161788-112161788 on Build GRCh38
rs140424886 Chr.5: 112161662-112161662 on Build GRCh38
rs771191410 Chr.5: 112161456-112161456 on Build GRCh38
rs775033407 Chr.5: 112161557-112161557 on Build GRCh38
rs926160321 Chr.5: 112161438-112161438 on Build GRCh38
rs540845114 Chr.5: 112161485-112161485 on Build GRCh38
rs770449141 Chr.5: 112161622-112161623 on Build GRCh38
rs539073923 Chr.5: 112161726-112161726 on Build GRCh38
rs954886526 Chr.5: 112161822-112161822 on Build GRCh38
rs992921970 Chr.5: 112161408-112161408 on Build GRCh38
rs114738159 Chr.5: 112161690-112161690 on Build GRCh38
rs201669130 Chr.5: 112161605-112161605 on Build GRCh38
rs150430305 Chr.5: 112161815-112161815 on Build GRCh38
rs781324475 Chr.5: 112161528-112161528 on Build GRCh38
rs760313894 Chr.5: 112161602-112161602 on Build GRCh38
rs778425282 Chr.5: 112161603-112161603 on Build GRCh38
rs980987563 Chr.5: 112161419-112161419 on Build GRCh38
rs369444005 Chr.5: 112161448-112161448 on Build GRCh38
rs951098075 Chr.5: 112161391-112161391 on Build GRCh38
rs759616321 Chr.5: 112161852-112161852 on Build GRCh38
rs3816657 Chr.5: 112161661-112161661 on Build GRCh38
rs933341591 Chr.5: 112161454-112161454 on Build GRCh38
rs964708502 Chr.5: 112161858-112161858 on Build GRCh38
rs1021561543 Chr.5: 112161811-112161811 on Build GRCh38
rs919754526 Chr.5: 112161779-112161779 on Build GRCh38
rs970078804 Chr.5: 112161417-112161417 on Build GRCh38
rs755595740 Chr.5: 112161497-112161497 on Build GRCh38
rs768866323 Chr.5: 112161640-112161640 on Build GRCh38
rs1025368136 Chr.5: 112161386-112161386 on Build GRCh38
rs779978612 Chr.5: 112161442-112161442 on Build GRCh38

View Full Product Details