Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22640821-22641328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN3
Entrez Gene ID
55909
Gene Name
bridging integrator 3
Gene Aliases
-
UniGene
Hs.433722
Gene Chromosome Location
Chr.8: 22620418-22669148 on Build GRCh38
Transcripts
NM_018688.4 XM_005273573.3 XM_011544586.2 XM_011544587.1 NR_027715.1

Gene Symbol
BIN3-IT1
Entrez Gene ID
80094
Gene Name
BIN3 intronic transcript 1
Gene Aliases
-
UniGene
Hs.675917
Gene Chromosome Location
Chr.8: 22640371-22642209 on Build GRCh38
Transcripts
NM_018688.4 XM_005273573.3 XM_011544586.2 XM_011544587.1 NR_027715.1

SNP Detail

SNP ID Chromosome Location
rs1054363720 Chr.8: 22640931-22640931 on Build GRCh38
rs896579551 Chr.8: 22640967-22640967 on Build GRCh38
rs955308634 Chr.8: 22641060-22641060 on Build GRCh38
rs911494897 Chr.8: 22641125-22641125 on Build GRCh38
rs771726313 Chr.8: 22641099-22641099 on Build GRCh38
rs572508337 Chr.8: 22640895-22640895 on Build GRCh38
rs911459008 Chr.8: 22640889-22640889 on Build GRCh38
rs536305485 Chr.8: 22641183-22641183 on Build GRCh38
rs112535446 Chr.8: 22641108-22641108 on Build GRCh38
rs4872528 Chr.8: 22640915-22640915 on Build GRCh38
rs139941090 Chr.8: 22640991-22640991 on Build GRCh38
rs962102702 Chr.8: 22641004-22641004 on Build GRCh38
rs190671564 Chr.8: 22640904-22640904 on Build GRCh38
rs918723934 Chr.8: 22640882-22640882 on Build GRCh38
rs918346738 Chr.8: 22640859-22640859 on Build GRCh38
rs866789951 Chr.8: 22641262-22641262 on Build GRCh38
rs993298968 Chr.8: 22641023-22641023 on Build GRCh38
rs990119013 Chr.8: 22641178-22641178 on Build GRCh38
rs115614249 Chr.8: 22641256-22641256 on Build GRCh38
rs946037097 Chr.8: 22641188-22641188 on Build GRCh38
rs746960747 Chr.8: 22641086-22641086 on Build GRCh38
rs551758081 Chr.8: 22641100-22641100 on Build GRCh38
rs933732639 Chr.8: 22640865-22640865 on Build GRCh38
rs778465045 Chr.8: 22640978-22640978 on Build GRCh38
rs768541561 Chr.8: 22641296-22641296 on Build GRCh38
rs1018365663 Chr.8: 22641295-22641295 on Build GRCh38
rs1023598261 Chr.8: 22640971-22640971 on Build GRCh38
rs1006194240 Chr.8: 22641253-22641253 on Build GRCh38
rs527330235 Chr.8: 22641002-22641002 on Build GRCh38
rs972999004 Chr.8: 22641001-22641001 on Build GRCh38
rs941886258 Chr.8: 22641228-22641228 on Build GRCh38
rs1055607076 Chr.8: 22641222-22641222 on Build GRCh38
rs544140177 Chr.8: 22640936-22640936 on Build GRCh38
rs964268131 Chr.8: 22641304-22641304 on Build GRCh38
rs910858484 Chr.8: 22640885-22640885 on Build GRCh38
rs748859634 Chr.8: 22640966-22640966 on Build GRCh38
rs146029871 Chr.8: 22640981-22640981 on Build GRCh38
rs185314738 Chr.8: 22640881-22640881 on Build GRCh38
rs542752800 Chr.8: 22641285-22641287 on Build GRCh38
rs1045151740 Chr.8: 22641227-22641227 on Build GRCh38
rs111994113 Chr.8: 22641209-22641210 on Build GRCh38
rs986582994 Chr.8: 22641093-22641093 on Build GRCh38
rs781163511 Chr.8: 22641030-22641030 on Build GRCh38
rs142617327 Chr.8: 22640970-22640970 on Build GRCh38
rs554645954 Chr.8: 22641288-22641288 on Build GRCh38
rs565229200 Chr.8: 22640905-22640905 on Build GRCh38
rs942725149 Chr.8: 22640911-22640914 on Build GRCh38
rs113326510 Chr.8: 22641210-22641210 on Build GRCh38
rs575251147 Chr.8: 22640976-22640976 on Build GRCh38
rs949545895 Chr.8: 22641195-22641195 on Build GRCh38
rs570312096 Chr.8: 22641105-22641105 on Build GRCh38
rs938552755 Chr.8: 22641217-22641217 on Build GRCh38
rs1008365584 Chr.8: 22641287-22641287 on Build GRCh38
rs143361125 Chr.8: 22641255-22641255 on Build GRCh38
rs546012807 Chr.8: 22640932-22640932 on Build GRCh38
rs905274890 Chr.8: 22641250-22641250 on Build GRCh38
rs771913261 Chr.8: 22640896-22640896 on Build GRCh38
rs918056012 Chr.8: 22641194-22641194 on Build GRCh38
rs894372367 Chr.8: 22640998-22640998 on Build GRCh38
rs143470152 Chr.8: 22641166-22641166 on Build GRCh38
rs775555066 Chr.8: 22641124-22641124 on Build GRCh38
rs925539312 Chr.8: 22641136-22641136 on Build GRCh38
rs935572968 Chr.8: 22641176-22641176 on Build GRCh38
rs554287361 Chr.8: 22640888-22640888 on Build GRCh38
rs1030103014 Chr.8: 22641281-22641281 on Build GRCh38
rs3735898 Chr.8: 22640957-22640957 on Build GRCh38
rs374072923 Chr.8: 22641034-22641034 on Build GRCh38
rs1037619784 Chr.8: 22641240-22641240 on Build GRCh38

View Full Product Details