Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73944484-73944982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JPX
Entrez Gene ID
554203
Gene Name
JPX transcript, XIST activator (non-protein coding)
Gene Aliases
DCBALD06, ENOX, LINC00183, NCRNA00183
UniGene
Hs.648316
Gene Chromosome Location
Chr.X: 73944324-74070384 on Build GRCh38
Transcripts
NR_024582.1

SNP Detail

SNP ID Chromosome Location
rs148535176 Chr.X: 73944653-73944653 on Build GRCh38
rs767596614 Chr.X: 73944841-73944841 on Build GRCh38
rs920542627 Chr.X: 73944884-73944884 on Build GRCh38
rs968865609 Chr.X: 73944837-73944837 on Build GRCh38
rs918859284 Chr.X: 73944697-73944697 on Build GRCh38
rs933997148 Chr.X: 73944948-73944948 on Build GRCh38
rs1052410867 Chr.X: 73944954-73944954 on Build GRCh38
rs930461550 Chr.X: 73944935-73944935 on Build GRCh38
rs969171642 Chr.X: 73944578-73944578 on Build GRCh38
rs754678663 Chr.X: 73944714-73944714 on Build GRCh38
rs951593543 Chr.X: 73944698-73944698 on Build GRCh38
rs910076014 Chr.X: 73944778-73944778 on Build GRCh38
rs886215094 Chr.X: 73944734-73944734 on Build GRCh38
rs971678126 Chr.X: 73944650-73944650 on Build GRCh38
rs369835441 Chr.X: 73944815-73944816 on Build GRCh38
rs191507498 Chr.X: 73944624-73944624 on Build GRCh38
rs142856779 Chr.X: 73944776-73944776 on Build GRCh38
rs774620752 Chr.X: 73944667-73944667 on Build GRCh38
rs1040051187 Chr.X: 73944774-73944774 on Build GRCh38
rs1020269451 Chr.X: 73944524-73944524 on Build GRCh38
rs984262162 Chr.X: 73944740-73944740 on Build GRCh38
rs965635520 Chr.X: 73944530-73944530 on Build GRCh38
rs979251598 Chr.X: 73944565-73944565 on Build GRCh38
rs970072607 Chr.X: 73944882-73944882 on Build GRCh38
rs979983108 Chr.X: 73944883-73944883 on Build GRCh38
rs998178050 Chr.X: 73944518-73944518 on Build GRCh38
rs997101277 Chr.X: 73944796-73944796 on Build GRCh38
rs996986776 Chr.X: 73944532-73944532 on Build GRCh38
rs371562434 Chr.X: 73944865-73944865 on Build GRCh38
rs992864921 Chr.X: 73944609-73944609 on Build GRCh38
rs1022096210 Chr.X: 73944570-73944570 on Build GRCh38
rs1044168929 Chr.X: 73944635-73944635 on Build GRCh38
rs917126080 Chr.X: 73944621-73944621 on Build GRCh38
rs68178357 Chr.X: 73944648-73944648 on Build GRCh38
rs149942356 Chr.X: 73944641-73944641 on Build GRCh38
rs892357512 Chr.X: 73944543-73944543 on Build GRCh38
rs747657529 Chr.X: 73944809-73944809 on Build GRCh38
rs182180379 Chr.X: 73944845-73944845 on Build GRCh38
rs1028618951 Chr.X: 73944797-73944797 on Build GRCh38
rs956162788 Chr.X: 73944862-73944862 on Build GRCh38
rs1047924499 Chr.X: 73944690-73944690 on Build GRCh38
rs1028501312 Chr.X: 73944537-73944537 on Build GRCh38
rs757241649 Chr.X: 73944855-73944855 on Build GRCh38
rs766027179 Chr.X: 73944649-73944649 on Build GRCh38
rs924753821 Chr.X: 73944568-73944568 on Build GRCh38
rs1008957752 Chr.X: 73944759-73944759 on Build GRCh38
rs968749098 Chr.X: 73944548-73944548 on Build GRCh38
rs187292207 Chr.X: 73944870-73944870 on Build GRCh38
rs956220720 Chr.X: 73944596-73944596 on Build GRCh38
rs563242155 Chr.X: 73944852-73944852 on Build GRCh38
rs113865234 Chr.X: 73944515-73944515 on Build GRCh38
rs983324094 Chr.X: 73944945-73944945 on Build GRCh38

View Full Product Details