Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34838158-34838705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM205BP
Entrez Gene ID
389715
Gene Name
transmembrane protein C9orf144B pseudogene
Gene Aliases
C9orf144, FAM205B
UniGene
Hs.713965
Gene Chromosome Location
Chr.9: 34830267-34838586 on Build GRCh38
Transcripts
NR_024481.1 XM_011517827.1 XM_017014554.1 XM_017014555.1

Gene Symbol
PHF24
Entrez Gene ID
23349
Gene Name
PHD finger protein 24
Gene Aliases
KIAA1045
UniGene
Hs.7989
Gene Chromosome Location
Chr.9: 34810040-34982544 on Build GRCh38
Transcripts
NR_024481.1 XM_011517827.1 XM_017014554.1 XM_017014555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5964322 c.112T>G p.L38V Chr.9: 34838382-34838382 on Build GRCh38
1598264 c.22C>A p.L8M Chr.9: 34838472-34838472 on Build GRCh38
7310055 c.84C>A p.I28I Chr.9: 34838410-34838410 on Build GRCh38
5573322 c.39T>G p.Y13* Chr.9: 34838455-34838455 on Build GRCh38
2773686 c.30T>A p.D10E Chr.9: 34838464-34838464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053825882 Chr.9: 34838480-34838480 on Build GRCh38
rs551844187 Chr.9: 34838593-34838593 on Build GRCh38
rs981396353 Chr.9: 34838181-34838181 on Build GRCh38
rs937382334 Chr.9: 34838286-34838286 on Build GRCh38
rs530031187 Chr.9: 34838507-34838507 on Build GRCh38
rs758158821 Chr.9: 34838536-34838536 on Build GRCh38
rs992887043 Chr.9: 34838315-34838315 on Build GRCh38
rs58682572 Chr.9: 34838523-34838523 on Build GRCh38
rs145144293 Chr.9: 34838544-34838544 on Build GRCh38
rs552566838 Chr.9: 34838225-34838225 on Build GRCh38
rs544698576 Chr.9: 34838240-34838240 on Build GRCh38
rs779006401 Chr.9: 34838444-34838444 on Build GRCh38
rs375446358 Chr.9: 34838508-34838508 on Build GRCh38
rs182360086 Chr.9: 34838232-34838232 on Build GRCh38
rs546950091 Chr.9: 34838512-34838512 on Build GRCh38
rs764727444 Chr.9: 34838313-34838313 on Build GRCh38
rs367739025 Chr.9: 34838670-34838670 on Build GRCh38
rs766361780 Chr.9: 34838475-34838475 on Build GRCh38
rs917361641 Chr.9: 34838336-34838336 on Build GRCh38
rs769437773 Chr.9: 34838364-34838364 on Build GRCh38
rs777585344 Chr.9: 34838537-34838537 on Build GRCh38
rs867132131 Chr.9: 34838466-34838466 on Build GRCh38
rs561208584 Chr.9: 34838437-34838437 on Build GRCh38
rs1005050009 Chr.9: 34838604-34838604 on Build GRCh38
rs748262479 Chr.9: 34838497-34838497 on Build GRCh38
rs537604373 Chr.9: 34838634-34838634 on Build GRCh38
rs575352853 Chr.9: 34838409-34838409 on Build GRCh38
rs763437236 Chr.9: 34838396-34838396 on Build GRCh38
rs755031634 Chr.9: 34838425-34838425 on Build GRCh38
rs139592160 Chr.9: 34838277-34838277 on Build GRCh38
rs544077348 Chr.9: 34838424-34838424 on Build GRCh38
rs777543247 Chr.9: 34838539-34838539 on Build GRCh38
rs760723070 Chr.9: 34838457-34838457 on Build GRCh38
rs1015134759 Chr.9: 34838621-34838621 on Build GRCh38
rs774719263 Chr.9: 34838419-34838419 on Build GRCh38
rs778589832 Chr.9: 34838521-34838521 on Build GRCh38
rs373564359 Chr.9: 34838516-34838516 on Build GRCh38
rs770469504 Chr.9: 34838356-34838356 on Build GRCh38
rs752143210 Chr.9: 34838474-34838474 on Build GRCh38
rs963701531 Chr.9: 34838217-34838217 on Build GRCh38
rs199645052 Chr.9: 34838386-34838386 on Build GRCh38
rs147132769 Chr.9: 34838602-34838602 on Build GRCh38
rs775196173 Chr.9: 34838397-34838397 on Build GRCh38

View Full Product Details