Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133084854-133085412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELP
Entrez Gene ID
1057
Gene Name
carboxyl ester lipase pseudogene
Gene Aliases
CELL, cell1, cell2, cell3
UniGene
Hs.654361
Gene Chromosome Location
Chr.9: 133082539-133087091 on Build GRCh38
Transcripts
NR_001275.2

SNP Detail

SNP ID Chromosome Location
rs664844 Chr.9: 133085387-133085387 on Build GRCh38
rs777222779 Chr.9: 133085009-133085009 on Build GRCh38
rs899239290 Chr.9: 133085254-133085254 on Build GRCh38
rs906676087 Chr.9: 133085342-133085342 on Build GRCh38
rs778617259 Chr.9: 133084944-133084944 on Build GRCh38
rs534539218 Chr.9: 133085060-133085060 on Build GRCh38
rs747632100 Chr.9: 133084952-133084952 on Build GRCh38
rs1049627080 Chr.9: 133085198-133085198 on Build GRCh38
rs755811106 Chr.9: 133084922-133084922 on Build GRCh38
rs763209122 Chr.9: 133084902-133084902 on Build GRCh38
rs770261402 Chr.9: 133085075-133085075 on Build GRCh38
rs536363234 Chr.9: 133084985-133084985 on Build GRCh38
rs959061556 Chr.9: 133085361-133085361 on Build GRCh38
rs572006051 Chr.9: 133084894-133084894 on Build GRCh38
rs777276942 Chr.9: 133084984-133084984 on Build GRCh38
rs753787800 Chr.9: 133084941-133084941 on Build GRCh38
rs978309766 Chr.9: 133085158-133085158 on Build GRCh38
rs754559320 Chr.9: 133084943-133084943 on Build GRCh38
rs555019062 Chr.9: 133084989-133084989 on Build GRCh38
rs552738310 Chr.9: 133085104-133085104 on Build GRCh38
rs943367738 Chr.9: 133085206-133085206 on Build GRCh38
rs539332430 Chr.9: 133084899-133084899 on Build GRCh38
rs573231713 Chr.9: 133085008-133085008 on Build GRCh38
rs544857927 Chr.9: 133085189-133085189 on Build GRCh38
rs745695391 Chr.9: 133084955-133084957 on Build GRCh38
rs1003768177 Chr.9: 133084885-133084885 on Build GRCh38
rs1004204098 Chr.9: 133085350-133085350 on Build GRCh38
rs4962092 Chr.9: 133084921-133084921 on Build GRCh38
rs993708316 Chr.9: 133085283-133085283 on Build GRCh38
rs776945767 Chr.9: 133084888-133084888 on Build GRCh38
rs933920073 Chr.9: 133085180-133085180 on Build GRCh38
rs922432905 Chr.9: 133085171-133085171 on Build GRCh38
rs776653348 Chr.9: 133085358-133085358 on Build GRCh38
rs958985301 Chr.9: 133084934-133084934 on Build GRCh38
rs1010572460 Chr.9: 133085362-133085362 on Build GRCh38
rs878975386 Chr.9: 133085098-133085099 on Build GRCh38
rs577560860 Chr.9: 133085112-133085112 on Build GRCh38
rs914985833 Chr.9: 133085055-133085055 on Build GRCh38
rs764220564 Chr.9: 133084914-133084914 on Build GRCh38
rs528180600 Chr.9: 133085330-133085330 on Build GRCh38
rs139783742 Chr.9: 133085315-133085315 on Build GRCh38
rs612716 Chr.9: 133085367-133085367 on Build GRCh38
rs13291768 Chr.9: 133085205-133085205 on Build GRCh38
rs765258131 Chr.9: 133084895-133084895 on Build GRCh38
rs370286695 Chr.9: 133084983-133084983 on Build GRCh38
rs59909528 Chr.9: 133085243-133085243 on Build GRCh38
rs866241688 Chr.9: 133085105-133085105 on Build GRCh38
rs780050039 Chr.9: 133084931-133084931 on Build GRCh38

View Full Product Details