Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86645968-86646443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA3P
Entrez Gene ID
9629
Gene Name
chloride channel accessory 3, pseudogene
Gene Aliases
CLCA3
UniGene
Hs.673847
Gene Chromosome Location
Chr.1: 86634276-86655376 on Build GRCh38
Transcripts
NR_024604.1 NR_135837.1

Gene Symbol
LOC105378828
Entrez Gene ID
105378828
Gene Name
uncharacterized LOC105378828
Gene Aliases
-
UniGene
Hs.175285
Gene Chromosome Location
Chr.1: 86571181-86704493 on Build GRCh38
Transcripts
NR_024604.1 NR_135837.1

SNP Detail

SNP ID Chromosome Location
rs940707087 Chr.1: 86646037-86646037 on Build GRCh38
rs533436237 Chr.1: 86646203-86646203 on Build GRCh38
rs750181683 Chr.1: 86646409-86646409 on Build GRCh38
rs1023890809 Chr.1: 86646086-86646086 on Build GRCh38
rs572834112 Chr.1: 86646141-86646141 on Build GRCh38
rs1018640457 Chr.1: 86646151-86646151 on Build GRCh38
rs1033179390 Chr.1: 86646127-86646127 on Build GRCh38
rs549026004 Chr.1: 86646366-86646366 on Build GRCh38
rs1034989482 Chr.1: 86646030-86646030 on Build GRCh38
rs563720896 Chr.1: 86646266-86646266 on Build GRCh38
rs920803330 Chr.1: 86646290-86646290 on Build GRCh38
rs965664969 Chr.1: 86646215-86646215 on Build GRCh38
rs531347803 Chr.1: 86646149-86646150 on Build GRCh38
rs896529461 Chr.1: 86646403-86646403 on Build GRCh38
rs953128918 Chr.1: 86646138-86646138 on Build GRCh38
rs556865696 Chr.1: 86646031-86646031 on Build GRCh38
rs573723282 Chr.1: 86646050-86646050 on Build GRCh38
rs1020166377 Chr.1: 86646049-86646049 on Build GRCh38
rs968945777 Chr.1: 86646097-86646097 on Build GRCh38
rs978561765 Chr.1: 86646118-86646118 on Build GRCh38
rs759780835 Chr.1: 86646124-86646124 on Build GRCh38
rs772103189 Chr.1: 86646240-86646240 on Build GRCh38
rs911521265 Chr.1: 86646233-86646233 on Build GRCh38
rs771285690 Chr.1: 86646000-86646000 on Build GRCh38
rs765277935 Chr.1: 86646326-86646326 on Build GRCh38
rs1056935081 Chr.1: 86646374-86646374 on Build GRCh38
rs529392840 Chr.1: 86646345-86646345 on Build GRCh38
rs896477096 Chr.1: 86646060-86646060 on Build GRCh38
rs907966755 Chr.1: 86646332-86646332 on Build GRCh38
rs1049644204 Chr.1: 86646006-86646006 on Build GRCh38
rs184603867 Chr.1: 86646113-86646113 on Build GRCh38
rs76943995 Chr.1: 86646248-86646248 on Build GRCh38
rs537280797 Chr.1: 86646007-86646007 on Build GRCh38
rs564740465 Chr.1: 86646174-86646174 on Build GRCh38
rs775682513 Chr.1: 86646296-86646296 on Build GRCh38
rs940816302 Chr.1: 86646352-86646352 on Build GRCh38
rs940987399 Chr.1: 86646238-86646243 on Build GRCh38
rs565753553 Chr.1: 86646368-86646368 on Build GRCh38
rs528441753 Chr.1: 86646415-86646415 on Build GRCh38
rs761635884 Chr.1: 86646322-86646322 on Build GRCh38
rs541760014 Chr.1: 86646171-86646171 on Build GRCh38
rs147200459 Chr.1: 86646137-86646137 on Build GRCh38
rs1010117284 Chr.1: 86646041-86646041 on Build GRCh38
rs1012528717 Chr.1: 86646084-86646084 on Build GRCh38
rs1056441451 Chr.1: 86646055-86646055 on Build GRCh38

View Full Product Details