Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86649217-86649764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA3P
Entrez Gene ID
9629
Gene Name
chloride channel accessory 3, pseudogene
Gene Aliases
CLCA3
UniGene
Hs.673847
Gene Chromosome Location
Chr.1: 86634276-86655376 on Build GRCh38
Transcripts
NR_024604.1 NR_135837.1

Gene Symbol
LOC105378828
Entrez Gene ID
105378828
Gene Name
uncharacterized LOC105378828
Gene Aliases
-
UniGene
Hs.175285
Gene Chromosome Location
Chr.1: 86571181-86704493 on Build GRCh38
Transcripts
NR_024604.1 NR_135837.1

SNP Detail

SNP ID Chromosome Location
rs941417028 Chr.1: 86649271-86649271 on Build GRCh38
rs544312228 Chr.1: 86649400-86649400 on Build GRCh38
rs369255240 Chr.1: 86649424-86649424 on Build GRCh38
rs933816944 Chr.1: 86649251-86649251 on Build GRCh38
rs1001664887 Chr.1: 86649449-86649449 on Build GRCh38
rs1012199192 Chr.1: 86649298-86649298 on Build GRCh38
rs190314525 Chr.1: 86649474-86649474 on Build GRCh38
rs767226119 Chr.1: 86649535-86649535 on Build GRCh38
rs530062360 Chr.1: 86649506-86649506 on Build GRCh38
rs915935299 Chr.1: 86649592-86649592 on Build GRCh38
rs760280999 Chr.1: 86649730-86649730 on Build GRCh38
rs759305703 Chr.1: 86649378-86649378 on Build GRCh38
rs888540093 Chr.1: 86649262-86649262 on Build GRCh38
rs1017929317 Chr.1: 86649408-86649408 on Build GRCh38
rs866302295 Chr.1: 86649411-86649411 on Build GRCh38
rs911309230 Chr.1: 86649699-86649699 on Build GRCh38
rs1021987456 Chr.1: 86649537-86649537 on Build GRCh38
rs185947160 Chr.1: 86649417-86649417 on Build GRCh38
rs933826888 Chr.1: 86649670-86649670 on Build GRCh38
rs959124678 Chr.1: 86649522-86649522 on Build GRCh38
rs114326325 Chr.1: 86649702-86649702 on Build GRCh38
rs941374240 Chr.1: 86649346-86649346 on Build GRCh38
rs139014337 Chr.1: 86649349-86649349 on Build GRCh38
rs1056240728 Chr.1: 86649280-86649280 on Build GRCh38
rs750954657 Chr.1: 86649350-86649350 on Build GRCh38
rs143256850 Chr.1: 86649332-86649332 on Build GRCh38
rs558751946 Chr.1: 86649347-86649347 on Build GRCh38
rs989241777 Chr.1: 86649360-86649360 on Build GRCh38
rs910050789 Chr.1: 86649508-86649509 on Build GRCh38
rs371729591 Chr.1: 86649469-86649469 on Build GRCh38
rs1045731569 Chr.1: 86649609-86649609 on Build GRCh38
rs1047199789 Chr.1: 86649252-86649252 on Build GRCh38
rs61802879 Chr.1: 86649519-86649519 on Build GRCh38
rs922870285 Chr.1: 86649435-86649435 on Build GRCh38
rs894473689 Chr.1: 86649282-86649282 on Build GRCh38
rs142971207 Chr.1: 86649588-86649588 on Build GRCh38
rs966778172 Chr.1: 86649562-86649562 on Build GRCh38
rs1038344713 Chr.1: 86649348-86649348 on Build GRCh38
rs976364580 Chr.1: 86649427-86649427 on Build GRCh38
rs1024559317 Chr.1: 86649358-86649358 on Build GRCh38
rs930210293 Chr.1: 86649447-86649447 on Build GRCh38
rs922798080 Chr.1: 86649243-86649243 on Build GRCh38
rs181769370 Chr.1: 86649303-86649303 on Build GRCh38
rs1017997543 Chr.1: 86649711-86649712 on Build GRCh38
rs929929261 Chr.1: 86649731-86649731 on Build GRCh38
rs985234395 Chr.1: 86649678-86649678 on Build GRCh38
rs906456201 Chr.1: 86649620-86649620 on Build GRCh38
rs138005088 Chr.1: 86649530-86649533 on Build GRCh38
rs540486465 Chr.1: 86649574-86649574 on Build GRCh38
rs573058123 Chr.1: 86649307-86649307 on Build GRCh38
rs950047764 Chr.1: 86649595-86649595 on Build GRCh38
rs78019701 Chr.1: 86649517-86649517 on Build GRCh38
rs1038842230 Chr.1: 86649710-86649710 on Build GRCh38
rs1052736780 Chr.1: 86649658-86649658 on Build GRCh38
rs922487744 Chr.1: 86649649-86649649 on Build GRCh38

View Full Product Details