Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 166604650-166605199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO9P
Entrez Gene ID
116123
Gene Name
flavin containing monooxygenase 9 pseudogene
Gene Aliases
-
UniGene
Hs.348539
Gene Chromosome Location
Chr.1: 166603916-166625238 on Build GRCh38
Transcripts
NR_002925.2

SNP Detail

SNP ID Chromosome Location
rs930557232 Chr.1: 166604740-166604740 on Build GRCh38
rs969447933 Chr.1: 166605103-166605103 on Build GRCh38
rs1003346874 Chr.1: 166604826-166604826 on Build GRCh38
rs911673447 Chr.1: 166605081-166605081 on Build GRCh38
rs538616175 Chr.1: 166604920-166604920 on Build GRCh38
rs558437846 Chr.1: 166604927-166604927 on Build GRCh38
rs1022954323 Chr.1: 166604948-166604948 on Build GRCh38
rs961992723 Chr.1: 166604726-166604726 on Build GRCh38
rs934800862 Chr.1: 166604671-166604671 on Build GRCh38
rs769200387 Chr.1: 166605059-166605059 on Build GRCh38
rs554742323 Chr.1: 166605149-166605149 on Build GRCh38
rs974835667 Chr.1: 166605127-166605127 on Build GRCh38
rs897900597 Chr.1: 166605051-166605051 on Build GRCh38
rs750134619 Chr.1: 166605135-166605135 on Build GRCh38
rs185913169 Chr.1: 166604857-166604857 on Build GRCh38
rs1014963310 Chr.1: 166604717-166604717 on Build GRCh38
rs189232974 Chr.1: 166604690-166604690 on Build GRCh38
rs537698015 Chr.1: 166605148-166605148 on Build GRCh38
rs575449000 Chr.1: 166605126-166605126 on Build GRCh38
rs774792429 Chr.1: 166605123-166605123 on Build GRCh38
rs539593517 Chr.1: 166605170-166605170 on Build GRCh38
rs954608079 Chr.1: 166604677-166604677 on Build GRCh38
rs567699338 Chr.1: 166604815-166604815 on Build GRCh38
rs979049224 Chr.1: 166604987-166604987 on Build GRCh38
rs890246452 Chr.1: 166604944-166604944 on Build GRCh38
rs1027684769 Chr.1: 166604746-166604746 on Build GRCh38
rs899105171 Chr.1: 166604738-166604738 on Build GRCh38
rs776168552 Chr.1: 166604852-166604852 on Build GRCh38
rs1008700582 Chr.1: 166605015-166605015 on Build GRCh38
rs536737329 Chr.1: 166604827-166604827 on Build GRCh38
rs1038685951 Chr.1: 166604735-166604735 on Build GRCh38
rs1051467182 Chr.1: 166604674-166604674 on Build GRCh38
rs764903466 Chr.1: 166605113-166605113 on Build GRCh38
rs956209103 Chr.1: 166605053-166605053 on Build GRCh38
rs1043104609 Chr.1: 166604911-166604911 on Build GRCh38
rs988321847 Chr.1: 166605128-166605128 on Build GRCh38
rs970025534 Chr.1: 166604769-166604769 on Build GRCh38
rs376630973 Chr.1: 166604682-166604682 on Build GRCh38
rs181629875 Chr.1: 166604787-166604787 on Build GRCh38
rs373055236 Chr.1: 166604798-166604798 on Build GRCh38
rs1031850007 Chr.1: 166604990-166604990 on Build GRCh38
rs978576240 Chr.1: 166605138-166605138 on Build GRCh38
rs928113263 Chr.1: 166604821-166604821 on Build GRCh38
rs1014930711 Chr.1: 166605022-166605022 on Build GRCh38
rs913004881 Chr.1: 166604865-166604865 on Build GRCh38
rs939508376 Chr.1: 166604822-166604822 on Build GRCh38
rs981084793 Chr.1: 166604781-166604781 on Build GRCh38
rs969400878 Chr.1: 166604959-166604959 on Build GRCh38
rs931890269 Chr.1: 166604919-166604919 on Build GRCh38
rs525948 Chr.1: 166604909-166604909 on Build GRCh38
rs945762986 Chr.1: 166605136-166605136 on Build GRCh38
rs546619349 Chr.1: 166604908-166604908 on Build GRCh38
rs904214299 Chr.1: 166604913-166604913 on Build GRCh38
rs763580815 Chr.1: 166604794-166604794 on Build GRCh38

View Full Product Details