Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100168342-100168890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA2P5
Entrez Gene ID
55592
Gene Name
golgin A2 pseudogene 5
Gene Aliases
GOLGA2B, GOLGA2L1
UniGene
Hs.524660
Gene Chromosome Location
Chr.12: 100156391-100173343 on Build GRCh38
Transcripts
NR_024261.2 NR_036632.1

SNP Detail

SNP ID Chromosome Location
rs541875382 Chr.12: 100168614-100168614 on Build GRCh38
rs146649120 Chr.12: 100168536-100168536 on Build GRCh38
rs897971561 Chr.12: 100168618-100168618 on Build GRCh38
rs926495256 Chr.12: 100168770-100168770 on Build GRCh38
rs994905084 Chr.12: 100168414-100168414 on Build GRCh38
rs953730138 Chr.12: 100168852-100168852 on Build GRCh38
rs898087373 Chr.12: 100168836-100168836 on Build GRCh38
rs527670825 Chr.12: 100168665-100168665 on Build GRCh38
rs953330249 Chr.12: 100168439-100168439 on Build GRCh38
rs913835974 Chr.12: 100168699-100168699 on Build GRCh38
rs553876368 Chr.12: 100168385-100168385 on Build GRCh38
rs574713664 Chr.12: 100168580-100168580 on Build GRCh38
rs565629346 Chr.12: 100168456-100168456 on Build GRCh38
rs897217386 Chr.12: 100168366-100168366 on Build GRCh38
rs912496717 Chr.12: 100168856-100168856 on Build GRCh38
rs1043429073 Chr.12: 100168488-100168488 on Build GRCh38
rs773724507 Chr.12: 100168741-100168741 on Build GRCh38
rs1050141872 Chr.12: 100168492-100168492 on Build GRCh38
rs560223074 Chr.12: 100168664-100168664 on Build GRCh38
rs1036586359 Chr.12: 100168827-100168827 on Build GRCh38
rs550068286 Chr.12: 100168779-100168779 on Build GRCh38
rs979235938 Chr.12: 100168743-100168743 on Build GRCh38
rs553905506 Chr.12: 100168442-100168442 on Build GRCh38
rs939720274 Chr.12: 100168823-100168823 on Build GRCh38
rs568134807 Chr.12: 100168409-100168409 on Build GRCh38
rs960526634 Chr.12: 100168443-100168443 on Build GRCh38
rs752760563 Chr.12: 100168448-100168448 on Build GRCh38
rs184107156 Chr.12: 100168564-100168564 on Build GRCh38
rs967822601 Chr.12: 100168712-100168712 on Build GRCh38
rs905909368 Chr.12: 100168679-100168679 on Build GRCh38
rs995016702 Chr.12: 100168637-100168637 on Build GRCh38
rs2303625 Chr.12: 100168447-100168447 on Build GRCh38
rs114732197 Chr.12: 100168698-100168698 on Build GRCh38
rs867062759 Chr.12: 100168500-100168500 on Build GRCh38
rs535511207 Chr.12: 100168423-100168423 on Build GRCh38
rs778297762 Chr.12: 100168525-100168525 on Build GRCh38
rs979110630 Chr.12: 100168468-100168468 on Build GRCh38
rs568112090 Chr.12: 100168844-100168844 on Build GRCh38
rs559198955 Chr.12: 100168499-100168499 on Build GRCh38
rs937748560 Chr.12: 100168773-100168773 on Build GRCh38
rs191665430 Chr.12: 100168505-100168505 on Build GRCh38
rs117855075 Chr.12: 100168696-100168696 on Build GRCh38
rs925016481 Chr.12: 100168778-100168778 on Build GRCh38
rs61940504 Chr.12: 100168471-100168471 on Build GRCh38
rs1036639267 Chr.12: 100168610-100168610 on Build GRCh38
rs1056243704 Chr.12: 100168799-100168799 on Build GRCh38
rs966453424 Chr.12: 100168768-100168768 on Build GRCh38
rs548567915 Chr.12: 100168370-100168370 on Build GRCh38
rs1014544743 Chr.12: 100168369-100168369 on Build GRCh38
rs748922539 Chr.12: 100168868-100168868 on Build GRCh38
rs986540832 Chr.12: 100168441-100168441 on Build GRCh38
rs140176993 Chr.12: 100168697-100168697 on Build GRCh38
rs917554527 Chr.12: 100168805-100168805 on Build GRCh38
rs754889078 Chr.12: 100168747-100168747 on Build GRCh38
rs1027758542 Chr.12: 100168418-100168418 on Build GRCh38
rs1027705862 Chr.12: 100168641-100168641 on Build GRCh38
rs953441163 Chr.12: 100168652-100168652 on Build GRCh38
rs946573226 Chr.12: 100168474-100168474 on Build GRCh38
rs970300225 Chr.12: 100168433-100168433 on Build GRCh38
rs1048392836 Chr.12: 100168601-100168601 on Build GRCh38
rs913671298 Chr.12: 100168451-100168451 on Build GRCh38
rs985123750 Chr.12: 100168855-100168855 on Build GRCh38

View Full Product Details