Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132085146-132085553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EP400NL
Entrez Gene ID
347918
Gene Name
EP400 N-terminal like
Gene Aliases
-
UniGene
Hs.122115
Gene Chromosome Location
Chr.12: 132084283-132126340 on Build GRCh38
Transcripts
NR_003290.2

SNP Detail

SNP ID Chromosome Location
rs1027160161 Chr.12: 132085408-132085408 on Build GRCh38
rs11246920 Chr.12: 132085529-132085529 on Build GRCh38
rs529311691 Chr.12: 132085325-132085325 on Build GRCh38
rs953559182 Chr.12: 132085287-132085287 on Build GRCh38
rs1035606831 Chr.12: 132085210-132085210 on Build GRCh38
rs944593220 Chr.12: 132085207-132085207 on Build GRCh38
rs931059873 Chr.12: 132085346-132085346 on Build GRCh38
rs368118314 Chr.12: 132085356-132085366 on Build GRCh38
rs766550455 Chr.12: 132085310-132085310 on Build GRCh38
rs937206118 Chr.12: 132085404-132085404 on Build GRCh38
rs1010201145 Chr.12: 132085513-132085513 on Build GRCh38
rs1010679398 Chr.12: 132085235-132085235 on Build GRCh38
rs906853533 Chr.12: 132085354-132085354 on Build GRCh38
rs61944627 Chr.12: 132085444-132085444 on Build GRCh38
rs895811017 Chr.12: 132085234-132085234 on Build GRCh38
rs533357784 Chr.12: 132085269-132085269 on Build GRCh38
rs972823332 Chr.12: 132085331-132085331 on Build GRCh38
rs944034374 Chr.12: 132085518-132085518 on Build GRCh38
rs982900793 Chr.12: 132085459-132085459 on Build GRCh38
rs1034601059 Chr.12: 132085385-132085385 on Build GRCh38
rs930055240 Chr.12: 132085168-132085168 on Build GRCh38
rs147848733 Chr.12: 132085521-132085521 on Build GRCh38
rs951421106 Chr.12: 132085412-132085412 on Build GRCh38
rs1002973699 Chr.12: 132085365-132085365 on Build GRCh38
rs762384260 Chr.12: 132085258-132085258 on Build GRCh38
rs61998162 Chr.12: 132085277-132085277 on Build GRCh38
rs1048172914 Chr.12: 132085389-132085389 on Build GRCh38
rs565488086 Chr.12: 132085498-132085498 on Build GRCh38
rs921817396 Chr.12: 132085520-132085520 on Build GRCh38
rs541698914 Chr.12: 132085186-132085186 on Build GRCh38
rs1001405661 Chr.12: 132085202-132085202 on Build GRCh38
rs912666915 Chr.12: 132085473-132085473 on Build GRCh38
rs895732332 Chr.12: 132085505-132085505 on Build GRCh38
rs117691658 Chr.12: 132085393-132085393 on Build GRCh38
rs543895008 Chr.12: 132085300-132085300 on Build GRCh38
rs554533994 Chr.12: 132085477-132085477 on Build GRCh38
rs1054223809 Chr.12: 132085293-132085293 on Build GRCh38
rs541078493 Chr.12: 132085342-132085342 on Build GRCh38
rs1020705981 Chr.12: 132085519-132085519 on Build GRCh38
rs912583781 Chr.12: 132085205-132085205 on Build GRCh38
rs75954421 Chr.12: 132085244-132085244 on Build GRCh38

View Full Product Details