Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132121284-132121831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EP400NL
Entrez Gene ID
347918
Gene Name
EP400 N-terminal like
Gene Aliases
-
UniGene
Hs.122115
Gene Chromosome Location
Chr.12: 132084283-132126340 on Build GRCh38
Transcripts
NR_003290.2

SNP Detail

SNP ID Chromosome Location
rs188373940 Chr.12: 132121735-132121735 on Build GRCh38
rs938034272 Chr.12: 132121343-132121343 on Build GRCh38
rs949508191 Chr.12: 132121662-132121662 on Build GRCh38
rs531231404 Chr.12: 132121405-132121405 on Build GRCh38
rs909165404 Chr.12: 132121565-132121565 on Build GRCh38
rs574861791 Chr.12: 132121765-132121765 on Build GRCh38
rs950633837 Chr.12: 132121529-132121529 on Build GRCh38
rs1042893457 Chr.12: 132121663-132121663 on Build GRCh38
rs966131295 Chr.12: 132121737-132121737 on Build GRCh38
rs780821930 Chr.12: 132121670-132121670 on Build GRCh38
rs902734109 Chr.12: 132121429-132121429 on Build GRCh38
rs527360413 Chr.12: 132121478-132121478 on Build GRCh38
rs569542908 Chr.12: 132121669-132121669 on Build GRCh38
rs951059274 Chr.12: 132121748-132121748 on Build GRCh38
rs145254150 Chr.12: 132121599-132121599 on Build GRCh38
rs745340862 Chr.12: 132121674-132121674 on Build GRCh38
rs59929481 Chr.12: 132121557-132121557 on Build GRCh38
rs1002702409 Chr.12: 132121655-132121655 on Build GRCh38
rs534295381 Chr.12: 132121805-132121805 on Build GRCh38
rs777882738 Chr.12: 132121458-132121458 on Build GRCh38
rs957032290 Chr.12: 132121459-132121459 on Build GRCh38
rs539194692 Chr.12: 132121555-132121555 on Build GRCh38
rs1041616871 Chr.12: 132121612-132121612 on Build GRCh38
rs879165731 Chr.12: 132121339-132121339 on Build GRCh38
rs75618105 Chr.12: 132121445-132121445 on Build GRCh38
rs1055210516 Chr.12: 132121388-132121388 on Build GRCh38
rs994759901 Chr.12: 132121575-132121575 on Build GRCh38
rs144259874 Chr.12: 132121489-132121489 on Build GRCh38
rs567058946 Chr.12: 132121528-132121528 on Build GRCh38
rs1007213387 Chr.12: 132121464-132121464 on Build GRCh38
rs562398916 Chr.12: 132121401-132121401 on Build GRCh38
rs185456032 Chr.12: 132121577-132121577 on Build GRCh38
rs1053752573 Chr.12: 132121561-132121561 on Build GRCh38
rs193025618 Chr.12: 132121806-132121806 on Build GRCh38
rs1007157562 Chr.12: 132121700-132121700 on Build GRCh38
rs1055130410 Chr.12: 132121589-132121589 on Build GRCh38
rs1017238137 Chr.12: 132121708-132121708 on Build GRCh38
rs748331908 Chr.12: 132121801-132121801 on Build GRCh38
rs867775339 Chr.12: 132121490-132121490 on Build GRCh38
rs139539728 Chr.12: 132121428-132121428 on Build GRCh38
rs192443246 Chr.12: 132121477-132121477 on Build GRCh38
rs568020497 Chr.12: 132121576-132121576 on Build GRCh38
rs1003143189 Chr.12: 132121427-132121427 on Build GRCh38
rs112660940 Chr.12: 132121687-132121687 on Build GRCh38
rs902661781 Chr.12: 132121665-132121665 on Build GRCh38
rs912827987 Chr.12: 132121480-132121480 on Build GRCh38
rs1020508024 Chr.12: 132121402-132121402 on Build GRCh38
rs1026612779 Chr.12: 132121460-132121460 on Build GRCh38
rs907143001 Chr.12: 132121623-132121623 on Build GRCh38
rs887761090 Chr.12: 132121328-132121328 on Build GRCh38

View Full Product Details