Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55773047-55773596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES1P1
Entrez Gene ID
51716
Gene Name
carboxylesterase 1 pseudogene 1
Gene Aliases
CES1A2, CES1A3, CES4, CESR, PCE-3
UniGene
Hs.721645
Gene Chromosome Location
Chr.16: 55760599-55774926 on Build GRCh38
Transcripts
NR_003276.2

SNP Detail

SNP ID Chromosome Location
rs566515919 Chr.16: 55773458-55773458 on Build GRCh38
rs150871113 Chr.16: 55773269-55773270 on Build GRCh38
rs376295531 Chr.16: 55773552-55773552 on Build GRCh38
rs111314076 Chr.16: 55773402-55773402 on Build GRCh38
rs758460628 Chr.16: 55773359-55773359 on Build GRCh38
rs540229179 Chr.16: 55773531-55773531 on Build GRCh38
rs561661845 Chr.16: 55773428-55773428 on Build GRCh38
rs79241300 Chr.16: 55773536-55773536 on Build GRCh38
rs28571062 Chr.16: 55773230-55773230 on Build GRCh38
rs780462081 Chr.16: 55773545-55773545 on Build GRCh38
rs776227182 Chr.16: 55773494-55773494 on Build GRCh38
rs563470107 Chr.16: 55773565-55773565 on Build GRCh38
rs77038480 Chr.16: 55773322-55773322 on Build GRCh38
rs534587127 Chr.16: 55773366-55773366 on Build GRCh38
rs746555621 Chr.16: 55773560-55773560 on Build GRCh38
rs865865803 Chr.16: 55773179-55773179 on Build GRCh38
rs557638225 Chr.16: 55773146-55773146 on Build GRCh38
rs749547710 Chr.16: 55773546-55773546 on Build GRCh38
rs540263745 Chr.16: 55773228-55773228 on Build GRCh38
rs564475380 Chr.16: 55773204-55773204 on Build GRCh38
rs28626221 Chr.16: 55773145-55773145 on Build GRCh38
rs750650822 Chr.16: 55773542-55773542 on Build GRCh38
rs746262373 Chr.16: 55773431-55773431 on Build GRCh38
rs749787192 Chr.16: 55773440-55773440 on Build GRCh38
rs28579748 Chr.16: 55773130-55773130 on Build GRCh38
rs28444662 Chr.16: 55773549-55773549 on Build GRCh38
rs58390444 Chr.16: 55773121-55773121 on Build GRCh38
rs546193413 Chr.16: 55773202-55773202 on Build GRCh38
rs772782289 Chr.16: 55773457-55773457 on Build GRCh38
rs528451356 Chr.16: 55773214-55773214 on Build GRCh38
rs764996873 Chr.16: 55773521-55773521 on Build GRCh38
rs756603772 Chr.16: 55773544-55773544 on Build GRCh38
rs28373962 Chr.16: 55773071-55773071 on Build GRCh38
rs528554767 Chr.16: 55773076-55773076 on Build GRCh38
rs557552553 Chr.16: 55773452-55773452 on Build GRCh38
rs550817699 Chr.16: 55773287-55773287 on Build GRCh38
rs569760232 Chr.16: 55773319-55773319 on Build GRCh38
rs201139605 Chr.16: 55773438-55773438 on Build GRCh38
rs756512208 Chr.16: 55773437-55773437 on Build GRCh38
rs770592707 Chr.16: 55773468-55773468 on Build GRCh38
rs902452862 Chr.16: 55773124-55773124 on Build GRCh38
rs62030211 Chr.16: 55773340-55773340 on Build GRCh38
rs757564128 Chr.16: 55773397-55773397 on Build GRCh38
rs190388070 Chr.16: 55773113-55773113 on Build GRCh38
rs529392466 Chr.16: 55773231-55773231 on Build GRCh38
rs60044394 Chr.16: 55773344-55773344 on Build GRCh38
rs764223925 Chr.16: 55773382-55773382 on Build GRCh38
rs374932054 Chr.16: 55773530-55773530 on Build GRCh38
rs575849682 Chr.16: 55773185-55773185 on Build GRCh38
rs553046509 Chr.16: 55773383-55773383 on Build GRCh38
rs533872023 Chr.16: 55773522-55773522 on Build GRCh38
rs540049968 Chr.16: 55773501-55773501 on Build GRCh38
rs568161800 Chr.16: 55773427-55773427 on Build GRCh38

View Full Product Details