Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6891905-6892471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX12P2
Entrez Gene ID
245
Gene Name
arachidonate 12-lipoxygenase pseudogene 2
Gene Aliases
ALOX12E
UniGene
Hs.378695
Gene Chromosome Location
Chr.17: 6853576-6900349 on Build GRCh38
Transcripts
NR_002710.2

SNP Detail

SNP ID Chromosome Location
rs994753156 Chr.17: 6892340-6892340 on Build GRCh38
rs918671340 Chr.17: 6892127-6892127 on Build GRCh38
rs941684154 Chr.17: 6892383-6892383 on Build GRCh38
rs767086125 Chr.17: 6892341-6892341 on Build GRCh38
rs891200551 Chr.17: 6892269-6892269 on Build GRCh38
rs369545534 Chr.17: 6892191-6892191 on Build GRCh38
rs990001461 Chr.17: 6892435-6892435 on Build GRCh38
rs144841381 Chr.17: 6892073-6892073 on Build GRCh38
rs8076947 Chr.17: 6892329-6892329 on Build GRCh38
rs929082068 Chr.17: 6892174-6892174 on Build GRCh38
rs1017103940 Chr.17: 6892103-6892103 on Build GRCh38
rs77401443 Chr.17: 6892426-6892426 on Build GRCh38
rs374780442 Chr.17: 6892364-6892364 on Build GRCh38
rs748413653 Chr.17: 6892072-6892072 on Build GRCh38
rs927284872 Chr.17: 6892387-6892387 on Build GRCh38
rs901484461 Chr.17: 6892050-6892050 on Build GRCh38
rs1025846568 Chr.17: 6892351-6892351 on Build GRCh38
rs1054328167 Chr.17: 6892208-6892208 on Build GRCh38
rs542609482 Chr.17: 6892122-6892122 on Build GRCh38
rs927356121 Chr.17: 6892168-6892168 on Build GRCh38
rs941902345 Chr.17: 6892199-6892199 on Build GRCh38
rs530722258 Chr.17: 6892081-6892081 on Build GRCh38
rs559285951 Chr.17: 6892004-6892004 on Build GRCh38
rs905783740 Chr.17: 6891931-6891931 on Build GRCh38
rs1054214423 Chr.17: 6891966-6891966 on Build GRCh38
rs985746927 Chr.17: 6892102-6892102 on Build GRCh38
rs775106956 Chr.17: 6892386-6892386 on Build GRCh38
rs866830577 Chr.17: 6892363-6892363 on Build GRCh38
rs184815206 Chr.17: 6892274-6892274 on Build GRCh38
rs770693748 Chr.17: 6892235-6892235 on Build GRCh38
rs550549143 Chr.17: 6892123-6892123 on Build GRCh38
rs568085242 Chr.17: 6891952-6891952 on Build GRCh38
rs766368159 Chr.17: 6892077-6892077 on Build GRCh38
rs1020465783 Chr.17: 6892010-6892010 on Build GRCh38
rs937294314 Chr.17: 6891938-6891938 on Build GRCh38
rs566454814 Chr.17: 6892267-6892267 on Build GRCh38
rs148578365 Chr.17: 6892152-6892152 on Build GRCh38
rs777683965 Chr.17: 6892167-6892167 on Build GRCh38
rs983219529 Chr.17: 6892367-6892367 on Build GRCh38
rs549077113 Chr.17: 6891982-6891982 on Build GRCh38
rs1025593292 Chr.17: 6892359-6892359 on Build GRCh38
rs992331166 Chr.17: 6892345-6892346 on Build GRCh38
rs528486454 Chr.17: 6892065-6892065 on Build GRCh38
rs888805427 Chr.17: 6892284-6892284 on Build GRCh38
rs544102391 Chr.17: 6892373-6892373 on Build GRCh38
rs908903374 Chr.17: 6892196-6892196 on Build GRCh38
rs921868992 Chr.17: 6892392-6892392 on Build GRCh38
rs749981204 Chr.17: 6892360-6892360 on Build GRCh38
rs945901187 Chr.17: 6892244-6892244 on Build GRCh38
rs80279081 Chr.17: 6892249-6892249 on Build GRCh38
rs557235082 Chr.17: 6892379-6892379 on Build GRCh38
rs149867587 Chr.17: 6891971-6891971 on Build GRCh38
rs571482888 Chr.17: 6892077-6892077 on Build GRCh38
rs764121486 Chr.17: 6892095-6892095 on Build GRCh38
rs1001040910 Chr.17: 6892300-6892300 on Build GRCh38
rs117145865 Chr.17: 6892231-6892231 on Build GRCh38
rs933265875 Chr.17: 6892396-6892396 on Build GRCh38
rs1041426930 Chr.17: 6892253-6892253 on Build GRCh38
rs997308049 Chr.17: 6892270-6892270 on Build GRCh38
rs75414248 Chr.17: 6892289-6892289 on Build GRCh38
rs1017218265 Chr.17: 6892321-6892321 on Build GRCh38
rs769889220 Chr.17: 6892116-6892116 on Build GRCh38

View Full Product Details