Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6894998-6895546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX12P2
Entrez Gene ID
245
Gene Name
arachidonate 12-lipoxygenase pseudogene 2
Gene Aliases
ALOX12E
UniGene
Hs.378695
Gene Chromosome Location
Chr.17: 6853576-6900349 on Build GRCh38
Transcripts
NR_002710.2 NR_120453.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7333576 c.435A>G p.G145G Chr.17: 6895183-6895183 on Build GRCh38
3701270 c.364A>C p.I122L Chr.17: 6895112-6895112 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs150500235 Chr.17: 6895299-6895299 on Build GRCh38
rs1023578402 Chr.17: 6895220-6895220 on Build GRCh38
rs923795529 Chr.17: 6895365-6895365 on Build GRCh38
rs906155622 Chr.17: 6895230-6895230 on Build GRCh38
rs761206264 Chr.17: 6895200-6895200 on Build GRCh38
rs540672273 Chr.17: 6895329-6895329 on Build GRCh38
rs527282383 Chr.17: 6895309-6895309 on Build GRCh38
rs1032259757 Chr.17: 6895259-6895259 on Build GRCh38
rs576171456 Chr.17: 6895245-6895245 on Build GRCh38
rs573836 Chr.17: 6895059-6895059 on Build GRCh38
rs987982736 Chr.17: 6895270-6895270 on Build GRCh38
rs775883609 Chr.17: 6895157-6895157 on Build GRCh38
rs911126488 Chr.17: 6895342-6895342 on Build GRCh38
rs181652977 Chr.17: 6895419-6895419 on Build GRCh38
rs943871367 Chr.17: 6895029-6895029 on Build GRCh38
rs956709259 Chr.17: 6895267-6895267 on Build GRCh38
rs184783382 Chr.17: 6895189-6895189 on Build GRCh38
rs554095378 Chr.17: 6895137-6895137 on Build GRCh38
rs1049338920 Chr.17: 6895074-6895074 on Build GRCh38
rs961096276 Chr.17: 6895257-6895257 on Build GRCh38
rs139475307 Chr.17: 6895069-6895071 on Build GRCh38
rs167478 Chr.17: 6895427-6895427 on Build GRCh38
rs1011798008 Chr.17: 6895112-6895112 on Build GRCh38
rs1049743707 Chr.17: 6895372-6895372 on Build GRCh38
rs311762 Chr.17: 6895519-6895519 on Build GRCh38
rs920910831 Chr.17: 6895341-6895341 on Build GRCh38
rs552612695 Chr.17: 6895471-6895471 on Build GRCh38
rs188806265 Chr.17: 6895219-6895219 on Build GRCh38
rs199610445 Chr.17: 6895310-6895310 on Build GRCh38
rs1015541881 Chr.17: 6895113-6895113 on Build GRCh38
rs979734572 Chr.17: 6895238-6895238 on Build GRCh38
rs200596965 Chr.17: 6895345-6895345 on Build GRCh38
rs947373846 Chr.17: 6895446-6895446 on Build GRCh38
rs1053140989 Chr.17: 6895397-6895397 on Build GRCh38
rs561473985 Chr.17: 6895300-6895300 on Build GRCh38
rs893140475 Chr.17: 6895106-6895106 on Build GRCh38
rs1035891612 Chr.17: 6895234-6895234 on Build GRCh38
rs914540486 Chr.17: 6895417-6895417 on Build GRCh38
rs1039382735 Chr.17: 6895032-6895032 on Build GRCh38
rs976541157 Chr.17: 6895346-6895346 on Build GRCh38
rs887903939 Chr.17: 6895096-6895096 on Build GRCh38
rs940980264 Chr.17: 6895461-6895461 on Build GRCh38
rs545780152 Chr.17: 6895211-6895211 on Build GRCh38
rs764122181 Chr.17: 6895232-6895232 on Build GRCh38
rs80048042 Chr.17: 6895104-6895104 on Build GRCh38
rs201488681 Chr.17: 6895084-6895084 on Build GRCh38
rs200633069 Chr.17: 6895068-6895068 on Build GRCh38
rs367716970 Chr.17: 6895073-6895075 on Build GRCh38
rs896574168 Chr.17: 6895172-6895172 on Build GRCh38
rs934717508 Chr.17: 6895369-6895369 on Build GRCh38
rs868493883 Chr.17: 6895288-6895288 on Build GRCh38

View Full Product Details