Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6896205-6896752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX12P2
Entrez Gene ID
245
Gene Name
arachidonate 12-lipoxygenase pseudogene 2
Gene Aliases
ALOX12E
UniGene
Hs.378695
Gene Chromosome Location
Chr.17: 6853576-6900349 on Build GRCh38
Transcripts
NR_002710.2 NR_120453.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5954062 c.757-4C>T p.? Chr.17: 6896550-6896550 on Build GRCh38
6202627 c.682C>T p.Q228* Chr.17: 6896385-6896385 on Build GRCh38
983643 c.684G>A p.Q228Q Chr.17: 6896387-6896387 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs557598563 Chr.17: 6896511-6896511 on Build GRCh38
rs932640227 Chr.17: 6896682-6896682 on Build GRCh38
rs527743553 Chr.17: 6896366-6896366 on Build GRCh38
rs1043816587 Chr.17: 6896605-6896605 on Build GRCh38
rs569898874 Chr.17: 6896473-6896473 on Build GRCh38
rs535615301 Chr.17: 6896435-6896435 on Build GRCh38
rs562228079 Chr.17: 6896695-6896695 on Build GRCh38
rs984397319 Chr.17: 6896304-6896304 on Build GRCh38
rs1052145665 Chr.17: 6896577-6896577 on Build GRCh38
rs897021983 Chr.17: 6896717-6896717 on Build GRCh38
rs918614699 Chr.17: 6896265-6896265 on Build GRCh38
rs1023912172 Chr.17: 6896546-6896546 on Build GRCh38
rs548844282 Chr.17: 6896302-6896302 on Build GRCh38
rs958349375 Chr.17: 6896581-6896581 on Build GRCh38
rs1023964471 Chr.17: 6896338-6896338 on Build GRCh38
rs910128259 Chr.17: 6896489-6896489 on Build GRCh38
rs970934913 Chr.17: 6896342-6896342 on Build GRCh38
rs990052833 Chr.17: 6896597-6896597 on Build GRCh38
rs933715832 Chr.17: 6896564-6896564 on Build GRCh38
rs188016600 Chr.17: 6896505-6896505 on Build GRCh38
rs145764765 Chr.17: 6896500-6896500 on Build GRCh38
rs553536148 Chr.17: 6896580-6896580 on Build GRCh38
rs577553556 Chr.17: 6896535-6896535 on Build GRCh38
rs1034879878 Chr.17: 6896446-6896446 on Build GRCh38
rs531935930 Chr.17: 6896252-6896252 on Build GRCh38
rs555104157 Chr.17: 6896490-6896490 on Build GRCh38
rs769190608 Chr.17: 6896536-6896536 on Build GRCh38
rs981016944 Chr.17: 6896353-6896353 on Build GRCh38
rs976847888 Chr.17: 6896645-6896645 on Build GRCh38
rs571939282 Chr.17: 6896632-6896632 on Build GRCh38
rs969707609 Chr.17: 6896571-6896571 on Build GRCh38
rs1014366923 Chr.17: 6896452-6896452 on Build GRCh38
rs980951417 Chr.17: 6896573-6896573 on Build GRCh38
rs960144031 Chr.17: 6896240-6896240 on Build GRCh38
rs144784950 Chr.17: 6896411-6896411 on Build GRCh38
rs1005276887 Chr.17: 6896260-6896260 on Build GRCh38
rs542547851 Chr.17: 6896663-6896663 on Build GRCh38
rs951416960 Chr.17: 6896463-6896463 on Build GRCh38
rs1049630190 Chr.17: 6896691-6896691 on Build GRCh38
rs961034846 Chr.17: 6896320-6896320 on Build GRCh38
rs1050125807 Chr.17: 6896443-6896443 on Build GRCh38
rs376408546 Chr.17: 6896448-6896448 on Build GRCh38
rs200069311 Chr.17: 6896369-6896369 on Build GRCh38
rs941326991 Chr.17: 6896701-6896701 on Build GRCh38
rs1041050988 Chr.17: 6896399-6896399 on Build GRCh38
rs922572257 Chr.17: 6896659-6896659 on Build GRCh38
rs946398808 Chr.17: 6896603-6896603 on Build GRCh38
rs535366247 Chr.17: 6896484-6896484 on Build GRCh38
rs984568417 Chr.17: 6896478-6896478 on Build GRCh38
rs888289086 Chr.17: 6896253-6896253 on Build GRCh38
rs140960268 Chr.17: 6896385-6896385 on Build GRCh38
rs901205653 Chr.17: 6896402-6896402 on Build GRCh38
rs910002047 Chr.17: 6896327-6896327 on Build GRCh38
rs922345218 Chr.17: 6896364-6896364 on Build GRCh38
rs896333017 Chr.17: 6896649-6896649 on Build GRCh38
rs73345728 Chr.17: 6896393-6896393 on Build GRCh38
rs776641030 Chr.17: 6896644-6896644 on Build GRCh38
rs866295419 Chr.17: 6896494-6896494 on Build GRCh38
rs1014309670 Chr.17: 6896651-6896651 on Build GRCh38
rs945485337 Chr.17: 6896396-6896396 on Build GRCh38
rs1002033460 Chr.17: 6896442-6896442 on Build GRCh38
rs1025621807 Chr.17: 6896457-6896457 on Build GRCh38
rs543094366 Chr.17: 6896575-6896575 on Build GRCh38
rs374665428 Chr.17: 6896634-6896634 on Build GRCh38
rs183260795 Chr.17: 6896334-6896334 on Build GRCh38
rs1037254110 Chr.17: 6896710-6896710 on Build GRCh38

View Full Product Details