Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6898689-6899245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX12P2
Entrez Gene ID
245
Gene Name
arachidonate 12-lipoxygenase pseudogene 2
Gene Aliases
ALOX12E
UniGene
Hs.378695
Gene Chromosome Location
Chr.17: 6853576-6900349 on Build GRCh38
Transcripts
NR_002710.2 NR_120453.1

SNP Detail

SNP ID Chromosome Location
rs1025958359 Chr.17: 6898752-6898752 on Build GRCh38
rs1008728244 Chr.17: 6898898-6898898 on Build GRCh38
rs1025904749 Chr.17: 6898993-6898993 on Build GRCh38
rs990803064 Chr.17: 6898741-6898741 on Build GRCh38
rs1007272633 Chr.17: 6898738-6898738 on Build GRCh38
rs547458503 Chr.17: 6898933-6898933 on Build GRCh38
rs11867644 Chr.17: 6898877-6898877 on Build GRCh38
rs1017480194 Chr.17: 6898966-6898966 on Build GRCh38
rs61742687 Chr.17: 6898915-6898915 on Build GRCh38
rs969179039 Chr.17: 6899165-6899165 on Build GRCh38
rs958135922 Chr.17: 6898739-6898739 on Build GRCh38
rs544534122 Chr.17: 6899190-6899190 on Build GRCh38
rs545427367 Chr.17: 6899039-6899039 on Build GRCh38
rs575448927 Chr.17: 6899094-6899094 on Build GRCh38
rs749393823 Chr.17: 6898771-6898771 on Build GRCh38
rs1017781673 Chr.17: 6898743-6898743 on Build GRCh38
rs960352651 Chr.17: 6899107-6899107 on Build GRCh38
rs992063952 Chr.17: 6899119-6899119 on Build GRCh38
rs180962928 Chr.17: 6898730-6898730 on Build GRCh38
rs311760 Chr.17: 6898811-6898811 on Build GRCh38
rs868384792 Chr.17: 6898946-6898946 on Build GRCh38
rs915973823 Chr.17: 6898824-6898824 on Build GRCh38
rs535533711 Chr.17: 6898879-6898879 on Build GRCh38
rs963195646 Chr.17: 6898745-6898745 on Build GRCh38
rs311759 Chr.17: 6898945-6898945 on Build GRCh38
rs775244479 Chr.17: 6899021-6899021 on Build GRCh38
rs538661721 Chr.17: 6898747-6898747 on Build GRCh38
rs985844304 Chr.17: 6899201-6899201 on Build GRCh38
rs944692581 Chr.17: 6898847-6898847 on Build GRCh38
rs1035910028 Chr.17: 6899025-6899025 on Build GRCh38
rs916110847 Chr.17: 6899150-6899150 on Build GRCh38
rs911679961 Chr.17: 6899203-6899203 on Build GRCh38
rs982370475 Chr.17: 6898759-6898759 on Build GRCh38
rs903131287 Chr.17: 6898926-6898926 on Build GRCh38
rs566320988 Chr.17: 6898742-6898742 on Build GRCh38
rs76171870 Chr.17: 6898938-6898938 on Build GRCh38
rs920345320 Chr.17: 6898871-6898871 on Build GRCh38
rs1012353090 Chr.17: 6898952-6898952 on Build GRCh38
rs78949693 Chr.17: 6898976-6898976 on Build GRCh38
rs962637804 Chr.17: 6899027-6899027 on Build GRCh38
rs950404023 Chr.17: 6899002-6899002 on Build GRCh38
rs1041802138 Chr.17: 6899223-6899223 on Build GRCh38
rs928781108 Chr.17: 6898786-6898786 on Build GRCh38
rs35969592 Chr.17: 6898956-6898957 on Build GRCh38
rs890263293 Chr.17: 6898944-6898944 on Build GRCh38
rs983007396 Chr.17: 6898780-6898780 on Build GRCh38
rs1000474559 Chr.17: 6898734-6898734 on Build GRCh38
rs138693111 Chr.17: 6898807-6898807 on Build GRCh38
rs9897943 Chr.17: 6899220-6899220 on Build GRCh38
rs893875505 Chr.17: 6898946-6898947 on Build GRCh38
rs1020501662 Chr.17: 6898717-6898717 on Build GRCh38
rs903198276 Chr.17: 6898896-6898896 on Build GRCh38
rs565031482 Chr.17: 6899060-6899060 on Build GRCh38
rs2271315 Chr.17: 6898970-6898970 on Build GRCh38
rs982561588 Chr.17: 6898988-6898988 on Build GRCh38

View Full Product Details