Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88786393-88786944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36BP2
Entrez Gene ID
645784
Gene Name
ankyrin repeat domain 36B pseudogene 2
Gene Aliases
-
UniGene
Hs.105323
Gene Chromosome Location
Chr.2: 88765902-88806596 on Build GRCh38
Transcripts
NR_015424.1

SNP Detail

SNP ID Chromosome Location
rs1843220 Chr.2: 88786418-88786418 on Build GRCh38
rs966689016 Chr.2: 88786486-88786486 on Build GRCh38
rs527852359 Chr.2: 88786689-88786689 on Build GRCh38
rs1022537142 Chr.2: 88786831-88786831 on Build GRCh38
rs191920182 Chr.2: 88786630-88786630 on Build GRCh38
rs1022589297 Chr.2: 88786476-88786476 on Build GRCh38
rs549355318 Chr.2: 88786911-88786911 on Build GRCh38
rs867595587 Chr.2: 88786512-88786512 on Build GRCh38
rs567763346 Chr.2: 88786913-88786913 on Build GRCh38
rs184303647 Chr.2: 88786636-88786636 on Build GRCh38
rs943244735 Chr.2: 88786529-88786529 on Build GRCh38
rs191480251 Chr.2: 88786488-88786488 on Build GRCh38
rs148683931 Chr.2: 88786836-88786836 on Build GRCh38
rs973232074 Chr.2: 88786919-88786919 on Build GRCh38
rs17037530 Chr.2: 88786840-88786840 on Build GRCh38
rs943192402 Chr.2: 88786912-88786912 on Build GRCh38
rs776104467 Chr.2: 88786652-88786652 on Build GRCh38
rs558972692 Chr.2: 88786540-88786540 on Build GRCh38
rs1026468514 Chr.2: 88786437-88786437 on Build GRCh38
rs910345796 Chr.2: 88786524-88786524 on Build GRCh38
rs761094839 Chr.2: 88786424-88786424 on Build GRCh38
rs1003256721 Chr.2: 88786715-88786715 on Build GRCh38
rs993155652 Chr.2: 88786436-88786436 on Build GRCh38
rs1011109059 Chr.2: 88786736-88786736 on Build GRCh38
rs753943357 Chr.2: 88786493-88786493 on Build GRCh38
rs922613072 Chr.2: 88786489-88786489 on Build GRCh38
rs769739574 Chr.2: 88786914-88786914 on Build GRCh38
rs765688020 Chr.2: 88786819-88786819 on Build GRCh38
rs761393988 Chr.2: 88786832-88786832 on Build GRCh38
rs955559683 Chr.2: 88786847-88786847 on Build GRCh38
rs1033821118 Chr.2: 88786444-88786444 on Build GRCh38
rs542495631 Chr.2: 88786666-88786666 on Build GRCh38
rs959810299 Chr.2: 88786462-88786462 on Build GRCh38
rs934062009 Chr.2: 88786492-88786492 on Build GRCh38
rs895310661 Chr.2: 88786732-88786732 on Build GRCh38
rs531339935 Chr.2: 88786845-88786845 on Build GRCh38
rs985956593 Chr.2: 88786502-88786502 on Build GRCh38
rs182855631 Chr.2: 88786545-88786545 on Build GRCh38
rs1033887170 Chr.2: 88786728-88786728 on Build GRCh38
rs899115093 Chr.2: 88786563-88786563 on Build GRCh38
rs188672792 Chr.2: 88786616-88786616 on Build GRCh38
rs373445367 Chr.2: 88786655-88786655 on Build GRCh38
rs989730449 Chr.2: 88786465-88786465 on Build GRCh38
rs187946699 Chr.2: 88786686-88786686 on Build GRCh38
rs111873397 Chr.2: 88786439-88786439 on Build GRCh38
rs911328941 Chr.2: 88786897-88786897 on Build GRCh38
rs928954352 Chr.2: 88786606-88786606 on Build GRCh38
rs985559173 Chr.2: 88786877-88786877 on Build GRCh38
rs372455971 Chr.2: 88786442-88786442 on Build GRCh38

View Full Product Details