Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88787938-88788484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36BP2
Entrez Gene ID
645784
Gene Name
ankyrin repeat domain 36B pseudogene 2
Gene Aliases
-
UniGene
Hs.105323
Gene Chromosome Location
Chr.2: 88765902-88806596 on Build GRCh38
Transcripts
NR_015424.1

SNP Detail

SNP ID Chromosome Location
rs977052315 Chr.2: 88788168-88788168 on Build GRCh38
rs539790298 Chr.2: 88788279-88788279 on Build GRCh38
rs986949461 Chr.2: 88788210-88788210 on Build GRCh38
rs1057055520 Chr.2: 88788335-88788335 on Build GRCh38
rs761918480 Chr.2: 88788066-88788066 on Build GRCh38
rs140312981 Chr.2: 88788211-88788211 on Build GRCh38
rs1013048915 Chr.2: 88788091-88788091 on Build GRCh38
rs559698142 Chr.2: 88788078-88788078 on Build GRCh38
rs1039922520 Chr.2: 88787968-88787968 on Build GRCh38
rs1035739913 Chr.2: 88788084-88788084 on Build GRCh38
rs931584787 Chr.2: 88787999-88787999 on Build GRCh38
rs145322830 Chr.2: 88788378-88788378 on Build GRCh38
rs189799842 Chr.2: 88787981-88787981 on Build GRCh38
rs1013023567 Chr.2: 88788380-88788380 on Build GRCh38
rs1005659446 Chr.2: 88788445-88788445 on Build GRCh38
rs1012114430 Chr.2: 88788087-88788087 on Build GRCh38
rs895495018 Chr.2: 88788086-88788086 on Build GRCh38
rs915360235 Chr.2: 88788260-88788260 on Build GRCh38
rs1022717145 Chr.2: 88788106-88788106 on Build GRCh38
rs776362800 Chr.2: 88787996-88787996 on Build GRCh38
rs924457377 Chr.2: 88788323-88788323 on Build GRCh38
rs956400220 Chr.2: 88788185-88788185 on Build GRCh38
rs985943793 Chr.2: 88788231-88788231 on Build GRCh38
rs1005710068 Chr.2: 88788119-88788119 on Build GRCh38
rs568486384 Chr.2: 88788107-88788107 on Build GRCh38
rs954073416 Chr.2: 88788226-88788226 on Build GRCh38
rs888227473 Chr.2: 88788388-88788388 on Build GRCh38
rs1015787969 Chr.2: 88788149-88788149 on Build GRCh38
rs1031047515 Chr.2: 88788132-88788132 on Build GRCh38
rs967931783 Chr.2: 88788115-88788115 on Build GRCh38
rs1047313540 Chr.2: 88788261-88788261 on Build GRCh38
rs934535512 Chr.2: 88788328-88788328 on Build GRCh38
rs942854780 Chr.2: 88788213-88788213 on Build GRCh38
rs1057459452 Chr.2: 88788365-88788365 on Build GRCh38
rs181744936 Chr.2: 88788448-88788448 on Build GRCh38
rs1012624988 Chr.2: 88788359-88788359 on Build GRCh38
rs895433420 Chr.2: 88788337-88788337 on Build GRCh38
rs568591968 Chr.2: 88787985-88787985 on Build GRCh38
rs1042934761 Chr.2: 88788281-88788281 on Build GRCh38
rs947188376 Chr.2: 88788267-88788267 on Build GRCh38
rs903130184 Chr.2: 88788068-88788068 on Build GRCh38
rs1003331083 Chr.2: 88788066-88788067 on Build GRCh38
rs1047023902 Chr.2: 88788036-88788037 on Build GRCh38
rs184575467 Chr.2: 88787958-88787958 on Build GRCh38
rs1044890116 Chr.2: 88788397-88788397 on Build GRCh38
rs931532354 Chr.2: 88788247-88788247 on Build GRCh38
rs779738787 Chr.2: 88788074-88788074 on Build GRCh38
rs938730554 Chr.2: 88788354-88788354 on Build GRCh38
rs535827866 Chr.2: 88788008-88788008 on Build GRCh38

View Full Product Details