Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 125930373-125930727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG1L
Entrez Gene ID
200810
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase like
Gene Aliases
ALG1L1
UniGene
Hs.591299
Gene Chromosome Location
Chr.3: 125929272-125993832 on Build GRCh38
Transcripts
NM_001015050.2 NM_001195223.1 XM_017005844.1 XM_017005845.1 XM_017005846.1 XM_017005847.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4848327 c.291G>A p.V97V Chr.3: 125930614-125930614 on Build GRCh38
7355196 c.320A>G p.E107G Chr.3: 125930585-125930585 on Build GRCh38
3392010 c.297T>C p.H99H Chr.3: 125930608-125930608 on Build GRCh38
1327907 c.298G>C p.E100Q Chr.3: 125930607-125930607 on Build GRCh38
5799409 c.276-6C>A p.? Chr.3: 125930635-125930635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773212429 Chr.3: 125930583-125930583 on Build GRCh38
rs544828238 Chr.3: 125930423-125930423 on Build GRCh38
rs764897297 Chr.3: 125930599-125930599 on Build GRCh38
rs781739929 Chr.3: 125930548-125930548 on Build GRCh38
rs148263244 Chr.3: 125930596-125930596 on Build GRCh38
rs769230664 Chr.3: 125930651-125930651 on Build GRCh38
rs371167256 Chr.3: 125930542-125930542 on Build GRCh38
rs753993286 Chr.3: 125930624-125930624 on Build GRCh38
rs558944876 Chr.3: 125930646-125930646 on Build GRCh38
rs7615952 Chr.3: 125930560-125930560 on Build GRCh38
rs761677350 Chr.3: 125930508-125930508 on Build GRCh38
rs569247561 Chr.3: 125930523-125930523 on Build GRCh38
rs7627571 Chr.3: 125930451-125930451 on Build GRCh38
rs570322638 Chr.3: 125930612-125930612 on Build GRCh38
rs753330905 Chr.3: 125930531-125930531 on Build GRCh38
rs750830267 Chr.3: 125930550-125930550 on Build GRCh38
rs540690257 Chr.3: 125930444-125930444 on Build GRCh38
rs914902516 Chr.3: 125930421-125930421 on Build GRCh38
rs777346811 Chr.3: 125930583-125930583 on Build GRCh38
rs780677551 Chr.3: 125930615-125930615 on Build GRCh38
rs7616019 Chr.3: 125930445-125930445 on Build GRCh38
rs776154533 Chr.3: 125930590-125930590 on Build GRCh38
rs1022076752 Chr.3: 125930620-125930620 on Build GRCh38
rs553708686 Chr.3: 125930609-125930609 on Build GRCh38
rs7618399 Chr.3: 125930656-125930656 on Build GRCh38
rs1046901294 Chr.3: 125930502-125930502 on Build GRCh38
rs750793577 Chr.3: 125930672-125930672 on Build GRCh38
rs867219788 Chr.3: 125930622-125930622 on Build GRCh38
rs779228491 Chr.3: 125930569-125930569 on Build GRCh38
rs941221892 Chr.3: 125930697-125930697 on Build GRCh38
rs11706376 Chr.3: 125930642-125930642 on Build GRCh38
rs776506857 Chr.3: 125930645-125930645 on Build GRCh38
rs1029551690 Chr.3: 125930668-125930668 on Build GRCh38
rs552154933 Chr.3: 125930493-125930493 on Build GRCh38
rs762196082 Chr.3: 125930679-125930679 on Build GRCh38
rs540611176 Chr.3: 125930449-125930449 on Build GRCh38
rs532563488 Chr.3: 125930476-125930476 on Build GRCh38
rs907912625 Chr.3: 125930503-125930503 on Build GRCh38
rs756234331 Chr.3: 125930556-125930556 on Build GRCh38
rs55828572 Chr.3: 125930675-125930675 on Build GRCh38
rs7615926 Chr.3: 125930400-125930400 on Build GRCh38
rs749376193 Chr.3: 125930558-125930558 on Build GRCh38
rs762571707 Chr.3: 125930654-125930654 on Build GRCh38
rs555240585 Chr.3: 125930695-125930695 on Build GRCh38
rs973497144 Chr.3: 125930708-125930708 on Build GRCh38
rs771696795 Chr.3: 125930577-125930577 on Build GRCh38
rs748322526 Chr.3: 125930637-125930637 on Build GRCh38
rs755069359 Chr.3: 125930625-125930625 on Build GRCh38
rs747985612 Chr.3: 125930571-125930571 on Build GRCh38
rs754687442 Chr.3: 125930537-125930537 on Build GRCh38
rs956483647 Chr.3: 125930410-125930410 on Build GRCh38
rs1003735437 Chr.3: 125930395-125930395 on Build GRCh38
rs760235282 Chr.3: 125930522-125930522 on Build GRCh38
rs7616044 Chr.3: 125930511-125930511 on Build GRCh38
rs370869581 Chr.3: 125930700-125930701 on Build GRCh38
rs752308735 Chr.3: 125930604-125930604 on Build GRCh38
rs772981071 Chr.3: 125930519-125930519 on Build GRCh38
rs770819855 Chr.3: 125930643-125930643 on Build GRCh38
rs989672472 Chr.3: 125930411-125930411 on Build GRCh38
rs750788815 Chr.3: 125930611-125930611 on Build GRCh38
rs751974678 Chr.3: 125930541-125930541 on Build GRCh38
rs779279829 Chr.3: 125930634-125930634 on Build GRCh38
rs567807193 Chr.3: 125930585-125930585 on Build GRCh38

View Full Product Details